Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 186838553-186839097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM2
Entrez Gene ID
151112
Gene Name
zinc finger SWIM-type containing 2
Gene Aliases
MEX, ZZZ2
UniGene
Hs.375054
Gene Chromosome Location
Chr.2: 186827480-186849170 on Build GRCh38
Transcripts
NM_182521.2 XM_011510712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5683699 c.439T>C p.C147R Chr.2: 186839014-186839014 on Build GRCh38
6245651 c.358C>A p.Q120K Chr.2: 186839095-186839095 on Build GRCh38
4404696 c.356C>T p.P119L Chr.2: 186839097-186839097 on Build GRCh38
3838118 c.414G>A p.Q138Q Chr.2: 186839039-186839039 on Build GRCh38
7135352 c.451C>A p.Q151K Chr.2: 186839002-186839002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891503793 Chr.2: 186838712-186838712 on Build GRCh38
rs575335909 Chr.2: 186838862-186838862 on Build GRCh38
rs767657931 Chr.2: 186838928-186838929 on Build GRCh38
rs1054576496 Chr.2: 186839011-186839011 on Build GRCh38
rs889563287 Chr.2: 186838690-186838690 on Build GRCh38
rs574500556 Chr.2: 186838817-186838817 on Build GRCh38
rs778497427 Chr.2: 186839052-186839052 on Build GRCh38
rs1021576743 Chr.2: 186838786-186838786 on Build GRCh38
rs550818855 Chr.2: 186838600-186838600 on Build GRCh38
rs545433328 Chr.2: 186838776-186838777 on Build GRCh38
rs908079143 Chr.2: 186838657-186838657 on Build GRCh38
rs960283568 Chr.2: 186838629-186838629 on Build GRCh38
rs769736941 Chr.2: 186838975-186838975 on Build GRCh38
rs545795528 Chr.2: 186838894-186838894 on Build GRCh38
rs980576188 Chr.2: 186838606-186838606 on Build GRCh38
rs992861374 Chr.2: 186838653-186838653 on Build GRCh38
rs553927408 Chr.2: 186838662-186838662 on Build GRCh38
rs758545231 Chr.2: 186838958-186838958 on Build GRCh38
rs746103464 Chr.2: 186838955-186838955 on Build GRCh38
rs758948190 Chr.2: 186839028-186839028 on Build GRCh38
rs907872529 Chr.2: 186838938-186838938 on Build GRCh38
rs1041232779 Chr.2: 186839062-186839062 on Build GRCh38
rs557996731 Chr.2: 186838678-186838678 on Build GRCh38
rs754363934 Chr.2: 186839033-186839033 on Build GRCh38
rs773971312 Chr.2: 186839019-186839019 on Build GRCh38
rs1010154892 Chr.2: 186838784-186838784 on Build GRCh38
rs534555562 Chr.2: 186838703-186838703 on Build GRCh38
rs757647707 Chr.2: 186838949-186838949 on Build GRCh38
rs1015654305 Chr.2: 186838801-186838801 on Build GRCh38
rs780243395 Chr.2: 186838959-186838959 on Build GRCh38
rs139345536 Chr.2: 186839008-186839008 on Build GRCh38
rs557259354 Chr.2: 186838852-186838852 on Build GRCh38
rs201322180 Chr.2: 186838688-186838689 on Build GRCh38
rs930531500 Chr.2: 186838685-186838685 on Build GRCh38
rs183673901 Chr.2: 186838658-186838658 on Build GRCh38
rs981935056 Chr.2: 186838799-186838800 on Build GRCh38
rs933816958 Chr.2: 186838804-186838825 on Build GRCh38
rs971927522 Chr.2: 186838643-186838643 on Build GRCh38
rs993203997 Chr.2: 186838853-186838853 on Build GRCh38
rs1040546002 Chr.2: 186838923-186838923 on Build GRCh38
rs144251004 Chr.2: 186839055-186839055 on Build GRCh38
rs754268070 Chr.2: 186838927-186838927 on Build GRCh38
rs748061279 Chr.2: 186838973-186838973 on Build GRCh38
rs113939717 Chr.2: 186838699-186838699 on Build GRCh38
rs542156548 Chr.2: 186838823-186838823 on Build GRCh38
rs773088669 Chr.2: 186839005-186839005 on Build GRCh38
rs768685520 Chr.2: 186838965-186838965 on Build GRCh38
rs553130708 Chr.2: 186838809-186838809 on Build GRCh38
rs1040091375 Chr.2: 186838704-186838705 on Build GRCh38
rs528183142 Chr.2: 186838964-186838964 on Build GRCh38
rs1013104415 Chr.2: 186838767-186838771 on Build GRCh38
rs868557830 Chr.2: 186839065-186839065 on Build GRCh38
rs920872606 Chr.2: 186838968-186838968 on Build GRCh38
rs957647745 Chr.2: 186838764-186838766 on Build GRCh38
rs757702862 Chr.2: 186839050-186839050 on Build GRCh38
rs1023421789 Chr.2: 186838791-186838792 on Build GRCh38
rs557740150 Chr.2: 186838905-186838905 on Build GRCh38
rs7581809 Chr.2: 186838665-186838665 on Build GRCh38
rs986060494 Chr.2: 186838825-186838825 on Build GRCh38
rs998956351 Chr.2: 186838721-186838724 on Build GRCh38
rs962057061 Chr.2: 186838953-186838953 on Build GRCh38
rs779198876 Chr.2: 186838954-186838954 on Build GRCh38
rs17852918 Chr.2: 186839075-186839075 on Build GRCh38
rs576352242 Chr.2: 186838743-186838745 on Build GRCh38
rs944350758 Chr.2: 186838897-186838897 on Build GRCh38
rs764945869 Chr.2: 186838650-186838651 on Build GRCh38
rs756713183 Chr.2: 186838924-186838924 on Build GRCh38
rs932112551 Chr.2: 186839018-186839018 on Build GRCh38
rs927429708 Chr.2: 186838619-186838619 on Build GRCh38
rs566940644 Chr.2: 186838691-186838691 on Build GRCh38
rs564064509 Chr.2: 186838915-186838915 on Build GRCh38
rs920758729 Chr.2: 186838664-186838664 on Build GRCh38
rs147735155 Chr.2: 186839029-186839029 on Build GRCh38
rs764498248 Chr.2: 186838926-186838926 on Build GRCh38
rs765944471 Chr.2: 186839016-186839016 on Build GRCh38
rs746358178 Chr.2: 186838925-186838925 on Build GRCh38
rs903366151 Chr.2: 186838717-186838717 on Build GRCh38
rs1033238757 Chr.2: 186838762-186838766 on Build GRCh38
rs971388931 Chr.2: 186838792-186838793 on Build GRCh38
rs752855462 Chr.2: 186839031-186839032 on Build GRCh38
rs766000917 Chr.2: 186838914-186838914 on Build GRCh38
rs776404112 Chr.2: 186838967-186838967 on Build GRCh38

View Full Product Details