Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78275608-78276109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA4
Entrez Gene ID
55466
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Gene Aliases
MST104, MSTP104, PRO1472
UniGene
Hs.513053
Gene Chromosome Location
Chr.15: 78264145-78282196 on Build GRCh38
Transcripts
NM_001130182.1 NM_001130183.1 NM_018602.3 XM_005254535.3 XM_006720601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
965343 c.859G>A p.V287I Chr.15: 78275710-78275710 on Build GRCh38
3504293 c.699C>T p.I233I Chr.15: 78275631-78275631 on Build GRCh38
4678649 c.831G>A p.T277T Chr.15: 78275682-78275682 on Build GRCh38
3816822 c.877+2T>C p.? Chr.15: 78275730-78275730 on Build GRCh38
965341 c.851G>A p.R284Q Chr.15: 78275702-78275702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569472097 Chr.15: 78275815-78275815 on Build GRCh38
rs776120715 Chr.15: 78275751-78275751 on Build GRCh38
rs775828560 Chr.15: 78275641-78275641 on Build GRCh38
rs927320535 Chr.15: 78276022-78276022 on Build GRCh38
rs904380792 Chr.15: 78276077-78276077 on Build GRCh38
rs965032777 Chr.15: 78275888-78275888 on Build GRCh38
rs1044485931 Chr.15: 78275718-78275718 on Build GRCh38
rs757504374 Chr.15: 78275710-78275710 on Build GRCh38
rs879719803 Chr.15: 78275893-78275893 on Build GRCh38
rs781515229 Chr.15: 78275744-78275745 on Build GRCh38
rs369601244 Chr.15: 78275713-78275713 on Build GRCh38
rs751859584 Chr.15: 78275702-78275702 on Build GRCh38
rs752694531 Chr.15: 78275761-78275761 on Build GRCh38
rs763264121 Chr.15: 78275734-78275735 on Build GRCh38
rs942115381 Chr.15: 78275708-78275708 on Build GRCh38
rs777655450 Chr.15: 78275701-78275701 on Build GRCh38
rs377270562 Chr.15: 78275745-78275745 on Build GRCh38
rs753342413 Chr.15: 78275669-78275669 on Build GRCh38
rs769010222 Chr.15: 78275739-78275739 on Build GRCh38
rs767180015 Chr.15: 78275815-78275817 on Build GRCh38
rs184860035 Chr.15: 78275863-78275863 on Build GRCh38
rs780664664 Chr.15: 78275726-78275726 on Build GRCh38
rs770623271 Chr.15: 78275746-78275746 on Build GRCh38
rs555443695 Chr.15: 78276053-78276053 on Build GRCh38
rs781248827 Chr.15: 78275712-78275712 on Build GRCh38
rs758326761 Chr.15: 78275699-78275699 on Build GRCh38
rs985208054 Chr.15: 78275827-78275827 on Build GRCh38
rs760251066 Chr.15: 78275658-78275658 on Build GRCh38
rs377119803 Chr.15: 78275714-78275714 on Build GRCh38
rs1057165604 Chr.15: 78276029-78276029 on Build GRCh38
rs532308692 Chr.15: 78276028-78276028 on Build GRCh38
rs201235137 Chr.15: 78275887-78275887 on Build GRCh38
rs750670908 Chr.15: 78275657-78275658 on Build GRCh38
rs751951146 Chr.15: 78275739-78275740 on Build GRCh38
rs749734650 Chr.15: 78275733-78275733 on Build GRCh38
rs78756785 Chr.15: 78275707-78275707 on Build GRCh38
rs905876057 Chr.15: 78275740-78275740 on Build GRCh38
rs1006657161 Chr.15: 78276078-78276078 on Build GRCh38
rs1017953458 Chr.15: 78275831-78275831 on Build GRCh38
rs372092147 Chr.15: 78275693-78275693 on Build GRCh38
rs753228318 Chr.15: 78275906-78275906 on Build GRCh38
rs755569100 Chr.15: 78275743-78275743 on Build GRCh38
rs563571693 Chr.15: 78275753-78275758 on Build GRCh38
rs1021454573 Chr.15: 78275813-78275813 on Build GRCh38
rs776756103 Chr.15: 78275648-78275648 on Build GRCh38
rs1054518973 Chr.15: 78275981-78275981 on Build GRCh38
rs148370822 Chr.15: 78275682-78275682 on Build GRCh38
rs1010560185 Chr.15: 78275799-78275799 on Build GRCh38
rs950329169 Chr.15: 78275875-78275875 on Build GRCh38
rs890992197 Chr.15: 78276079-78276079 on Build GRCh38
rs368143683 Chr.15: 78275650-78275650 on Build GRCh38
rs765263538 Chr.15: 78275681-78275681 on Build GRCh38
rs891787574 Chr.15: 78276038-78276038 on Build GRCh38
rs765663586 Chr.15: 78275880-78275880 on Build GRCh38
rs573284331 Chr.15: 78275942-78275942 on Build GRCh38
rs765907325 Chr.15: 78275661-78275661 on Build GRCh38
rs983646442 Chr.15: 78275847-78275847 on Build GRCh38
rs753068873 Chr.15: 78275774-78275776 on Build GRCh38
rs963715518 Chr.15: 78275798-78275798 on Build GRCh38
rs951031827 Chr.15: 78275923-78275923 on Build GRCh38
rs908082248 Chr.15: 78275862-78275862 on Build GRCh38
rs764641416 Chr.15: 78275755-78275755 on Build GRCh38
rs142259465 Chr.15: 78275741-78275741 on Build GRCh38
rs758956962 Chr.15: 78275988-78275988 on Build GRCh38
rs764000890 Chr.15: 78275774-78275774 on Build GRCh38
rs909408951 Chr.15: 78275954-78275954 on Build GRCh38
rs545943043 Chr.15: 78276048-78276048 on Build GRCh38
rs373981227 Chr.15: 78275742-78275742 on Build GRCh38
rs189185776 Chr.15: 78275940-78275940 on Build GRCh38
rs371732302 Chr.15: 78275753-78275753 on Build GRCh38
rs976407312 Chr.15: 78275901-78275901 on Build GRCh38
rs766891817 Chr.15: 78275735-78275736 on Build GRCh38
rs193218284 Chr.15: 78275674-78275674 on Build GRCh38
rs534325737 Chr.15: 78276016-78276016 on Build GRCh38
rs780592289 Chr.15: 78275849-78275849 on Build GRCh38
rs887941803 Chr.15: 78275705-78275705 on Build GRCh38
rs762919789 Chr.15: 78275773-78275773 on Build GRCh38

View Full Product Details