Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122187307-122187873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC43
Entrez Gene ID
254050
Gene Name
leucine rich repeat containing 43
Gene Aliases
-
UniGene
Hs.374856
Gene Chromosome Location
Chr.12: 122167719-122203471 on Build GRCh38
Transcripts
NM_001098519.1 NM_152759.4 XM_017019124.1 XM_017019125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1359564 c.537C>T p.Y179Y Chr.12: 122187715-122187715 on Build GRCh38
5042580 c.73C>T p.P25S Chr.12: 122187806-122187806 on Build GRCh38
7375602 c.652G>A p.A218T Chr.12: 122187830-122187830 on Build GRCh38
5135895 c.538G>A p.G180S Chr.12: 122187716-122187716 on Build GRCh38
2157024 c.650C>T p.T217I Chr.12: 122187828-122187828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916253769 Chr.12: 122187433-122187433 on Build GRCh38
rs530340667 Chr.12: 122187565-122187565 on Build GRCh38
rs768033305 Chr.12: 122187848-122187848 on Build GRCh38
rs202034748 Chr.12: 122187677-122187677 on Build GRCh38
rs773196895 Chr.12: 122187566-122187566 on Build GRCh38
rs994375102 Chr.12: 122187345-122187345 on Build GRCh38
rs761495780 Chr.12: 122187337-122187337 on Build GRCh38
rs747099274 Chr.12: 122187756-122187756 on Build GRCh38
rs768353118 Chr.12: 122187656-122187656 on Build GRCh38
rs550705478 Chr.12: 122187506-122187506 on Build GRCh38
rs764831908 Chr.12: 122187603-122187603 on Build GRCh38
rs374452899 Chr.12: 122187748-122187748 on Build GRCh38
rs942058734 Chr.12: 122187798-122187798 on Build GRCh38
rs74941994 Chr.12: 122187401-122187401 on Build GRCh38
rs779608980 Chr.12: 122187651-122187651 on Build GRCh38
rs963742738 Chr.12: 122187402-122187402 on Build GRCh38
rs770928602 Chr.12: 122187758-122187758 on Build GRCh38
rs749959009 Chr.12: 122187621-122187621 on Build GRCh38
rs149000870 Chr.12: 122187847-122187847 on Build GRCh38
rs371086435 Chr.12: 122187743-122187743 on Build GRCh38
rs562121684 Chr.12: 122187593-122187593 on Build GRCh38
rs181503685 Chr.12: 122187624-122187624 on Build GRCh38
rs762312410 Chr.12: 122187668-122187668 on Build GRCh38
rs765842778 Chr.12: 122187779-122187779 on Build GRCh38
rs947480237 Chr.12: 122187573-122187573 on Build GRCh38
rs114611063 Chr.12: 122187830-122187830 on Build GRCh38
rs548143956 Chr.12: 122187491-122187491 on Build GRCh38
rs777796530 Chr.12: 122187824-122187824 on Build GRCh38
rs778299089 Chr.12: 122187697-122187697 on Build GRCh38
rs746924097 Chr.12: 122187826-122187826 on Build GRCh38
rs778875166 Chr.12: 122187730-122187730 on Build GRCh38
rs758326623 Chr.12: 122187823-122187823 on Build GRCh38
rs771002440 Chr.12: 122187829-122187829 on Build GRCh38
rs909204076 Chr.12: 122187790-122187790 on Build GRCh38
rs749899890 Chr.12: 122187701-122187701 on Build GRCh38
rs772231630 Chr.12: 122187673-122187673 on Build GRCh38
rs753483392 Chr.12: 122187787-122187787 on Build GRCh38
rs1040274400 Chr.12: 122187535-122187535 on Build GRCh38
rs1030781212 Chr.12: 122187740-122187740 on Build GRCh38
rs983834640 Chr.12: 122187389-122187389 on Build GRCh38
rs776844842 Chr.12: 122187760-122187760 on Build GRCh38
rs765920622 Chr.12: 122187672-122187672 on Build GRCh38
rs145831421 Chr.12: 122187769-122187769 on Build GRCh38
rs896082237 Chr.12: 122187633-122187633 on Build GRCh38
rs371628171 Chr.12: 122187848-122187848 on Build GRCh38
rs967748450 Chr.12: 122187465-122187466 on Build GRCh38
rs764653926 Chr.12: 122187744-122187745 on Build GRCh38
rs374469863 Chr.12: 122187678-122187678 on Build GRCh38
rs923742999 Chr.12: 122187498-122187498 on Build GRCh38
rs149822167 Chr.12: 122187365-122187365 on Build GRCh38
rs763287758 Chr.12: 122187849-122187849 on Build GRCh38
rs952458885 Chr.12: 122187385-122187385 on Build GRCh38
rs1050824092 Chr.12: 122187623-122187623 on Build GRCh38
rs566749536 Chr.12: 122187670-122187670 on Build GRCh38
rs749100969 Chr.12: 122187653-122187653 on Build GRCh38
rs779702868 Chr.12: 122187716-122187716 on Build GRCh38
rs765586541 Chr.12: 122187761-122187761 on Build GRCh38
rs775753716 Chr.12: 122187833-122187833 on Build GRCh38
rs765523628 Chr.12: 122187686-122187686 on Build GRCh38
rs761342873 Chr.12: 122187682-122187695 on Build GRCh38
rs747954740 Chr.12: 122187821-122187821 on Build GRCh38
rs368344594 Chr.12: 122187764-122187764 on Build GRCh38
rs754766273 Chr.12: 122187727-122187727 on Build GRCh38
rs979196973 Chr.12: 122187487-122187488 on Build GRCh38
rs764576552 Chr.12: 122187766-122187766 on Build GRCh38
rs755324432 Chr.12: 122187847-122187848 on Build GRCh38
rs776455413 Chr.12: 122187665-122187666 on Build GRCh38
rs774282480 Chr.12: 122187669-122187669 on Build GRCh38
rs773406086 Chr.12: 122187676-122187676 on Build GRCh38
rs990924442 Chr.12: 122187407-122187407 on Build GRCh38
rs772104845 Chr.12: 122187739-122187739 on Build GRCh38
rs753161019 Chr.12: 122187687-122187687 on Build GRCh38
rs569502869 Chr.12: 122187545-122187545 on Build GRCh38
rs754567305 Chr.12: 122187808-122187808 on Build GRCh38
rs748007760 Chr.12: 122187733-122187733 on Build GRCh38
rs755588187 Chr.12: 122187775-122187775 on Build GRCh38
rs750183053 Chr.12: 122187754-122187754 on Build GRCh38
rs565270953 Chr.12: 122187467-122187467 on Build GRCh38
rs762678375 Chr.12: 122187811-122187816 on Build GRCh38
rs1023701484 Chr.12: 122187696-122187696 on Build GRCh38
rs200354949 Chr.12: 122187443-122187443 on Build GRCh38
rs61745357 Chr.12: 122187715-122187715 on Build GRCh38
rs923222154 Chr.12: 122187560-122187560 on Build GRCh38
rs199935953 Chr.12: 122187845-122187852 on Build GRCh38
rs556698413 Chr.12: 122187763-122187763 on Build GRCh38
rs778750097 Chr.12: 122187818-122187818 on Build GRCh38
rs766176761 Chr.12: 122187817-122187818 on Build GRCh38
rs748900768 Chr.12: 122187720-122187720 on Build GRCh38
rs753149618 Chr.12: 122187848-122187849 on Build GRCh38

View Full Product Details