Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30753095-30753683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHKG2
Entrez Gene ID
5261
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Gene Aliases
GSD9C
UniGene
Hs.65735
Gene Chromosome Location
Chr.16: 30748299-30761176 on Build GRCh38
Transcripts
NM_000294.2 NM_001172432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479101 c.471G>T p.E157D Chr.16: 30753472-30753472 on Build GRCh38
1377490 c.455G>A p.R152Q Chr.16: 30753456-30753456 on Build GRCh38
6451383 c.551T>C p.L184P Chr.16: 30753552-30753552 on Build GRCh38
4932081 c.327-2A>T p.? Chr.16: 30753230-30753230 on Build GRCh38
1301888 c.340G>A p.E114K Chr.16: 30753245-30753245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774298280 Chr.16: 30753322-30753322 on Build GRCh38
rs777689254 Chr.16: 30753310-30753310 on Build GRCh38
rs759361949 Chr.16: 30753247-30753247 on Build GRCh38
rs61733974 Chr.16: 30753555-30753555 on Build GRCh38
rs755646444 Chr.16: 30753161-30753161 on Build GRCh38
rs370423261 Chr.16: 30753258-30753258 on Build GRCh38
rs762578361 Chr.16: 30753540-30753540 on Build GRCh38
rs778952896 Chr.16: 30753503-30753503 on Build GRCh38
rs201964148 Chr.16: 30753359-30753359 on Build GRCh38
rs748159574 Chr.16: 30753307-30753308 on Build GRCh38
rs752961445 Chr.16: 30753470-30753470 on Build GRCh38
rs535319513 Chr.16: 30753362-30753362 on Build GRCh38
rs772912966 Chr.16: 30753455-30753455 on Build GRCh38
rs756255612 Chr.16: 30753373-30753373 on Build GRCh38
rs932084569 Chr.16: 30753493-30753493 on Build GRCh38
rs555472522 Chr.16: 30753276-30753276 on Build GRCh38
rs771861833 Chr.16: 30753325-30753325 on Build GRCh38
rs137853591 Chr.16: 30753434-30753434 on Build GRCh38
rs759012661 Chr.16: 30753173-30753173 on Build GRCh38
rs545582783 Chr.16: 30753492-30753492 on Build GRCh38
rs757490714 Chr.16: 30753498-30753498 on Build GRCh38
rs368904522 Chr.16: 30753536-30753536 on Build GRCh38
rs768238080 Chr.16: 30753284-30753285 on Build GRCh38
rs149163610 Chr.16: 30753265-30753265 on Build GRCh38
rs769766375 Chr.16: 30753194-30753194 on Build GRCh38
rs745848943 Chr.16: 30753182-30753182 on Build GRCh38
rs760885327 Chr.16: 30753547-30753547 on Build GRCh38
rs55751949 Chr.16: 30753394-30753394 on Build GRCh38
rs779998350 Chr.16: 30753518-30753518 on Build GRCh38
rs45612445 Chr.16: 30753530-30753530 on Build GRCh38
rs543453602 Chr.16: 30753573-30753573 on Build GRCh38
rs780269438 Chr.16: 30753175-30753175 on Build GRCh38
rs778083432 Chr.16: 30753376-30753376 on Build GRCh38
rs769454501 Chr.16: 30753343-30753344 on Build GRCh38
rs764834643 Chr.16: 30753441-30753441 on Build GRCh38
rs758496261 Chr.16: 30753588-30753588 on Build GRCh38
rs369684098 Chr.16: 30753562-30753562 on Build GRCh38
rs746449699 Chr.16: 30753126-30753129 on Build GRCh38
rs756414222 Chr.16: 30753309-30753309 on Build GRCh38
rs751510713 Chr.16: 30753598-30753598 on Build GRCh38
rs770738163 Chr.16: 30753319-30753319 on Build GRCh38
rs761161501 Chr.16: 30753433-30753433 on Build GRCh38
rs748373892 Chr.16: 30753303-30753303 on Build GRCh38
rs919599212 Chr.16: 30753356-30753356 on Build GRCh38
rs372239376 Chr.16: 30753544-30753544 on Build GRCh38
rs767112052 Chr.16: 30753253-30753253 on Build GRCh38
rs201959068 Chr.16: 30753582-30753582 on Build GRCh38
rs758301470 Chr.16: 30753517-30753517 on Build GRCh38
rs1020156539 Chr.16: 30753237-30753237 on Build GRCh38
rs371900227 Chr.16: 30753372-30753372 on Build GRCh38
rs1011510687 Chr.16: 30753612-30753612 on Build GRCh38
rs766949870 Chr.16: 30753266-30753266 on Build GRCh38
rs776394065 Chr.16: 30753296-30753300 on Build GRCh38
rs985964135 Chr.16: 30753205-30753205 on Build GRCh38
rs374131174 Chr.16: 30753334-30753334 on Build GRCh38
rs1042747324 Chr.16: 30753282-30753282 on Build GRCh38
rs201659307 Chr.16: 30753289-30753289 on Build GRCh38
rs747427432 Chr.16: 30753180-30753180 on Build GRCh38
rs375764924 Chr.16: 30753423-30753423 on Build GRCh38
rs559193568 Chr.16: 30753618-30753618 on Build GRCh38
rs540905727 Chr.16: 30753608-30753608 on Build GRCh38
rs750553595 Chr.16: 30753509-30753509 on Build GRCh38
rs923206955 Chr.16: 30753178-30753178 on Build GRCh38
rs762002537 Chr.16: 30753565-30753565 on Build GRCh38
rs886051911 Chr.16: 30753449-30753449 on Build GRCh38
rs768128006 Chr.16: 30753424-30753424 on Build GRCh38
rs768174256 Chr.16: 30753469-30753469 on Build GRCh38
rs752165708 Chr.16: 30753159-30753159 on Build GRCh38
rs886051912 Chr.16: 30753562-30753563 on Build GRCh38
rs749420596 Chr.16: 30753318-30753318 on Build GRCh38
rs1028989522 Chr.16: 30753124-30753124 on Build GRCh38
rs986429508 Chr.16: 30753591-30753591 on Build GRCh38
rs775973433 Chr.16: 30753339-30753339 on Build GRCh38
rs760096451 Chr.16: 30753459-30753459 on Build GRCh38
rs773982918 Chr.16: 30753236-30753236 on Build GRCh38
rs768501872 Chr.16: 30753520-30753520 on Build GRCh38
rs148036324 Chr.16: 30753404-30753404 on Build GRCh38
rs978598416 Chr.16: 30753660-30753660 on Build GRCh38
rs775332699 Chr.16: 30753333-30753333 on Build GRCh38
rs776358711 Chr.16: 30753426-30753426 on Build GRCh38
rs779420230 Chr.16: 30753162-30753162 on Build GRCh38
rs752115033 Chr.16: 30753358-30753358 on Build GRCh38
rs768958688 Chr.16: 30753181-30753181 on Build GRCh38
rs762783981 Chr.16: 30753226-30753226 on Build GRCh38
rs745607814 Chr.16: 30753323-30753323 on Build GRCh38
rs759161792 Chr.16: 30753152-30753152 on Build GRCh38
rs773199410 Chr.16: 30753472-30753472 on Build GRCh38
rs765485630 Chr.16: 30753571-30753571 on Build GRCh38
rs755884169 Chr.16: 30753221-30753221 on Build GRCh38
rs757383260 Chr.16: 30753157-30753157 on Build GRCh38
rs761408369 Chr.16: 30753341-30753341 on Build GRCh38
rs774068311 Chr.16: 30753442-30753442 on Build GRCh38
rs749282755 Chr.16: 30753377-30753377 on Build GRCh38
rs767202097 Chr.16: 30753154-30753154 on Build GRCh38
rs752039795 Chr.16: 30753215-30753215 on Build GRCh38
rs767789467 Chr.16: 30753559-30753560 on Build GRCh38
rs34885557 Chr.16: 30753232-30753232 on Build GRCh38
rs377761632 Chr.16: 30753580-30753580 on Build GRCh38
rs762844937 Chr.16: 30753523-30753525 on Build GRCh38
rs139116127 Chr.16: 30753531-30753531 on Build GRCh38
rs764516590 Chr.16: 30753560-30753560 on Build GRCh38
rs138740083 Chr.16: 30753490-30753490 on Build GRCh38
rs773405423 Chr.16: 30753202-30753202 on Build GRCh38
rs780913532 Chr.16: 30753524-30753524 on Build GRCh38
rs146181866 Chr.16: 30753496-30753496 on Build GRCh38
rs537463666 Chr.16: 30753165-30753165 on Build GRCh38
rs775246080 Chr.16: 30753405-30753405 on Build GRCh38
rs191368660 Chr.16: 30753594-30753594 on Build GRCh38
rs766736830 Chr.16: 30753354-30753354 on Build GRCh38
rs752363595 Chr.16: 30753254-30753254 on Build GRCh38
rs763509347 Chr.16: 30753262-30753262 on Build GRCh38
rs770947204 Chr.16: 30753231-30753231 on Build GRCh38
rs578228524 Chr.16: 30753401-30753401 on Build GRCh38
rs767977936 Chr.16: 30753364-30753364 on Build GRCh38
rs748179831 Chr.16: 30753189-30753189 on Build GRCh38
rs754926114 Chr.16: 30753603-30753603 on Build GRCh38
rs967173442 Chr.16: 30753241-30753241 on Build GRCh38
rs775875751 Chr.16: 30753567-30753568 on Build GRCh38

View Full Product Details