Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7861322-7861911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5D1
Entrez Gene ID
124637
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
-
UniGene
Hs.27475
Gene Chromosome Location
Chr.17: 7857746-7864417 on Build GRCh38
Transcripts
NM_144607.4 XM_005256440.3 XM_011523638.2 XM_011524037.2 XM_017025225.1 XM_017025226.1

Gene Symbol
NAA38
Entrez Gene ID
84316
Gene Name
N(alpha)-acetyltransferase 38, NatC auxiliary subunit
Gene Aliases
LSMD1, PFAAP2
UniGene
Hs.565094
Gene Chromosome Location
Chr.17: 7856685-7885465 on Build GRCh38
Transcripts
NM_144607.4 XM_005256440.3 XM_011523638.2 XM_011524037.2 XM_017025225.1 XM_017025226.1

SNP Detail

SNP ID Chromosome Location
rs761670695 Chr.17: 7861483-7861483 on Build GRCh38
rs577632170 Chr.17: 7861706-7861706 on Build GRCh38
rs562689072 Chr.17: 7861414-7861418 on Build GRCh38
rs564950677 Chr.17: 7861410-7861412 on Build GRCh38
rs895968258 Chr.17: 7861718-7861718 on Build GRCh38
rs774180971 Chr.17: 7861412-7861418 on Build GRCh38
rs1017243566 Chr.17: 7861514-7861514 on Build GRCh38
rs933769104 Chr.17: 7861598-7861598 on Build GRCh38
rs567648960 Chr.17: 7861844-7861844 on Build GRCh38
rs763860159 Chr.17: 7861410-7861410 on Build GRCh38
rs944143182 Chr.17: 7861662-7861662 on Build GRCh38
rs1025085276 Chr.17: 7861408-7861408 on Build GRCh38
rs1025666432 Chr.17: 7861451-7861451 on Build GRCh38
rs1055926269 Chr.17: 7861714-7861714 on Build GRCh38
rs1024697179 Chr.17: 7861829-7861829 on Build GRCh38
rs76080656 Chr.17: 7861636-7861636 on Build GRCh38
rs1029526974 Chr.17: 7861816-7861816 on Build GRCh38
rs984070346 Chr.17: 7861509-7861509 on Build GRCh38
rs372344502 Chr.17: 7861410-7861417 on Build GRCh38
rs1014287376 Chr.17: 7861419-7861419 on Build GRCh38
rs531757414 Chr.17: 7861880-7861880 on Build GRCh38
rs937585325 Chr.17: 7861714-7861715 on Build GRCh38
rs565915413 Chr.17: 7861363-7861363 on Build GRCh38
rs951444109 Chr.17: 7861481-7861481 on Build GRCh38
rs888663302 Chr.17: 7861792-7861792 on Build GRCh38
rs980696139 Chr.17: 7861464-7861464 on Build GRCh38
rs1013332216 Chr.17: 7861404-7861404 on Build GRCh38
rs957866309 Chr.17: 7861497-7861497 on Build GRCh38
rs963877367 Chr.17: 7861521-7861521 on Build GRCh38
rs913580239 Chr.17: 7861664-7861664 on Build GRCh38
rs537401954 Chr.17: 7861631-7861631 on Build GRCh38
rs976852268 Chr.17: 7861682-7861682 on Build GRCh38
rs988414763 Chr.17: 7861505-7861505 on Build GRCh38
rs573313512 Chr.17: 7861590-7861590 on Build GRCh38
rs1034404283 Chr.17: 7861415-7861415 on Build GRCh38
rs932632585 Chr.17: 7861771-7861771 on Build GRCh38
rs969029674 Chr.17: 7861418-7861418 on Build GRCh38
rs1026122319 Chr.17: 7861729-7861729 on Build GRCh38
rs962134407 Chr.17: 7861384-7861384 on Build GRCh38
rs749967212 Chr.17: 7861809-7861809 on Build GRCh38
rs755296263 Chr.17: 7861752-7861752 on Build GRCh38
rs1032235644 Chr.17: 7861489-7861489 on Build GRCh38
rs758697132 Chr.17: 7861350-7861350 on Build GRCh38
rs572259977 Chr.17: 7861815-7861815 on Build GRCh38
rs191037211 Chr.17: 7861343-7861343 on Build GRCh38
rs542951884 Chr.17: 7861822-7861822 on Build GRCh38
rs755572976 Chr.17: 7861882-7861882 on Build GRCh38
rs549151230 Chr.17: 7861366-7861366 on Build GRCh38
rs554835999 Chr.17: 7861527-7861527 on Build GRCh38
rs887135980 Chr.17: 7861746-7861746 on Build GRCh38
rs79533239 Chr.17: 7861813-7861813 on Build GRCh38
rs147577981 Chr.17: 7861825-7861825 on Build GRCh38
rs1014368269 Chr.17: 7861728-7861728 on Build GRCh38
rs921279818 Chr.17: 7861698-7861698 on Build GRCh38
rs940267092 Chr.17: 7861806-7861806 on Build GRCh38
rs536597874 Chr.17: 7861526-7861526 on Build GRCh38
rs145133723 Chr.17: 7861785-7861785 on Build GRCh38
rs751860151 Chr.17: 7861583-7861583 on Build GRCh38

View Full Product Details