Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59154835-59155387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKA2
Entrez Gene ID
348235
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
FAM33A
UniGene
Hs.463607
Gene Chromosome Location
Chr.17: 59109947-59155439 on Build GRCh38
Transcripts
NM_001100595.1 NM_182620.3 XM_011524745.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338492 c.69C>A p.V23V Chr.17: 59155015-59155015 on Build GRCh38
3958599 c.30G>T p.E10D Chr.17: 59155054-59155054 on Build GRCh38
7120900 c.69C>G p.V23V Chr.17: 59155015-59155015 on Build GRCh38
5048441 c.71A>T p.E24V Chr.17: 59155013-59155013 on Build GRCh38
7319088 c.61A>T p.T21S Chr.17: 59155023-59155023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200500858 Chr.17: 59154989-59154989 on Build GRCh38
rs551507658 Chr.17: 59154897-59154897 on Build GRCh38
rs770429469 Chr.17: 59154927-59154927 on Build GRCh38
rs533123424 Chr.17: 59154857-59154857 on Build GRCh38
rs759733194 Chr.17: 59155185-59155185 on Build GRCh38
rs778187675 Chr.17: 59155113-59155113 on Build GRCh38
rs754182359 Chr.17: 59155190-59155190 on Build GRCh38
rs979549377 Chr.17: 59154880-59154880 on Build GRCh38
rs910293077 Chr.17: 59155000-59155000 on Build GRCh38
rs747758423 Chr.17: 59155131-59155131 on Build GRCh38
rs781690084 Chr.17: 59154921-59154921 on Build GRCh38
rs1021047670 Chr.17: 59154856-59154856 on Build GRCh38
rs370436807 Chr.17: 59155210-59155210 on Build GRCh38
rs758076885 Chr.17: 59155191-59155191 on Build GRCh38
rs774813820 Chr.17: 59155352-59155352 on Build GRCh38
rs768122388 Chr.17: 59155134-59155134 on Build GRCh38
rs747451516 Chr.17: 59155079-59155080 on Build GRCh38
rs189395759 Chr.17: 59155279-59155279 on Build GRCh38
rs774607925 Chr.17: 59155180-59155180 on Build GRCh38
rs771541476 Chr.17: 59155035-59155035 on Build GRCh38
rs745512955 Chr.17: 59155020-59155020 on Build GRCh38
rs769607242 Chr.17: 59155065-59155065 on Build GRCh38
rs925413321 Chr.17: 59154888-59154888 on Build GRCh38
rs748948409 Chr.17: 59154926-59154926 on Build GRCh38
rs755455739 Chr.17: 59154920-59154920 on Build GRCh38
rs749054743 Chr.17: 59155205-59155205 on Build GRCh38
rs761626619 Chr.17: 59155098-59155101 on Build GRCh38
rs199620882 Chr.17: 59155088-59155088 on Build GRCh38
rs775920420 Chr.17: 59155188-59155188 on Build GRCh38
rs767685059 Chr.17: 59155187-59155187 on Build GRCh38
rs908188184 Chr.17: 59155339-59155339 on Build GRCh38
rs777456292 Chr.17: 59155204-59155204 on Build GRCh38
rs546489176 Chr.17: 59154974-59154974 on Build GRCh38
rs970912174 Chr.17: 59155288-59155288 on Build GRCh38
rs776352377 Chr.17: 59154964-59154964 on Build GRCh38
rs547241095 Chr.17: 59155078-59155078 on Build GRCh38
rs746482031 Chr.17: 59155133-59155133 on Build GRCh38
rs773361645 Chr.17: 59155067-59155067 on Build GRCh38
rs924443473 Chr.17: 59155316-59155316 on Build GRCh38
rs768414988 Chr.17: 59154963-59154963 on Build GRCh38
rs747499200 Chr.17: 59154916-59154916 on Build GRCh38
rs535796522 Chr.17: 59155186-59155186 on Build GRCh38
rs775286657 Chr.17: 59154952-59154952 on Build GRCh38
rs748100480 Chr.17: 59155143-59155143 on Build GRCh38
rs934490457 Chr.17: 59155340-59155340 on Build GRCh38
rs770821255 Chr.17: 59155206-59155206 on Build GRCh38
rs762119090 Chr.17: 59154978-59154978 on Build GRCh38
rs763944292 Chr.17: 59154963-59154963 on Build GRCh38
rs752174353 Chr.17: 59154993-59154993 on Build GRCh38
rs199522186 Chr.17: 59154946-59154946 on Build GRCh38
rs374246041 Chr.17: 59155165-59155165 on Build GRCh38
rs760145872 Chr.17: 59155079-59155079 on Build GRCh38
rs754538131 Chr.17: 59155198-59155198 on Build GRCh38
rs763229439 Chr.17: 59155071-59155071 on Build GRCh38
rs368214976 Chr.17: 59155247-59155247 on Build GRCh38
rs776360470 Chr.17: 59155000-59155002 on Build GRCh38
rs758460273 Chr.17: 59154991-59154991 on Build GRCh38
rs750629863 Chr.17: 59154981-59154981 on Build GRCh38
rs552595435 Chr.17: 59155018-59155018 on Build GRCh38
rs776440495 Chr.17: 59155055-59155055 on Build GRCh38
rs765338616 Chr.17: 59154979-59154979 on Build GRCh38
rs377016243 Chr.17: 59154945-59154945 on Build GRCh38
rs983674916 Chr.17: 59155303-59155303 on Build GRCh38
rs918203540 Chr.17: 59154917-59154917 on Build GRCh38
rs1050102876 Chr.17: 59155160-59155160 on Build GRCh38
rs781703711 Chr.17: 59155002-59155002 on Build GRCh38
rs991461044 Chr.17: 59154908-59154908 on Build GRCh38
rs755480328 Chr.17: 59155001-59155001 on Build GRCh38
rs1032451227 Chr.17: 59155216-59155216 on Build GRCh38
rs1023677852 Chr.17: 59155170-59155170 on Build GRCh38
rs774765602 Chr.17: 59155207-59155207 on Build GRCh38
rs781155337 Chr.17: 59155142-59155142 on Build GRCh38
rs77282282 Chr.17: 59155004-59155004 on Build GRCh38
rs769540487 Chr.17: 59155150-59155150 on Build GRCh38
rs756017494 Chr.17: 59155201-59155201 on Build GRCh38
rs757978823 Chr.17: 59155129-59155129 on Build GRCh38
rs766058628 Chr.17: 59155192-59155192 on Build GRCh38
rs771233708 Chr.17: 59155178-59155178 on Build GRCh38
rs572650229 Chr.17: 59155041-59155041 on Build GRCh38
rs952330140 Chr.17: 59155226-59155226 on Build GRCh38
rs569760584 Chr.17: 59154901-59154901 on Build GRCh38
rs985756356 Chr.17: 59155264-59155264 on Build GRCh38
rs760592346 Chr.17: 59154962-59154962 on Build GRCh38
rs780746160 Chr.17: 59155199-59155199 on Build GRCh38
rs779562136 Chr.17: 59155132-59155132 on Build GRCh38
rs775574296 Chr.17: 59155208-59155208 on Build GRCh38
rs746962020 Chr.17: 59155049-59155049 on Build GRCh38
rs750110383 Chr.17: 59155126-59155126 on Build GRCh38
rs903044458 Chr.17: 59155019-59155019 on Build GRCh38
rs376103947 Chr.17: 59155208-59155209 on Build GRCh38
rs768100587 Chr.17: 59155086-59155086 on Build GRCh38
rs761551881 Chr.17: 59155058-59155058 on Build GRCh38
rs959791930 Chr.17: 59155241-59155241 on Build GRCh38
rs775455069 Chr.17: 59154961-59154961 on Build GRCh38
rs778252373 Chr.17: 59155040-59155040 on Build GRCh38
rs116256937 Chr.17: 59155171-59155171 on Build GRCh38
rs998752239 Chr.17: 59155021-59155021 on Build GRCh38
rs760645278 Chr.17: 59154955-59154957 on Build GRCh38
rs536714934 Chr.17: 59154933-59154933 on Build GRCh38
rs764371581 Chr.17: 59155189-59155189 on Build GRCh38
rs1040188833 Chr.17: 59155005-59155005 on Build GRCh38
rs766557649 Chr.17: 59154992-59154992 on Build GRCh38
rs966746330 Chr.17: 59154864-59154864 on Build GRCh38
rs768531629 Chr.17: 59155051-59155051 on Build GRCh38

View Full Product Details