Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 45983408-45983812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSTPIP2
Entrez Gene ID
9050
Gene Name
proline-serine-threonine phosphatase interacting protein 2
Gene Aliases
MAYP
UniGene
Hs.567384
Gene Chromosome Location
Chr.18: 45983536-46072327 on Build GRCh38
Transcripts
NM_024430.3 XM_011526253.2 XM_011526254.1 XM_011526256.1

SNP Detail

SNP ID Chromosome Location
rs1050985561 Chr.18: 45983605-45983605 on Build GRCh38
rs149279561 Chr.18: 45983734-45983734 on Build GRCh38
rs974432089 Chr.18: 45983485-45983485 on Build GRCh38
rs892660221 Chr.18: 45983570-45983570 on Build GRCh38
rs925030787 Chr.18: 45983767-45983767 on Build GRCh38
rs946197155 Chr.18: 45983505-45983505 on Build GRCh38
rs961937636 Chr.18: 45983631-45983631 on Build GRCh38
rs113478050 Chr.18: 45983457-45983457 on Build GRCh38
rs996268174 Chr.18: 45983527-45983527 on Build GRCh38
rs1006646415 Chr.18: 45983614-45983614 on Build GRCh38
rs535363825 Chr.18: 45983528-45983528 on Build GRCh38
rs996530425 Chr.18: 45983708-45983708 on Build GRCh38
rs572308999 Chr.18: 45983579-45983579 on Build GRCh38
rs900061196 Chr.18: 45983632-45983632 on Build GRCh38
rs890778603 Chr.18: 45983726-45983726 on Build GRCh38
rs36023797 Chr.18: 45983740-45983741 on Build GRCh38
rs955081474 Chr.18: 45983433-45983433 on Build GRCh38
rs755741265 Chr.18: 45983784-45983784 on Build GRCh38
rs67746112 Chr.18: 45983711-45983711 on Build GRCh38
rs1014825527 Chr.18: 45983623-45983623 on Build GRCh38
rs1054603785 Chr.18: 45983441-45983441 on Build GRCh38
rs980505508 Chr.18: 45983738-45983738 on Build GRCh38
rs962870884 Chr.18: 45983776-45983776 on Build GRCh38
rs887747126 Chr.18: 45983609-45983609 on Build GRCh38
rs761715245 Chr.18: 45983627-45983627 on Build GRCh38
rs962571805 Chr.18: 45983466-45983466 on Build GRCh38
rs879457222 Chr.18: 45983499-45983499 on Build GRCh38
rs553571465 Chr.18: 45983611-45983611 on Build GRCh38
rs879347768 Chr.18: 45983510-45983510 on Build GRCh38
rs1024803840 Chr.18: 45983731-45983731 on Build GRCh38
rs924968239 Chr.18: 45983427-45983427 on Build GRCh38
rs9630785 Chr.18: 45983506-45983506 on Build GRCh38
rs376450101 Chr.18: 45983440-45983440 on Build GRCh38
rs879379863 Chr.18: 45983497-45983497 on Build GRCh38

View Full Product Details