Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51861904-51862362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC70
Entrez Gene ID
83446
Gene Name
coiled-coil domain containing 70
Gene Aliases
-
UniGene
Hs.120573
Gene Chromosome Location
Chr.13: 51861981-51866236 on Build GRCh38
Transcripts
NM_031290.2

SNP Detail

SNP ID Chromosome Location
rs973465483 Chr.13: 51862288-51862288 on Build GRCh38
rs1035918041 Chr.13: 51862272-51862272 on Build GRCh38
rs370253619 Chr.13: 51861962-51861962 on Build GRCh38
rs977653600 Chr.13: 51861999-51861999 on Build GRCh38
rs761842675 Chr.13: 51861981-51861981 on Build GRCh38
rs940717587 Chr.13: 51862006-51862006 on Build GRCh38
rs1021371118 Chr.13: 51862162-51862162 on Build GRCh38
rs558392927 Chr.13: 51862021-51862021 on Build GRCh38
rs916027530 Chr.13: 51862041-51862041 on Build GRCh38
rs931158369 Chr.13: 51862054-51862054 on Build GRCh38
rs1036281602 Chr.13: 51862145-51862145 on Build GRCh38
rs907895102 Chr.13: 51861991-51861991 on Build GRCh38
rs993209088 Chr.13: 51861986-51861986 on Build GRCh38
rs566392518 Chr.13: 51862102-51862102 on Build GRCh38
rs889670372 Chr.13: 51862097-51862097 on Build GRCh38
rs552781190 Chr.13: 51862056-51862056 on Build GRCh38
rs1021297654 Chr.13: 51862257-51862257 on Build GRCh38
rs940471363 Chr.13: 51862144-51862144 on Build GRCh38
rs993098672 Chr.13: 51862241-51862241 on Build GRCh38
rs146463174 Chr.13: 51862037-51862037 on Build GRCh38
rs960557258 Chr.13: 51862287-51862287 on Build GRCh38
rs34322019 Chr.13: 51862182-51862182 on Build GRCh38
rs565000569 Chr.13: 51862002-51862002 on Build GRCh38
rs561467879 Chr.13: 51862292-51862292 on Build GRCh38
rs369177510 Chr.13: 51862106-51862106 on Build GRCh38
rs933012904 Chr.13: 51862177-51862177 on Build GRCh38
rs1011772040 Chr.13: 51861932-51861932 on Build GRCh38
rs554048277 Chr.13: 51862146-51862146 on Build GRCh38
rs185013376 Chr.13: 51862293-51862293 on Build GRCh38
rs115987271 Chr.13: 51862239-51862239 on Build GRCh38
rs920705962 Chr.13: 51862308-51862308 on Build GRCh38
rs891468334 Chr.13: 51862138-51862138 on Build GRCh38
rs984660249 Chr.13: 51862057-51862057 on Build GRCh38
rs1050498126 Chr.13: 51862181-51862181 on Build GRCh38
rs1041068417 Chr.13: 51862104-51862104 on Build GRCh38
rs903364569 Chr.13: 51862265-51862265 on Build GRCh38
rs991528245 Chr.13: 51862332-51862332 on Build GRCh38
rs1002140431 Chr.13: 51862207-51862207 on Build GRCh38
rs200671997 Chr.13: 51862026-51862026 on Build GRCh38
rs111611094 Chr.13: 51862211-51862211 on Build GRCh38
rs781510217 Chr.13: 51862244-51862244 on Build GRCh38
rs998956189 Chr.13: 51862268-51862268 on Build GRCh38

View Full Product Details