Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94593437-94594033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBP2
Entrez Gene ID
8789
Gene Name
fructose-bisphosphatase 2
Gene Aliases
-
UniGene
Hs.61255
Gene Chromosome Location
Chr.9: 94558714-94593832 on Build GRCh38
Transcripts
NM_003837.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111363 c.22G>A p.E8K Chr.9: 94593705-94593705 on Build GRCh38
1206754 c.119C>T p.T40M Chr.9: 94593608-94593608 on Build GRCh38
3927284 c.40C>T p.L14L Chr.9: 94593687-94593687 on Build GRCh38
6661806 c.157G>A p.G53S Chr.9: 94593570-94593570 on Build GRCh38
3909124 c.148C>T p.R50C Chr.9: 94593579-94593579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376760370 Chr.9: 94593508-94593508 on Build GRCh38
rs181595760 Chr.9: 94593785-94593785 on Build GRCh38
rs753736391 Chr.9: 94593601-94593601 on Build GRCh38
rs757012592 Chr.9: 94593538-94593538 on Build GRCh38
rs749092184 Chr.9: 94593535-94593535 on Build GRCh38
rs778439796 Chr.9: 94593964-94593964 on Build GRCh38
rs868519987 Chr.9: 94593844-94593844 on Build GRCh38
rs772284798 Chr.9: 94593747-94593747 on Build GRCh38
rs1019191387 Chr.9: 94593691-94593691 on Build GRCh38
rs189097557 Chr.9: 94593703-94593703 on Build GRCh38
rs754512471 Chr.9: 94593711-94593711 on Build GRCh38
rs759667090 Chr.9: 94593680-94593680 on Build GRCh38
rs996808373 Chr.9: 94593914-94593914 on Build GRCh38
rs10993255 Chr.9: 94593949-94593949 on Build GRCh38
rs751160942 Chr.9: 94593707-94593707 on Build GRCh38
rs771646420 Chr.9: 94593563-94593563 on Build GRCh38
rs764092748 Chr.9: 94593593-94593593 on Build GRCh38
rs761904090 Chr.9: 94593752-94593752 on Build GRCh38
rs752148715 Chr.9: 94593713-94593713 on Build GRCh38
rs893561330 Chr.9: 94593671-94593671 on Build GRCh38
rs370129734 Chr.9: 94593586-94593586 on Build GRCh38
rs774461696 Chr.9: 94593735-94593735 on Build GRCh38
rs375620140 Chr.9: 94593654-94593654 on Build GRCh38
rs753859617 Chr.9: 94593686-94593686 on Build GRCh38
rs757142905 Chr.9: 94593607-94593607 on Build GRCh38
rs145086444 Chr.9: 94593706-94593706 on Build GRCh38
rs939953138 Chr.9: 94593520-94593520 on Build GRCh38
rs750516241 Chr.9: 94593510-94593510 on Build GRCh38
rs939646721 Chr.9: 94593832-94593832 on Build GRCh38
rs73531485 Chr.9: 94594000-94594000 on Build GRCh38
rs552506256 Chr.9: 94593923-94593923 on Build GRCh38
rs761779339 Chr.9: 94593700-94593700 on Build GRCh38
rs758815381 Chr.9: 94593518-94593518 on Build GRCh38
rs745464258 Chr.9: 94593552-94593552 on Build GRCh38
rs775657117 Chr.9: 94593751-94593751 on Build GRCh38
rs769142942 Chr.9: 94593571-94593571 on Build GRCh38
rs778688824 Chr.9: 94593546-94593546 on Build GRCh38
rs910542584 Chr.9: 94593503-94593503 on Build GRCh38
rs900756543 Chr.9: 94593913-94593913 on Build GRCh38
rs35600708 Chr.9: 94593961-94593961 on Build GRCh38
rs763293145 Chr.9: 94593673-94593673 on Build GRCh38
rs762140130 Chr.9: 94593572-94593572 on Build GRCh38
rs548702030 Chr.9: 94593977-94593977 on Build GRCh38
rs898955362 Chr.9: 94593943-94593943 on Build GRCh38
rs527469627 Chr.9: 94593811-94593811 on Build GRCh38
rs902221747 Chr.9: 94593864-94593864 on Build GRCh38
rs754566439 Chr.9: 94593778-94593778 on Build GRCh38
rs777769948 Chr.9: 94593523-94593523 on Build GRCh38
rs748666964 Chr.9: 94593720-94593720 on Build GRCh38
rs371067816 Chr.9: 94593631-94593631 on Build GRCh38
rs796654815 Chr.9: 94593889-94593889 on Build GRCh38
rs200620421 Chr.9: 94593587-94593587 on Build GRCh38
rs773104262 Chr.9: 94593758-94593758 on Build GRCh38
rs1018993218 Chr.9: 94593907-94593907 on Build GRCh38
rs114230965 Chr.9: 94593661-94593661 on Build GRCh38
rs780611933 Chr.9: 94593714-94593714 on Build GRCh38
rs1026130728 Chr.9: 94593951-94593951 on Build GRCh38
rs965281795 Chr.9: 94593693-94593693 on Build GRCh38
rs769197968 Chr.9: 94593659-94593659 on Build GRCh38
rs765050214 Chr.9: 94593757-94593757 on Build GRCh38
rs779678092 Chr.9: 94593635-94593635 on Build GRCh38
rs759794555 Chr.9: 94593737-94593737 on Build GRCh38
rs766630515 Chr.9: 94593681-94593681 on Build GRCh38
rs1009178802 Chr.9: 94593690-94593690 on Build GRCh38
rs993208534 Chr.9: 94593950-94593950 on Build GRCh38
rs376034537 Chr.9: 94593579-94593579 on Build GRCh38
rs770271718 Chr.9: 94593721-94593721 on Build GRCh38
rs757934614 Chr.9: 94593632-94593632 on Build GRCh38
rs773390438 Chr.9: 94593665-94593665 on Build GRCh38
rs141358375 Chr.9: 94593588-94593588 on Build GRCh38
rs758526263 Chr.9: 94593511-94593511 on Build GRCh38
rs893603406 Chr.9: 94593796-94593796 on Build GRCh38
rs575545531 Chr.9: 94593570-94593570 on Build GRCh38
rs952199942 Chr.9: 94593938-94593938 on Build GRCh38
rs755946534 Chr.9: 94593522-94593522 on Build GRCh38
rs538285249 Chr.9: 94594006-94594006 on Build GRCh38
rs771054913 Chr.9: 94593675-94593675 on Build GRCh38
rs556448037 Chr.9: 94593504-94593504 on Build GRCh38
rs906381897 Chr.9: 94593559-94593559 on Build GRCh38
rs765534622 Chr.9: 94593578-94593578 on Build GRCh38
rs567100303 Chr.9: 94593978-94593978 on Build GRCh38
rs774531216 Chr.9: 94593676-94593676 on Build GRCh38
rs766329123 Chr.9: 94593521-94593521 on Build GRCh38
rs527401557 Chr.9: 94593962-94593962 on Build GRCh38
rs201242837 Chr.9: 94593759-94593759 on Build GRCh38
rs748612104 Chr.9: 94593660-94593660 on Build GRCh38
rs1051809223 Chr.9: 94593499-94593499 on Build GRCh38
rs971846577 Chr.9: 94593456-94593456 on Build GRCh38
rs774799689 Chr.9: 94593564-94593564 on Build GRCh38
rs61465993 Chr.9: 94593468-94593468 on Build GRCh38
rs751215910 Chr.9: 94593768-94593768 on Build GRCh38
rs902169475 Chr.9: 94593689-94593689 on Build GRCh38
rs758106624 Chr.9: 94593705-94593705 on Build GRCh38
rs557921747 Chr.9: 94593608-94593608 on Build GRCh38
rs933200798 Chr.9: 94593495-94593495 on Build GRCh38
rs766155930 Chr.9: 94593762-94593762 on Build GRCh38
rs532751754 Chr.9: 94593867-94593867 on Build GRCh38
rs148648693 Chr.9: 94593806-94593806 on Build GRCh38
rs374397427 Chr.9: 94593722-94593722 on Build GRCh38

View Full Product Details