Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 54100210-54100725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSX2
Entrez Gene ID
170825
Gene Name
GS homeobox 2
Gene Aliases
GSH2
UniGene
Hs.371899
Gene Chromosome Location
Chr.4: 54100081-54101955 on Build GRCh38
Transcripts
NM_133267.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4159109 c.319G>A p.G107S Chr.4: 54100663-54100663 on Build GRCh38
281581 c.75G>A p.P25P Chr.4: 54100419-54100419 on Build GRCh38
6269290 c.324G>T p.Q108H Chr.4: 54100668-54100668 on Build GRCh38
4135809 c.255C>T p.A85A Chr.4: 54100599-54100599 on Build GRCh38
4546894 c.40G>A p.D14N Chr.4: 54100384-54100384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567632527 Chr.4: 54100374-54100374 on Build GRCh38
rs367974428 Chr.4: 54100308-54100308 on Build GRCh38
rs897750791 Chr.4: 54100627-54100627 on Build GRCh38
rs781328528 Chr.4: 54100298-54100298 on Build GRCh38
rs757156464 Chr.4: 54100589-54100589 on Build GRCh38
rs1056109404 Chr.4: 54100623-54100623 on Build GRCh38
rs939289682 Chr.4: 54100388-54100388 on Build GRCh38
rs926321201 Chr.4: 54100614-54100614 on Build GRCh38
rs761947645 Chr.4: 54100435-54100437 on Build GRCh38
rs750756097 Chr.4: 54100474-54100474 on Build GRCh38
rs993340679 Chr.4: 54100418-54100418 on Build GRCh38
rs1018044237 Chr.4: 54100518-54100518 on Build GRCh38
rs779734371 Chr.4: 54100615-54100615 on Build GRCh38
rs897696944 Chr.4: 54100407-54100407 on Build GRCh38
rs13144341 Chr.4: 54100663-54100663 on Build GRCh38
rs370144954 Chr.4: 54100301-54100301 on Build GRCh38
rs144560606 Chr.4: 54100365-54100365 on Build GRCh38
rs752052904 Chr.4: 54100463-54100463 on Build GRCh38
rs776143162 Chr.4: 54100350-54100350 on Build GRCh38
rs766743168 Chr.4: 54100667-54100667 on Build GRCh38
rs770861562 Chr.4: 54100503-54100503 on Build GRCh38
rs768017023 Chr.4: 54100558-54100558 on Build GRCh38
rs537141111 Chr.4: 54100361-54100361 on Build GRCh38
rs557068792 Chr.4: 54100362-54100362 on Build GRCh38
rs750584127 Chr.4: 54100375-54100375 on Build GRCh38
rs745621052 Chr.4: 54100509-54100509 on Build GRCh38
rs961855466 Chr.4: 54100243-54100243 on Build GRCh38
rs759142365 Chr.4: 54100422-54100422 on Build GRCh38
rs754099416 Chr.4: 54100565-54100565 on Build GRCh38
rs917591766 Chr.4: 54100609-54100609 on Build GRCh38
rs1027452125 Chr.4: 54100495-54100495 on Build GRCh38
rs1031890961 Chr.4: 54100551-54100551 on Build GRCh38
rs555599449 Chr.4: 54100381-54100381 on Build GRCh38
rs150979273 Chr.4: 54100508-54100508 on Build GRCh38
rs745718839 Chr.4: 54100611-54100611 on Build GRCh38
rs768136730 Chr.4: 54100527-54100527 on Build GRCh38
rs755102374 Chr.4: 54100496-54100496 on Build GRCh38
rs140713069 Chr.4: 54100353-54100353 on Build GRCh38
rs201536623 Chr.4: 54100390-54100390 on Build GRCh38
rs952131842 Chr.4: 54100289-54100289 on Build GRCh38
rs773160470 Chr.4: 54100336-54100336 on Build GRCh38
rs774877384 Chr.4: 54100447-54100447 on Build GRCh38
rs749328981 Chr.4: 54100386-54100386 on Build GRCh38
rs548038657 Chr.4: 54100316-54100316 on Build GRCh38
rs563266158 Chr.4: 54100579-54100579 on Build GRCh38
rs781120072 Chr.4: 54100601-54100601 on Build GRCh38
rs867902995 Chr.4: 54100567-54100567 on Build GRCh38
rs375038462 Chr.4: 54100562-54100562 on Build GRCh38
rs541178150 Chr.4: 54100473-54100473 on Build GRCh38
rs762265876 Chr.4: 54100624-54100624 on Build GRCh38
rs976336764 Chr.4: 54100540-54100540 on Build GRCh38
rs775207811 Chr.4: 54100522-54100522 on Build GRCh38
rs772458969 Chr.4: 54100648-54100648 on Build GRCh38
rs776387289 Chr.4: 54100420-54100420 on Build GRCh38
rs571063101 Chr.4: 54100337-54100337 on Build GRCh38
rs762217977 Chr.4: 54100465-54100465 on Build GRCh38
rs371417798 Chr.4: 54100477-54100477 on Build GRCh38
rs140790882 Chr.4: 54100512-54100512 on Build GRCh38
rs746920585 Chr.4: 54100502-54100502 on Build GRCh38
rs148460532 Chr.4: 54100404-54100404 on Build GRCh38
rs764654441 Chr.4: 54100462-54100462 on Build GRCh38
rs990242170 Chr.4: 54100323-54100323 on Build GRCh38
rs773762938 Chr.4: 54100653-54100653 on Build GRCh38
rs796644145 Chr.4: 54100702-54100702 on Build GRCh38
rs759143507 Chr.4: 54100351-54100351 on Build GRCh38
rs528852767 Chr.4: 54100612-54100612 on Build GRCh38
rs962405464 Chr.4: 54100253-54100253 on Build GRCh38
rs542267886 Chr.4: 54100616-54100616 on Build GRCh38
rs996827359 Chr.4: 54100449-54100449 on Build GRCh38
rs756310653 Chr.4: 54100476-54100476 on Build GRCh38
rs762385629 Chr.4: 54100453-54100453 on Build GRCh38
rs946486362 Chr.4: 54100602-54100602 on Build GRCh38
rs866843178 Chr.4: 54100658-54100658 on Build GRCh38
rs780026264 Chr.4: 54100383-54100383 on Build GRCh38
rs766132573 Chr.4: 54100344-54100344 on Build GRCh38
rs769441688 Chr.4: 54100429-54100429 on Build GRCh38
rs966066757 Chr.4: 54100529-54100529 on Build GRCh38
rs759859762 Chr.4: 54100681-54100681 on Build GRCh38
rs3194383 Chr.4: 54100500-54100500 on Build GRCh38
rs752777000 Chr.4: 54100585-54100585 on Build GRCh38
rs1008093986 Chr.4: 54100706-54100706 on Build GRCh38
rs200996880 Chr.4: 54100483-54100483 on Build GRCh38
rs777034082 Chr.4: 54100676-54100676 on Build GRCh38
rs924819025 Chr.4: 54100315-54100315 on Build GRCh38
rs375280653 Chr.4: 54100305-54100305 on Build GRCh38
rs768253284 Chr.4: 54100631-54100631 on Build GRCh38
rs1027733604 Chr.4: 54100274-54100274 on Build GRCh38
rs993622489 Chr.4: 54100255-54100255 on Build GRCh38
rs145597698 Chr.4: 54100378-54100378 on Build GRCh38
rs370613209 Chr.4: 54100488-54100488 on Build GRCh38
rs762336257 Chr.4: 54100549-54100549 on Build GRCh38
rs559949387 Chr.4: 54100652-54100652 on Build GRCh38
rs1056393577 Chr.4: 54100406-54100406 on Build GRCh38
rs774874463 Chr.4: 54100545-54100545 on Build GRCh38
rs865868726 Chr.4: 54100544-54100544 on Build GRCh38
rs774392752 Chr.4: 54100307-54100307 on Build GRCh38
rs772092478 Chr.4: 54100316-54100316 on Build GRCh38
rs887606692 Chr.4: 54100651-54100651 on Build GRCh38
rs765499335 Chr.4: 54100687-54100687 on Build GRCh38
rs917472618 Chr.4: 54100313-54100313 on Build GRCh38
rs371317920 Chr.4: 54100526-54100526 on Build GRCh38
rs761032211 Chr.4: 54100559-54100559 on Build GRCh38
rs755157514 Chr.4: 54100566-54100566 on Build GRCh38
rs773869658 Chr.4: 54100639-54100639 on Build GRCh38
rs377377329 Chr.4: 54100352-54100352 on Build GRCh38

View Full Product Details