Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 118143385-118143917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDST3
Entrez Gene ID
9348
Gene Name
N-deacetylase and N-sulfotransferase 3
Gene Aliases
HSST3
UniGene
Hs.480596
Gene Chromosome Location
Chr.4: 118033596-118258945 on Build GRCh38
Transcripts
NM_004784.2 XM_006714416.3 XM_011532415.2 XM_011532416.2 XM_017008839.1 XM_017008840.1 XM_017008841.1 XM_017008842.1 XM_017008843.1 XM_017008844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618313 c.1435C>T p.L479F Chr.4: 118143580-118143580 on Build GRCh38
3722731 c.1416C>T p.L472L Chr.4: 118143561-118143561 on Build GRCh38
1050328 c.1445A>G p.H482R Chr.4: 118143590-118143590 on Build GRCh38
3409022 c.1455C>T p.F485F Chr.4: 118143600-118143600 on Build GRCh38
3007558 c.1509A>G p.G503G Chr.4: 118143654-118143654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760223789 Chr.4: 118143684-118143684 on Build GRCh38
rs978990330 Chr.4: 118143833-118143833 on Build GRCh38
rs573884040 Chr.4: 118143892-118143892 on Build GRCh38
rs1011795396 Chr.4: 118143507-118143507 on Build GRCh38
rs541601885 Chr.4: 118143512-118143513 on Build GRCh38
rs1046675537 Chr.4: 118143436-118143436 on Build GRCh38
rs568725430 Chr.4: 118143696-118143696 on Build GRCh38
rs938594338 Chr.4: 118143458-118143458 on Build GRCh38
rs982448226 Chr.4: 118143828-118143828 on Build GRCh38
rs780574562 Chr.4: 118143870-118143870 on Build GRCh38
rs796104058 Chr.4: 118143513-118143514 on Build GRCh38
rs749435873 Chr.4: 118143485-118143485 on Build GRCh38
rs746846813 Chr.4: 118143720-118143720 on Build GRCh38
rs535128415 Chr.4: 118143685-118143685 on Build GRCh38
rs534123995 Chr.4: 118143848-118143848 on Build GRCh38
rs776546871 Chr.4: 118143655-118143655 on Build GRCh38
rs767102073 Chr.4: 118143522-118143522 on Build GRCh38
rs267599996 Chr.4: 118143622-118143622 on Build GRCh38
rs367825303 Chr.4: 118143653-118143653 on Build GRCh38
rs755832650 Chr.4: 118143565-118143565 on Build GRCh38
rs370396586 Chr.4: 118143726-118143726 on Build GRCh38
rs750650307 Chr.4: 118143525-118143526 on Build GRCh38
rs929714597 Chr.4: 118143789-118143789 on Build GRCh38
rs751242927 Chr.4: 118143417-118143417 on Build GRCh38
rs755120141 Chr.4: 118143725-118143725 on Build GRCh38
rs565016977 Chr.4: 118143753-118143753 on Build GRCh38
rs548687829 Chr.4: 118143689-118143689 on Build GRCh38
rs562693464 Chr.4: 118143427-118143427 on Build GRCh38
rs773724696 Chr.4: 118143672-118143672 on Build GRCh38
rs780354655 Chr.4: 118143635-118143635 on Build GRCh38
rs753438148 Chr.4: 118143590-118143590 on Build GRCh38
rs1049749546 Chr.4: 118143740-118143740 on Build GRCh38
rs926091611 Chr.4: 118143415-118143415 on Build GRCh38
rs761988786 Chr.4: 118143656-118143656 on Build GRCh38
rs748642066 Chr.4: 118143513-118143514 on Build GRCh38
rs751980811 Chr.4: 118143678-118143678 on Build GRCh38
rs756813881 Chr.4: 118143695-118143695 on Build GRCh38
rs763238949 Chr.4: 118143673-118143673 on Build GRCh38
rs1011846989 Chr.4: 118143762-118143762 on Build GRCh38
rs756443530 Chr.4: 118143865-118143865 on Build GRCh38
rs988916037 Chr.4: 118143860-118143860 on Build GRCh38
rs900404339 Chr.4: 118143459-118143459 on Build GRCh38
rs867038662 Chr.4: 118143466-118143466 on Build GRCh38
rs1018516767 Chr.4: 118143721-118143721 on Build GRCh38
rs567526951 Chr.4: 118143572-118143572 on Build GRCh38
rs577865250 Chr.4: 118143847-118143847 on Build GRCh38
rs779271438 Chr.4: 118143514-118143514 on Build GRCh38
rs67092892 Chr.4: 118143512-118143512 on Build GRCh38
rs750717824 Chr.4: 118143506-118143506 on Build GRCh38
rs938624118 Chr.4: 118143844-118143844 on Build GRCh38
rs201448015 Chr.4: 118143509-118143509 on Build GRCh38
rs750389120 Chr.4: 118143704-118143704 on Build GRCh38
rs1053161656 Chr.4: 118143426-118143426 on Build GRCh38
rs557494997 Chr.4: 118143742-118143742 on Build GRCh38
rs745607453 Chr.4: 118143625-118143625 on Build GRCh38
rs879137812 Chr.4: 118143723-118143723 on Build GRCh38
rs770155552 Chr.4: 118143664-118143664 on Build GRCh38
rs1049367220 Chr.4: 118143469-118143469 on Build GRCh38
rs10033308 Chr.4: 118143438-118143438 on Build GRCh38
rs748122184 Chr.4: 118143729-118143729 on Build GRCh38
rs956205349 Chr.4: 118143851-118143851 on Build GRCh38
rs758474312 Chr.4: 118143706-118143706 on Build GRCh38
rs911911124 Chr.4: 118143428-118143428 on Build GRCh38
rs888469876 Chr.4: 118143778-118143778 on Build GRCh38
rs3749506 Chr.4: 118143513-118143513 on Build GRCh38
rs267599997 Chr.4: 118143623-118143623 on Build GRCh38
rs758456807 Chr.4: 118143703-118143706 on Build GRCh38
rs769925025 Chr.4: 118143731-118143731 on Build GRCh38
rs553970811 Chr.4: 118143887-118143887 on Build GRCh38
rs369281202 Chr.4: 118143445-118143445 on Build GRCh38
rs537831723 Chr.4: 118143712-118143712 on Build GRCh38
rs757111328 Chr.4: 118143595-118143595 on Build GRCh38
rs778939851 Chr.4: 118143617-118143617 on Build GRCh38
rs772004450 Chr.4: 118143633-118143633 on Build GRCh38
rs752353564 Chr.4: 118143541-118143541 on Build GRCh38
rs764629361 Chr.4: 118143416-118143416 on Build GRCh38
rs1023200120 Chr.4: 118143770-118143770 on Build GRCh38
rs970706352 Chr.4: 118143808-118143808 on Build GRCh38
rs886234874 Chr.4: 118143594-118143594 on Build GRCh38
rs113975339 Chr.4: 118143700-118143700 on Build GRCh38
rs531756725 Chr.4: 118143434-118143434 on Build GRCh38
rs894327215 Chr.4: 118143468-118143468 on Build GRCh38
rs528006849 Chr.4: 118143484-118143484 on Build GRCh38
rs747227062 Chr.4: 118143639-118143639 on Build GRCh38
rs780030371 Chr.4: 118143732-118143733 on Build GRCh38
rs766727875 Chr.4: 118143676-118143676 on Build GRCh38

View Full Product Details