Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75313302-75313901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
128007 c.2430C>A p.V810V Chr.6: 75313402-75313402 on Build GRCh38
743101 c.2131C>T p.R711W Chr.6: 75313701-75313701 on Build GRCh38
239880 c.2005A>G p.T669A Chr.6: 75313827-75313827 on Build GRCh38
6876904 c.2481C>T p.S827S Chr.6: 75313351-75313351 on Build GRCh38
4428313 c.2152C>T p.R718* Chr.6: 75313680-75313680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367963442 Chr.6: 75313728-75313728 on Build GRCh38
rs764736807 Chr.6: 75313387-75313388 on Build GRCh38
rs764200052 Chr.6: 75313535-75313535 on Build GRCh38
rs746495331 Chr.6: 75313389-75313389 on Build GRCh38
rs759249502 Chr.6: 75313794-75313794 on Build GRCh38
rs777289478 Chr.6: 75313605-75313605 on Build GRCh38
rs781231109 Chr.6: 75313580-75313580 on Build GRCh38
rs1023596934 Chr.6: 75313663-75313663 on Build GRCh38
rs777282127 Chr.6: 75313708-75313708 on Build GRCh38
rs542938844 Chr.6: 75313491-75313491 on Build GRCh38
rs150255059 Chr.6: 75313335-75313335 on Build GRCh38
rs761316787 Chr.6: 75313399-75313399 on Build GRCh38
rs371768415 Chr.6: 75313787-75313787 on Build GRCh38
rs1014403478 Chr.6: 75313460-75313460 on Build GRCh38
rs746259489 Chr.6: 75313645-75313645 on Build GRCh38
rs767280732 Chr.6: 75313798-75313798 on Build GRCh38
rs780180968 Chr.6: 75313380-75313380 on Build GRCh38
rs200898378 Chr.6: 75313678-75313678 on Build GRCh38
rs145877579 Chr.6: 75313452-75313452 on Build GRCh38
rs773410611 Chr.6: 75313418-75313418 on Build GRCh38
rs760615391 Chr.6: 75313838-75313838 on Build GRCh38
rs769301224 Chr.6: 75313503-75313503 on Build GRCh38
rs763294278 Chr.6: 75313511-75313511 on Build GRCh38
rs370823048 Chr.6: 75313376-75313376 on Build GRCh38
rs141477236 Chr.6: 75313736-75313736 on Build GRCh38
rs531280086 Chr.6: 75313472-75313472 on Build GRCh38
rs141786046 Chr.6: 75313792-75313792 on Build GRCh38
rs752450803 Chr.6: 75313824-75313824 on Build GRCh38
rs747726048 Chr.6: 75313492-75313492 on Build GRCh38
rs747008532 Chr.6: 75313490-75313490 on Build GRCh38
rs776109907 Chr.6: 75313665-75313665 on Build GRCh38
rs776455963 Chr.6: 75313701-75313701 on Build GRCh38
rs751424079 Chr.6: 75313557-75313557 on Build GRCh38
rs765589504 Chr.6: 75313688-75313688 on Build GRCh38
rs778083612 Chr.6: 75313454-75313454 on Build GRCh38
rs747481132 Chr.6: 75313743-75313743 on Build GRCh38
rs767520996 Chr.6: 75313439-75313439 on Build GRCh38
rs780125831 Chr.6: 75313475-75313475 on Build GRCh38
rs542112601 Chr.6: 75313375-75313375 on Build GRCh38
rs772424046 Chr.6: 75313654-75313654 on Build GRCh38
rs760845175 Chr.6: 75313532-75313532 on Build GRCh38
rs758261361 Chr.6: 75313547-75313547 on Build GRCh38
rs772545086 Chr.6: 75313741-75313741 on Build GRCh38
rs752749763 Chr.6: 75313705-75313705 on Build GRCh38
rs766854436 Chr.6: 75313425-75313425 on Build GRCh38
rs751607164 Chr.6: 75313325-75313325 on Build GRCh38
rs769055972 Chr.6: 75313755-75313755 on Build GRCh38
rs768337770 Chr.6: 75313397-75313397 on Build GRCh38
rs896231495 Chr.6: 75313456-75313456 on Build GRCh38
rs763966569 Chr.6: 75313856-75313856 on Build GRCh38
rs146541750 Chr.6: 75313540-75313540 on Build GRCh38
rs921803616 Chr.6: 75313661-75313661 on Build GRCh38
rs147346630 Chr.6: 75313534-75313534 on Build GRCh38
rs757717364 Chr.6: 75313550-75313552 on Build GRCh38
rs373828183 Chr.6: 75313710-75313710 on Build GRCh38
rs142709604 Chr.6: 75313513-75313513 on Build GRCh38
rs753312674 Chr.6: 75313875-75313875 on Build GRCh38
rs768030723 Chr.6: 75313525-75313525 on Build GRCh38
rs775856904 Chr.6: 75313742-75313742 on Build GRCh38
rs78566620 Chr.6: 75313430-75313430 on Build GRCh38
rs867498559 Chr.6: 75313643-75313643 on Build GRCh38
rs776854335 Chr.6: 75313679-75313679 on Build GRCh38
rs578080275 Chr.6: 75313398-75313398 on Build GRCh38
rs774706544 Chr.6: 75313516-75313516 on Build GRCh38
rs148968530 Chr.6: 75313429-75313429 on Build GRCh38
rs759981199 Chr.6: 75313524-75313524 on Build GRCh38
rs573177009 Chr.6: 75313352-75313352 on Build GRCh38
rs762026679 Chr.6: 75313765-75313765 on Build GRCh38
rs267601126 Chr.6: 75313357-75313357 on Build GRCh38
rs753495863 Chr.6: 75313487-75313487 on Build GRCh38
rs1023114778 Chr.6: 75313466-75313466 on Build GRCh38
rs906002829 Chr.6: 75313478-75313478 on Build GRCh38
rs762067637 Chr.6: 75313680-75313680 on Build GRCh38
rs910634974 Chr.6: 75313479-75313479 on Build GRCh38
rs766207449 Chr.6: 75313554-75313554 on Build GRCh38
rs1031387301 Chr.6: 75313703-75313703 on Build GRCh38
rs746206187 Chr.6: 75313737-75313737 on Build GRCh38
rs1044004957 Chr.6: 75313392-75313392 on Build GRCh38
rs760657416 Chr.6: 75313666-75313666 on Build GRCh38
rs756013863 Chr.6: 75313706-75313706 on Build GRCh38
rs752675547 Chr.6: 75313530-75313530 on Build GRCh38
rs770586450 Chr.6: 75313635-75313635 on Build GRCh38
rs781382151 Chr.6: 75313346-75313346 on Build GRCh38
rs750150915 Chr.6: 75313471-75313471 on Build GRCh38
rs748875756 Chr.6: 75313629-75313629 on Build GRCh38
rs375456304 Chr.6: 75313648-75313648 on Build GRCh38
rs201842783 Chr.6: 75313591-75313591 on Build GRCh38
rs201906969 Chr.6: 75313877-75313877 on Build GRCh38
rs368729229 Chr.6: 75313441-75313441 on Build GRCh38
rs747667417 Chr.6: 75313761-75313761 on Build GRCh38
rs545378552 Chr.6: 75313419-75313419 on Build GRCh38
rs752090345 Chr.6: 75313449-75313451 on Build GRCh38
rs772967207 Chr.6: 75313508-75313508 on Build GRCh38
rs754928211 Chr.6: 75313569-75313569 on Build GRCh38
rs769511155 Chr.6: 75313404-75313404 on Build GRCh38
rs756361284 Chr.6: 75313358-75313358 on Build GRCh38
rs974803954 Chr.6: 75313656-75313656 on Build GRCh38
rs747590253 Chr.6: 75313590-75313590 on Build GRCh38
rs755925479 Chr.6: 75313359-75313359 on Build GRCh38
rs35449948 Chr.6: 75313698-75313698 on Build GRCh38
rs754618124 Chr.6: 75313636-75313636 on Build GRCh38

View Full Product Details