Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102407251-102407737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100630923
Entrez Gene ID
100630923
Gene Name
LOC100289561-PRKRIP1 readthrough
Gene Aliases
-
UniGene
Hs.406395
Gene Chromosome Location
Chr.7: 102363861-102426682 on Build GRCh38
Transcripts
NR_038967.1 NM_024653.3

Gene Symbol
PRKRIP1
Entrez Gene ID
79706
Gene Name
PRKR interacting protein 1 (IL11 inducible)
Gene Aliases
C114, KRBOX3
UniGene
Hs.722104
Gene Chromosome Location
Chr.7: 102396357-102426682 on Build GRCh38
Transcripts
NR_038967.1 NM_024653.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1222101 c.422C>T p.A141V Chr.7: 102407463-102407463 on Build GRCh38
6175349 c.429G>A p.K143K Chr.7: 102407470-102407470 on Build GRCh38
314453 c.421G>T p.A141S Chr.7: 102407462-102407462 on Build GRCh38
6398761 c.436C>A p.L146I Chr.7: 102407477-102407477 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1012635858 Chr.7: 102407647-102407647 on Build GRCh38
rs782274032 Chr.7: 102407478-102407478 on Build GRCh38
rs778711858 Chr.7: 102407275-102407275 on Build GRCh38
rs375884258 Chr.7: 102407511-102407511 on Build GRCh38
rs573180045 Chr.7: 102407303-102407303 on Build GRCh38
rs1021430949 Chr.7: 102407287-102407287 on Build GRCh38
rs781892046 Chr.7: 102407516-102407516 on Build GRCh38
rs781850942 Chr.7: 102407392-102407392 on Build GRCh38
rs1031604288 Chr.7: 102407375-102407375 on Build GRCh38
rs201991805 Chr.7: 102407390-102407390 on Build GRCh38
rs929494754 Chr.7: 102407568-102407568 on Build GRCh38
rs977304836 Chr.7: 102407341-102407341 on Build GRCh38
rs562560007 Chr.7: 102407575-102407575 on Build GRCh38
rs782793835 Chr.7: 102407513-102407513 on Build GRCh38
rs782065098 Chr.7: 102407499-102407499 on Build GRCh38
rs782372299 Chr.7: 102407477-102407477 on Build GRCh38
rs782592697 Chr.7: 102407697-102407697 on Build GRCh38
rs144259551 Chr.7: 102407670-102407670 on Build GRCh38
rs782521102 Chr.7: 102407426-102407426 on Build GRCh38
rs1048428617 Chr.7: 102407580-102407580 on Build GRCh38
rs894147943 Chr.7: 102407605-102407605 on Build GRCh38
rs201385162 Chr.7: 102407406-102407406 on Build GRCh38
rs187603433 Chr.7: 102407340-102407340 on Build GRCh38
rs782445135 Chr.7: 102407461-102407461 on Build GRCh38
rs191367889 Chr.7: 102407523-102407523 on Build GRCh38
rs564161834 Chr.7: 102407276-102407276 on Build GRCh38
rs782673975 Chr.7: 102407428-102407428 on Build GRCh38
rs782596621 Chr.7: 102407462-102407462 on Build GRCh38
rs373993073 Chr.7: 102407548-102407548 on Build GRCh38
rs782419065 Chr.7: 102407482-102407482 on Build GRCh38
rs552739947 Chr.7: 102407676-102407676 on Build GRCh38
rs782485431 Chr.7: 102407410-102407410 on Build GRCh38
rs782705379 Chr.7: 102407525-102407525 on Build GRCh38
rs782735024 Chr.7: 102407506-102407506 on Build GRCh38
rs373441521 Chr.7: 102407485-102407485 on Build GRCh38
rs782697770 Chr.7: 102407396-102407396 on Build GRCh38
rs782542328 Chr.7: 102407389-102407389 on Build GRCh38
rs987516701 Chr.7: 102407467-102407467 on Build GRCh38
rs781853529 Chr.7: 102407594-102407594 on Build GRCh38
rs782739866 Chr.7: 102407412-102407412 on Build GRCh38
rs149978574 Chr.7: 102407595-102407595 on Build GRCh38
rs782220241 Chr.7: 102407466-102407466 on Build GRCh38
rs77150298 Chr.7: 102407581-102407581 on Build GRCh38
rs782303686 Chr.7: 102407458-102407458 on Build GRCh38
rs781856645 Chr.7: 102407420-102407420 on Build GRCh38

View Full Product Details