Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92454287-92454711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD1
Entrez Gene ID
57798
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
CMD2B, ODAG, RG083M05.2
UniGene
Hs.21145
Gene Chromosome Location
Chr.7: 92447448-92494631 on Build GRCh38
Transcripts
NM_021167.4 NR_052016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5710742 c.505C>T p.Q169* Chr.7: 92454571-92454571 on Build GRCh38
5710743 c.557C>T p.T186M Chr.7: 92454623-92454623 on Build GRCh38
6669593 c.436-7delC p.? Chr.7: 92454495-92454495 on Build GRCh38
5633207 c.467C>T p.S156F Chr.7: 92454533-92454533 on Build GRCh38
402904 c.561G>T p.W187C Chr.7: 92454627-92454627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761271091 Chr.7: 92454585-92454585 on Build GRCh38
rs138831121 Chr.7: 92454580-92454580 on Build GRCh38
rs564747350 Chr.7: 92454670-92454670 on Build GRCh38
rs935126885 Chr.7: 92454334-92454334 on Build GRCh38
rs761360783 Chr.7: 92454454-92454454 on Build GRCh38
rs540654499 Chr.7: 92454578-92454578 on Build GRCh38
rs1003809257 Chr.7: 92454522-92454522 on Build GRCh38
rs761551157 Chr.7: 92454495-92454495 on Build GRCh38
rs762414487 Chr.7: 92454674-92454674 on Build GRCh38
rs769038754 Chr.7: 92454648-92454648 on Build GRCh38
rs557622563 Chr.7: 92454342-92454342 on Build GRCh38
rs915361712 Chr.7: 92454367-92454367 on Build GRCh38
rs727503061 Chr.7: 92454603-92454603 on Build GRCh38
rs761228374 Chr.7: 92454451-92454451 on Build GRCh38
rs150863199 Chr.7: 92454669-92454669 on Build GRCh38
rs764819654 Chr.7: 92454474-92454474 on Build GRCh38
rs764733372 Chr.7: 92454586-92454586 on Build GRCh38
rs753314550 Chr.7: 92454453-92454453 on Build GRCh38
rs922458277 Chr.7: 92454333-92454333 on Build GRCh38
rs576944000 Chr.7: 92454618-92454618 on Build GRCh38
rs776757859 Chr.7: 92454378-92454378 on Build GRCh38
rs922487847 Chr.7: 92454677-92454677 on Build GRCh38
rs1017952508 Chr.7: 92454616-92454616 on Build GRCh38
rs781323574 Chr.7: 92454534-92454534 on Build GRCh38
rs904608535 Chr.7: 92454613-92454613 on Build GRCh38
rs969433320 Chr.7: 92454371-92454371 on Build GRCh38
rs778933082 Chr.7: 92454553-92454553 on Build GRCh38
rs767407947 Chr.7: 92454525-92454527 on Build GRCh38
rs370960226 Chr.7: 92454641-92454641 on Build GRCh38
rs754965302 Chr.7: 92454532-92454532 on Build GRCh38
rs931176011 Chr.7: 92454423-92454423 on Build GRCh38
rs779034033 Chr.7: 92454619-92454619 on Build GRCh38
rs397517295 Chr.7: 92454558-92454558 on Build GRCh38
rs764831879 Chr.7: 92454605-92454605 on Build GRCh38
rs377491014 Chr.7: 92454543-92454543 on Build GRCh38
rs943985310 Chr.7: 92454461-92454461 on Build GRCh38
rs569540564 Chr.7: 92454392-92454392 on Build GRCh38
rs569662774 Chr.7: 92454363-92454364 on Build GRCh38
rs908476780 Chr.7: 92454448-92454448 on Build GRCh38
rs758063690 Chr.7: 92454491-92454491 on Build GRCh38
rs762654572 Chr.7: 92454591-92454591 on Build GRCh38
rs919637934 Chr.7: 92454391-92454391 on Build GRCh38
rs775370055 Chr.7: 92454325-92454325 on Build GRCh38
rs794729022 Chr.7: 92454679-92454679 on Build GRCh38
rs141932124 Chr.7: 92454624-92454624 on Build GRCh38
rs187484662 Chr.7: 92454569-92454569 on Build GRCh38
rs905177333 Chr.7: 92454473-92454473 on Build GRCh38
rs1031781653 Chr.7: 92454657-92454657 on Build GRCh38
rs780188711 Chr.7: 92454644-92454644 on Build GRCh38
rs775915716 Chr.7: 92454573-92454573 on Build GRCh38
rs876657465 Chr.7: 92454489-92454489 on Build GRCh38
rs964024778 Chr.7: 92454317-92454317 on Build GRCh38
rs760623919 Chr.7: 92454340-92454340 on Build GRCh38
rs976639416 Chr.7: 92454655-92454655 on Build GRCh38
rs746113520 Chr.7: 92454559-92454559 on Build GRCh38
rs766013276 Chr.7: 92454597-92454597 on Build GRCh38
rs559216917 Chr.7: 92454363-92454363 on Build GRCh38
rs960306591 Chr.7: 92454339-92454339 on Build GRCh38
rs767515474 Chr.7: 92454604-92454604 on Build GRCh38
rs868277813 Chr.7: 92454328-92454328 on Build GRCh38
rs980526092 Chr.7: 92454372-92454372 on Build GRCh38
rs749359981 Chr.7: 92454535-92454535 on Build GRCh38
rs554905494 Chr.7: 92454526-92454526 on Build GRCh38
rs145592666 Chr.7: 92454590-92454590 on Build GRCh38
rs758612559 Chr.7: 92454642-92454642 on Build GRCh38
rs748669338 Chr.7: 92454667-92454667 on Build GRCh38
rs764652132 Chr.7: 92454452-92454452 on Build GRCh38
rs112028916 Chr.7: 92454431-92454431 on Build GRCh38
rs373528467 Chr.7: 92454495-92454495 on Build GRCh38
rs777057775 Chr.7: 92454659-92454659 on Build GRCh38
rs370413559 Chr.7: 92454357-92454357 on Build GRCh38
rs1056705241 Chr.7: 92454540-92454540 on Build GRCh38

View Full Product Details