Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151056378-151056959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5
Entrez Gene ID
1020
Gene Name
cyclin dependent kinase 5
Gene Aliases
LIS7, PSSALRE
UniGene
Hs.647078
Gene Chromosome Location
Chr.7: 151053812-151057965 on Build GRCh38
Transcripts
NM_001164410.2 NM_004935.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3832313 c.301G>C p.E101Q Chr.7: 151056591-151056591 on Build GRCh38
3374552 c.294C>T p.L98L Chr.7: 151056598-151056598 on Build GRCh38
3261437 c.168G>T p.K56N Chr.7: 151056934-151056934 on Build GRCh38
1087357 c.201T>C p.H67H Chr.7: 151056790-151056790 on Build GRCh38
5528499 c.298C>T p.P100S Chr.7: 151056594-151056594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762731440 Chr.7: 151056786-151056786 on Build GRCh38
rs375655625 Chr.7: 151056844-151056844 on Build GRCh38
rs554159803 Chr.7: 151056647-151056647 on Build GRCh38
rs1022629799 Chr.7: 151056727-151056727 on Build GRCh38
rs746425237 Chr.7: 151056887-151056887 on Build GRCh38
rs759883551 Chr.7: 151056766-151056766 on Build GRCh38
rs777878897 Chr.7: 151056738-151056738 on Build GRCh38
rs755933600 Chr.7: 151056866-151056866 on Build GRCh38
rs781027595 Chr.7: 151056699-151056699 on Build GRCh38
rs202081553 Chr.7: 151056724-151056724 on Build GRCh38
rs776043437 Chr.7: 151056890-151056890 on Build GRCh38
rs779010975 Chr.7: 151056625-151056625 on Build GRCh38
rs747044107 Chr.7: 151056880-151056880 on Build GRCh38
rs763865593 Chr.7: 151056829-151056829 on Build GRCh38
rs1043815260 Chr.7: 151056405-151056405 on Build GRCh38
rs1017390795 Chr.7: 151056601-151056601 on Build GRCh38
rs879019629 Chr.7: 151056812-151056812 on Build GRCh38
rs190447046 Chr.7: 151056422-151056422 on Build GRCh38
rs371590011 Chr.7: 151056713-151056713 on Build GRCh38
rs368886325 Chr.7: 151056587-151056587 on Build GRCh38
rs369848852 Chr.7: 151056885-151056885 on Build GRCh38
rs1056106626 Chr.7: 151056474-151056474 on Build GRCh38
rs764876563 Chr.7: 151056658-151056658 on Build GRCh38
rs182778117 Chr.7: 151056657-151056657 on Build GRCh38
rs546262888 Chr.7: 151056665-151056665 on Build GRCh38
rs775128740 Chr.7: 151056799-151056800 on Build GRCh38
rs777007468 Chr.7: 151056761-151056761 on Build GRCh38
rs974834525 Chr.7: 151056886-151056886 on Build GRCh38
rs369446117 Chr.7: 151056745-151056745 on Build GRCh38
rs561605555 Chr.7: 151056879-151056879 on Build GRCh38
rs1036372253 Chr.7: 151056834-151056834 on Build GRCh38
rs768156497 Chr.7: 151056845-151056848 on Build GRCh38
rs752895368 Chr.7: 151056867-151056867 on Build GRCh38
rs749022122 Chr.7: 151056441-151056441 on Build GRCh38
rs551975005 Chr.7: 151056896-151056896 on Build GRCh38
rs979667306 Chr.7: 151056721-151056721 on Build GRCh38
rs748438212 Chr.7: 151056715-151056715 on Build GRCh38
rs981723295 Chr.7: 151056503-151056503 on Build GRCh38
rs768158895 Chr.7: 151056527-151056527 on Build GRCh38
rs370736023 Chr.7: 151056822-151056822 on Build GRCh38
rs752407282 Chr.7: 151056664-151056664 on Build GRCh38
rs2069450 Chr.7: 151056682-151056682 on Build GRCh38
rs200729612 Chr.7: 151056582-151056582 on Build GRCh38
rs548283397 Chr.7: 151056452-151056452 on Build GRCh38
rs905557030 Chr.7: 151056774-151056774 on Build GRCh38
rs373690199 Chr.7: 151056837-151056837 on Build GRCh38
rs879089142 Chr.7: 151056811-151056811 on Build GRCh38
rs531146226 Chr.7: 151056775-151056775 on Build GRCh38
rs775274881 Chr.7: 151056784-151056784 on Build GRCh38
rs753704210 Chr.7: 151056526-151056526 on Build GRCh38
rs761211846 Chr.7: 151056655-151056655 on Build GRCh38
rs1003109582 Chr.7: 151056777-151056777 on Build GRCh38
rs374660006 Chr.7: 151056889-151056889 on Build GRCh38
rs750386255 Chr.7: 151056861-151056861 on Build GRCh38
rs757172305 Chr.7: 151056692-151056692 on Build GRCh38
rs879174395 Chr.7: 151056740-151056740 on Build GRCh38
rs950885523 Chr.7: 151056853-151056853 on Build GRCh38
rs767434391 Chr.7: 151056858-151056858 on Build GRCh38
rs143488987 Chr.7: 151056521-151056521 on Build GRCh38
rs748993694 Chr.7: 151056819-151056828 on Build GRCh38
rs376160817 Chr.7: 151056726-151056726 on Build GRCh38
rs904853918 Chr.7: 151056414-151056414 on Build GRCh38
rs774164033 Chr.7: 151056914-151056914 on Build GRCh38
rs777790756 Chr.7: 151056551-151056551 on Build GRCh38
rs374501108 Chr.7: 151056577-151056577 on Build GRCh38
rs759673695 Chr.7: 151056524-151056524 on Build GRCh38
rs781093521 Chr.7: 151056571-151056571 on Build GRCh38
rs534137476 Chr.7: 151056540-151056540 on Build GRCh38
rs755876265 Chr.7: 151056711-151056711 on Build GRCh38
rs371217173 Chr.7: 151056559-151056559 on Build GRCh38
rs868359935 Chr.7: 151056686-151056686 on Build GRCh38
rs970294640 Chr.7: 151056472-151056472 on Build GRCh38
rs771429450 Chr.7: 151056757-151056757 on Build GRCh38
rs751076575 Chr.7: 151056831-151056831 on Build GRCh38
rs750169157 Chr.7: 151056705-151056705 on Build GRCh38
rs757297273 Chr.7: 151056882-151056882 on Build GRCh38
rs950973528 Chr.7: 151056519-151056519 on Build GRCh38
rs773975565 Chr.7: 151056539-151056539 on Build GRCh38
rs754863632 Chr.7: 151056616-151056616 on Build GRCh38
rs896335945 Chr.7: 151056509-151056509 on Build GRCh38
rs773706186 Chr.7: 151056651-151056651 on Build GRCh38
rs1016313768 Chr.7: 151056877-151056877 on Build GRCh38
rs749704291 Chr.7: 151056894-151056894 on Build GRCh38
rs769672901 Chr.7: 151056558-151056558 on Build GRCh38
rs758565818 Chr.7: 151056869-151056869 on Build GRCh38
rs2069451 Chr.7: 151056667-151056667 on Build GRCh38
rs954726581 Chr.7: 151056915-151056915 on Build GRCh38
rs975504693 Chr.7: 151056648-151056648 on Build GRCh38
rs777004002 Chr.7: 151056656-151056656 on Build GRCh38

View Full Product Details