Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144452622-144453196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPK1
Entrez Gene ID
27010
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
HTPK1, PP20, THMD5
UniGene
Hs.660232
Gene Chromosome Location
Chr.7: 144451941-144836424 on Build GRCh38
Transcripts
NM_001042482.1 NM_022445.3 XM_005249970.1 XM_011516031.1 XM_011516032.2 XM_011516033.2 XM_011516034.2 XM_011516037.2 XM_011516039.2 XM_011516041.2 XM_011516043.1 XM_011516044.1 XM_011516047.2 XM_011516048.1 XM_011516050.1 XM_017011970.1 XM_017011971.1 XM_017011973.1 XM_017011974.1 XM_017011975.1 XM_017011976.1 XM_017011977.1 XM_017011978.1 XM_017011979.1 XM_017011980.1 XM_017011981.1 XM_017011982.1 XM_017011983.1 XM_017011984.1 XM_017011985.1 XM_017011986.1

SNP Detail

SNP ID Chromosome Location
rs571219388 Chr.7: 144452859-144452859 on Build GRCh38
rs972257196 Chr.7: 144452985-144452985 on Build GRCh38
rs750793171 Chr.7: 144452874-144452874 on Build GRCh38
rs943170146 Chr.7: 144453074-144453074 on Build GRCh38
rs987125201 Chr.7: 144453066-144453066 on Build GRCh38
rs569988300 Chr.7: 144453071-144453072 on Build GRCh38
rs898630354 Chr.7: 144452956-144452956 on Build GRCh38
rs553169447 Chr.7: 144452764-144452764 on Build GRCh38
rs979460059 Chr.7: 144452851-144452851 on Build GRCh38
rs994547737 Chr.7: 144452729-144452729 on Build GRCh38
rs918887139 Chr.7: 144452686-144452686 on Build GRCh38
rs868680569 Chr.7: 144452744-144452744 on Build GRCh38
rs372934267 Chr.7: 144452894-144452894 on Build GRCh38
rs910284305 Chr.7: 144453071-144453071 on Build GRCh38
rs142197214 Chr.7: 144453084-144453084 on Build GRCh38
rs149043256 Chr.7: 144452886-144452886 on Build GRCh38
rs1004570203 Chr.7: 144452779-144452779 on Build GRCh38
rs950404537 Chr.7: 144452735-144452735 on Build GRCh38
rs963883195 Chr.7: 144452973-144452973 on Build GRCh38
rs978919363 Chr.7: 144453081-144453081 on Build GRCh38
rs919525300 Chr.7: 144453026-144453026 on Build GRCh38
rs1053000138 Chr.7: 144453137-144453137 on Build GRCh38
rs577998625 Chr.7: 144452655-144452655 on Build GRCh38
rs763780658 Chr.7: 144452695-144452695 on Build GRCh38
rs1005074969 Chr.7: 144453055-144453055 on Build GRCh38
rs969528971 Chr.7: 144452904-144452904 on Build GRCh38
rs564032610 Chr.7: 144452722-144452722 on Build GRCh38
rs193069601 Chr.7: 144453082-144453082 on Build GRCh38
rs996015431 Chr.7: 144453072-144453073 on Build GRCh38
rs542340947 Chr.7: 144452902-144452902 on Build GRCh38
rs61594332 Chr.7: 144452781-144452781 on Build GRCh38
rs149912480 Chr.7: 144452716-144452716 on Build GRCh38
rs561505864 Chr.7: 144452843-144452843 on Build GRCh38
rs1016471913 Chr.7: 144452944-144452944 on Build GRCh38
rs536179132 Chr.7: 144452998-144452998 on Build GRCh38
rs532154714 Chr.7: 144452885-144452885 on Build GRCh38
rs762859688 Chr.7: 144453003-144453003 on Build GRCh38
rs879453505 Chr.7: 144452790-144452790 on Build GRCh38
rs200480908 Chr.7: 144452916-144452916 on Build GRCh38
rs569140093 Chr.7: 144452903-144452903 on Build GRCh38
rs528675328 Chr.7: 144452844-144452844 on Build GRCh38
rs549792609 Chr.7: 144453017-144453018 on Build GRCh38
rs954355523 Chr.7: 144453040-144453040 on Build GRCh38
rs769920534 Chr.7: 144452900-144452900 on Build GRCh38
rs776297285 Chr.7: 144452711-144452711 on Build GRCh38
rs965983419 Chr.7: 144452791-144452791 on Build GRCh38
rs895759001 Chr.7: 144453162-144453162 on Build GRCh38
rs190747119 Chr.7: 144452731-144452731 on Build GRCh38
rs552066831 Chr.7: 144452766-144452766 on Build GRCh38
rs144891862 Chr.7: 144452854-144452854 on Build GRCh38
rs140880403 Chr.7: 144452916-144452917 on Build GRCh38
rs886170908 Chr.7: 144453045-144453045 on Build GRCh38

View Full Product Details