Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28890805-28891301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMBOX1
Entrez Gene ID
79618
Gene Name
homeobox containing 1
Gene Aliases
HNF1LA, HOT1, PBHNF
UniGene
Hs.563560
Gene Chromosome Location
Chr.8: 28890370-29064939 on Build GRCh38
Transcripts
NM_001135726.2 NM_001324383.1 NM_001324384.1 NM_001324385.1 NM_001324386.1 NM_001324387.1 NM_001324389.1 NM_001324390.1 NM_001324391.1 NM_001324392.1 NM_001324393.1 NM_001324394.1 NM_001324395.1 NM_024567.4 NR_136757.1 NR_136758.1 XM_005273635.1 XM_005273641.4 XM_006716398.2 XM_011544642.2 XM_017013825.1

SNP Detail

SNP ID Chromosome Location
rs958958456 Chr.8: 28890924-28890924 on Build GRCh38
rs889154310 Chr.8: 28891037-28891037 on Build GRCh38
rs144108418 Chr.8: 28890995-28890995 on Build GRCh38
rs1009300670 Chr.8: 28890875-28890875 on Build GRCh38
rs1021445356 Chr.8: 28890945-28890945 on Build GRCh38
rs532903284 Chr.8: 28891016-28891016 on Build GRCh38
rs900403797 Chr.8: 28890864-28890864 on Build GRCh38
rs907597907 Chr.8: 28891056-28891056 on Build GRCh38
rs931856179 Chr.8: 28891129-28891129 on Build GRCh38
rs967659044 Chr.8: 28890919-28890919 on Build GRCh38
rs1008263240 Chr.8: 28890932-28890932 on Build GRCh38
rs1009829554 Chr.8: 28891125-28891125 on Build GRCh38
rs927511813 Chr.8: 28890923-28890923 on Build GRCh38
rs867199257 Chr.8: 28890942-28890942 on Build GRCh38
rs574238709 Chr.8: 28891119-28891122 on Build GRCh38
rs546650386 Chr.8: 28890944-28890944 on Build GRCh38
rs146476780 Chr.8: 28891250-28891250 on Build GRCh38
rs930706597 Chr.8: 28891274-28891274 on Build GRCh38
rs983165661 Chr.8: 28891050-28891050 on Build GRCh38
rs1051624287 Chr.8: 28891138-28891138 on Build GRCh38
rs554843242 Chr.8: 28891211-28891211 on Build GRCh38
rs1021979276 Chr.8: 28891128-28891128 on Build GRCh38
rs941806562 Chr.8: 28891065-28891065 on Build GRCh38
rs941844707 Chr.8: 28890856-28890856 on Build GRCh38
rs970789865 Chr.8: 28891042-28891042 on Build GRCh38
rs1025267722 Chr.8: 28891018-28891018 on Build GRCh38
rs529789064 Chr.8: 28890911-28890911 on Build GRCh38
rs1037519534 Chr.8: 28890863-28890863 on Build GRCh38
rs902568970 Chr.8: 28891175-28891175 on Build GRCh38
rs902622465 Chr.8: 28890958-28890958 on Build GRCh38
rs1024990281 Chr.8: 28891240-28891240 on Build GRCh38
rs903617480 Chr.8: 28891136-28891136 on Build GRCh38
rs993523190 Chr.8: 28890926-28890926 on Build GRCh38
rs1032527810 Chr.8: 28890988-28890988 on Build GRCh38
rs188847766 Chr.8: 28891272-28891272 on Build GRCh38
rs568576220 Chr.8: 28891232-28891232 on Build GRCh38
rs1001420439 Chr.8: 28890970-28890970 on Build GRCh38
rs996062673 Chr.8: 28890865-28890865 on Build GRCh38
rs1001794625 Chr.8: 28891155-28891155 on Build GRCh38
rs944020112 Chr.8: 28891147-28891147 on Build GRCh38
rs552905684 Chr.8: 28891087-28891087 on Build GRCh38
rs930590525 Chr.8: 28891027-28891027 on Build GRCh38
rs371954919 Chr.8: 28890881-28890881 on Build GRCh38
rs1050425581 Chr.8: 28891032-28891032 on Build GRCh38
rs891124685 Chr.8: 28890922-28890922 on Build GRCh38
rs183389546 Chr.8: 28891166-28891166 on Build GRCh38
rs777301591 Chr.8: 28891219-28891219 on Build GRCh38
rs570418250 Chr.8: 28891069-28891069 on Build GRCh38
rs1000960554 Chr.8: 28891200-28891200 on Build GRCh38
rs921776736 Chr.8: 28891094-28891094 on Build GRCh38
rs895311034 Chr.8: 28891265-28891265 on Build GRCh38
rs746542295 Chr.8: 28891024-28891027 on Build GRCh38
rs907649946 Chr.8: 28890844-28890844 on Build GRCh38
rs940881948 Chr.8: 28890973-28890973 on Build GRCh38
rs780519636 Chr.8: 28891040-28891040 on Build GRCh38
rs569408021 Chr.8: 28891139-28891139 on Build GRCh38

View Full Product Details