Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103885090-103885687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIMS2
Entrez Gene ID
9699
Gene Name
regulating synaptic membrane exocytosis 2
Gene Aliases
OBOE, RAB3IP3, RIM2
UniGene
Hs.655271
Gene Chromosome Location
Chr.8: 103500026-104254430 on Build GRCh38
Transcripts
NM_001100117.2 NM_001282881.1 NM_014677.4 XM_005251106.3 XM_005251107.3 XM_011517395.2 XM_011517398.2 XM_017014006.1 XM_017014007.1 XM_017014008.1 XM_017014009.1 XM_017014010.1 XM_017014011.1 XM_017014012.1 XM_017014013.1 XM_017014014.1 XM_017014015.1 XM_017014016.1 XM_017014017.1 XM_017014018.1 XM_017014019.1 XM_017014020.1 XM_017014021.1 XM_017014022.1 XM_017014023.1 XM_017014024.1 XM_017014025.1 XM_017014026.1 XM_017014027.1 XM_017014028.1 XM_017014029.1 XM_017014030.1 XM_017014031.1 XM_017014032.1 XM_017014033.1 XM_017014034.1 XM_017014035.1 XM_017014036.1 XM_017014037.1 XM_017014038.1 XM_017014039.1 XM_017014040.1 XM_017014041.1 XM_017014042.1 XM_017014043.1 XM_017014044.1 XM_017014045.1 XM_017014046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6111602 c.352G>T p.A118S Chr.8: 103885617-103885617 on Build GRCh38
1453746 c.126C>T p.T42T Chr.8: 103885391-103885391 on Build GRCh38
4563547 c.1087G>A p.E363K Chr.8: 103885578-103885578 on Build GRCh38
5408511 c.709C>T p.P237S Chr.8: 103885308-103885308 on Build GRCh38
1453758 c.447T>C p.R149R Chr.8: 103885622-103885622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767296516 Chr.8: 103885626-103885626 on Build GRCh38
rs963041852 Chr.8: 103885155-103885155 on Build GRCh38
rs754649407 Chr.8: 103885510-103885510 on Build GRCh38
rs768211118 Chr.8: 103885305-103885305 on Build GRCh38
rs762927342 Chr.8: 103885617-103885617 on Build GRCh38
rs750171549 Chr.8: 103885392-103885392 on Build GRCh38
rs72681354 Chr.8: 103885163-103885163 on Build GRCh38
rs757064818 Chr.8: 103885284-103885284 on Build GRCh38
rs149422371 Chr.8: 103885222-103885222 on Build GRCh38
rs769876416 Chr.8: 103885600-103885600 on Build GRCh38
rs374551478 Chr.8: 103885340-103885340 on Build GRCh38
rs747383172 Chr.8: 103885660-103885660 on Build GRCh38
rs748229985 Chr.8: 103885588-103885588 on Build GRCh38
rs766129122 Chr.8: 103885620-103885620 on Build GRCh38
rs200264682 Chr.8: 103885578-103885578 on Build GRCh38
rs768762604 Chr.8: 103885454-103885454 on Build GRCh38
rs377220871 Chr.8: 103885258-103885258 on Build GRCh38
rs761807288 Chr.8: 103885150-103885150 on Build GRCh38
rs1005391456 Chr.8: 103885162-103885162 on Build GRCh38
rs562430054 Chr.8: 103885575-103885575 on Build GRCh38
rs190433281 Chr.8: 103885314-103885314 on Build GRCh38
rs780330474 Chr.8: 103885356-103885359 on Build GRCh38
rs760675914 Chr.8: 103885338-103885338 on Build GRCh38
rs780920782 Chr.8: 103885523-103885523 on Build GRCh38
rs749854815 Chr.8: 103885429-103885429 on Build GRCh38
rs756231924 Chr.8: 103885356-103885357 on Build GRCh38
rs764120867 Chr.8: 103885341-103885341 on Build GRCh38
rs758937074 Chr.8: 103885652-103885652 on Build GRCh38
rs747295124 Chr.8: 103885568-103885568 on Build GRCh38
rs758150011 Chr.8: 103885394-103885394 on Build GRCh38
rs373254707 Chr.8: 103885289-103885289 on Build GRCh38
rs118008121 Chr.8: 103885248-103885248 on Build GRCh38
rs377313890 Chr.8: 103885502-103885502 on Build GRCh38
rs772415017 Chr.8: 103885434-103885434 on Build GRCh38
rs3824117 Chr.8: 103885637-103885637 on Build GRCh38
rs754559656 Chr.8: 103885411-103885411 on Build GRCh38
rs17854256 Chr.8: 103885432-103885432 on Build GRCh38
rs76903489 Chr.8: 103885433-103885433 on Build GRCh38
rs761796354 Chr.8: 103885469-103885469 on Build GRCh38
rs929660257 Chr.8: 103885412-103885412 on Build GRCh38
rs750304080 Chr.8: 103885493-103885493 on Build GRCh38
rs34681663 Chr.8: 103885391-103885391 on Build GRCh38
rs780830800 Chr.8: 103885414-103885414 on Build GRCh38
rs146399863 Chr.8: 103885378-103885378 on Build GRCh38
rs745573912 Chr.8: 103885291-103885291 on Build GRCh38
rs752714528 Chr.8: 103885250-103885250 on Build GRCh38
rs560220838 Chr.8: 103885252-103885252 on Build GRCh38
rs779291464 Chr.8: 103885462-103885462 on Build GRCh38
rs751603406 Chr.8: 103885129-103885129 on Build GRCh38
rs755631676 Chr.8: 103885530-103885530 on Build GRCh38
rs762684104 Chr.8: 103885494-103885494 on Build GRCh38
rs752335393 Chr.8: 103885628-103885628 on Build GRCh38
rs750932195 Chr.8: 103885645-103885645 on Build GRCh38
rs774284988 Chr.8: 103885623-103885623 on Build GRCh38
rs560445160 Chr.8: 103885538-103885538 on Build GRCh38
rs369996192 Chr.8: 103885331-103885331 on Build GRCh38
rs1018659910 Chr.8: 103885625-103885625 on Build GRCh38
rs751181714 Chr.8: 103885409-103885409 on Build GRCh38
rs950334846 Chr.8: 103885608-103885608 on Build GRCh38
rs912736593 Chr.8: 103885213-103885213 on Build GRCh38
rs1035954784 Chr.8: 103885123-103885123 on Build GRCh38
rs551220305 Chr.8: 103885161-103885161 on Build GRCh38
rs35925435 Chr.8: 103885459-103885459 on Build GRCh38
rs559067260 Chr.8: 103885407-103885407 on Build GRCh38
rs139492859 Chr.8: 103885397-103885397 on Build GRCh38
rs200726457 Chr.8: 103885558-103885558 on Build GRCh38
rs530434507 Chr.8: 103885200-103885200 on Build GRCh38
rs760862293 Chr.8: 103885448-103885448 on Build GRCh38
rs138898746 Chr.8: 103885142-103885142 on Build GRCh38
rs888236827 Chr.8: 103885149-103885149 on Build GRCh38
rs759873512 Chr.8: 103885320-103885320 on Build GRCh38
rs200878422 Chr.8: 103885504-103885504 on Build GRCh38
rs369246895 Chr.8: 103885273-103885273 on Build GRCh38
rs374557980 Chr.8: 103885444-103885444 on Build GRCh38
rs535730678 Chr.8: 103885364-103885364 on Build GRCh38
rs779295630 Chr.8: 103885536-103885536 on Build GRCh38
rs369681508 Chr.8: 103885663-103885663 on Build GRCh38
rs185138315 Chr.8: 103885238-103885238 on Build GRCh38
rs746511179 Chr.8: 103885293-103885293 on Build GRCh38
rs551774397 Chr.8: 103885632-103885632 on Build GRCh38
rs201322998 Chr.8: 103885311-103885311 on Build GRCh38
rs987214509 Chr.8: 103885205-103885205 on Build GRCh38
rs373874945 Chr.8: 103885395-103885395 on Build GRCh38
rs780336959 Chr.8: 103885659-103885659 on Build GRCh38
rs570226693 Chr.8: 103885281-103885281 on Build GRCh38
rs781422909 Chr.8: 103885667-103885667 on Build GRCh38
rs998881190 Chr.8: 103885118-103885118 on Build GRCh38
rs371773136 Chr.8: 103885438-103885438 on Build GRCh38
rs565414312 Chr.8: 103885127-103885127 on Build GRCh38
rs1013263136 Chr.8: 103885135-103885135 on Build GRCh38
rs200152573 Chr.8: 103885559-103885559 on Build GRCh38
rs749766002 Chr.8: 103885313-103885313 on Build GRCh38
rs757981777 Chr.8: 103885292-103885292 on Build GRCh38
rs773044369 Chr.8: 103885603-103885603 on Build GRCh38
rs918914147 Chr.8: 103885194-103885194 on Build GRCh38
rs371608826 Chr.8: 103885487-103885487 on Build GRCh38
rs144225573 Chr.8: 103885624-103885624 on Build GRCh38

View Full Product Details