Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32972789-32973357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APTX
Entrez Gene ID
54840
Gene Name
aprataxin
Gene Aliases
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
UniGene
Hs.20158
Gene Chromosome Location
Chr.9: 32883872-33025129 on Build GRCh38
Transcripts
NM_001195248.1 NM_001195249.1 NM_001195250.1 NM_001195251.1 NM_001195252.1 NM_001195254.1 NM_175069.2 NM_175073.2 NR_036576.1 NR_036577.1 NR_036578.1 NR_036579.1 XM_006716791.3 XM_006716792.2 XM_011517938.1 XM_011517939.2 XM_017014831.1 XM_017014832.1 XM_017014833.1 XM_017014834.1 XM_017014835.1 XM_017014836.1 XM_017014837.1 XM_017014838.1

SNP Detail

SNP ID Chromosome Location
rs191436643 Chr.9: 32973008-32973008 on Build GRCh38
rs138490250 Chr.9: 32973044-32973044 on Build GRCh38
rs896086786 Chr.9: 32973164-32973164 on Build GRCh38
rs898096564 Chr.9: 32973251-32973251 on Build GRCh38
rs367715736 Chr.9: 32972913-32972913 on Build GRCh38
rs994414292 Chr.9: 32973185-32973185 on Build GRCh38
rs532688347 Chr.9: 32973205-32973205 on Build GRCh38
rs1040449763 Chr.9: 32972902-32972902 on Build GRCh38
rs897953806 Chr.9: 32972988-32972988 on Build GRCh38
rs1006923029 Chr.9: 32972977-32972977 on Build GRCh38
rs1025632277 Chr.9: 32972944-32972944 on Build GRCh38
rs781025624 Chr.9: 32973153-32973153 on Build GRCh38
rs778040753 Chr.9: 32973076-32973076 on Build GRCh38
rs563055303 Chr.9: 32973188-32973188 on Build GRCh38
rs931949925 Chr.9: 32972930-32972930 on Build GRCh38
rs750884035 Chr.9: 32973134-32973134 on Build GRCh38
rs984204614 Chr.9: 32972958-32972958 on Build GRCh38
rs756510915 Chr.9: 32973186-32973186 on Build GRCh38
rs1018372403 Chr.9: 32973235-32973235 on Build GRCh38
rs964100401 Chr.9: 32973253-32973253 on Build GRCh38
rs866139662 Chr.9: 32973220-32973220 on Build GRCh38
rs780379330 Chr.9: 32972955-32972955 on Build GRCh38
rs762629781 Chr.9: 32973256-32973256 on Build GRCh38
rs991102979 Chr.9: 32973030-32973030 on Build GRCh38
rs745612904 Chr.9: 32973204-32973204 on Build GRCh38
rs914473152 Chr.9: 32972818-32972818 on Build GRCh38
rs973159126 Chr.9: 32972929-32972929 on Build GRCh38
rs922527511 Chr.9: 32973020-32973020 on Build GRCh38
rs568783017 Chr.9: 32973283-32973283 on Build GRCh38
rs111430445 Chr.9: 32972892-32972892 on Build GRCh38
rs746922480 Chr.9: 32973115-32973115 on Build GRCh38
rs750414984 Chr.9: 32972826-32972826 on Build GRCh38
rs1035380211 Chr.9: 32973017-32973017 on Build GRCh38
rs574067595 Chr.9: 32972996-32972996 on Build GRCh38
rs886063857 Chr.9: 32973135-32973135 on Build GRCh38
rs923335458 Chr.9: 32973092-32973092 on Build GRCh38
rs563932179 Chr.9: 32972982-32972982 on Build GRCh38
rs1010598831 Chr.9: 32972830-32972830 on Build GRCh38
rs901945278 Chr.9: 32972926-32972926 on Build GRCh38
rs760824792 Chr.9: 32973313-32973313 on Build GRCh38
rs113638548 Chr.9: 32973311-32973311 on Build GRCh38
rs946507857 Chr.9: 32973055-32973055 on Build GRCh38
rs1057272176 Chr.9: 32973135-32973135 on Build GRCh38
rs143023388 Chr.9: 32973061-32973061 on Build GRCh38
rs779137120 Chr.9: 32973053-32973053 on Build GRCh38
rs766841162 Chr.9: 32973324-32973324 on Build GRCh38
rs761257788 Chr.9: 32973319-32973319 on Build GRCh38
rs1012662246 Chr.9: 32973282-32973282 on Build GRCh38
rs775131687 Chr.9: 32972888-32972888 on Build GRCh38
rs960296817 Chr.9: 32972916-32972916 on Build GRCh38
rs754188583 Chr.9: 32973022-32973022 on Build GRCh38
rs758547802 Chr.9: 32973070-32973070 on Build GRCh38
rs762743887 Chr.9: 32972960-32972960 on Build GRCh38
rs911089390 Chr.9: 32972970-32972970 on Build GRCh38
rs977003268 Chr.9: 32973296-32973296 on Build GRCh38
rs559954042 Chr.9: 32973042-32973042 on Build GRCh38
rs530692252 Chr.9: 32972862-32972862 on Build GRCh38
rs1026107483 Chr.9: 32973202-32973202 on Build GRCh38
rs759273556 Chr.9: 32972861-32972861 on Build GRCh38
rs545625482 Chr.9: 32972893-32972893 on Build GRCh38
rs924560935 Chr.9: 32973048-32973048 on Build GRCh38
rs770855079 Chr.9: 32973151-32973151 on Build GRCh38
rs548870588 Chr.9: 32973250-32973250 on Build GRCh38
rs894220671 Chr.9: 32973277-32973277 on Build GRCh38
rs761289876 Chr.9: 32973002-32973002 on Build GRCh38
rs768175241 Chr.9: 32972826-32972833 on Build GRCh38
rs1057260827 Chr.9: 32973270-32973270 on Build GRCh38

View Full Product Details