Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21861800-21862370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTAP
Entrez Gene ID
4507
Gene Name
methylthioadenosine phosphorylase
Gene Aliases
BDMF, DMSFH, DMSMFH, HEL-249, LGMBF, MSAP, c86fus
UniGene
Hs.193268
Gene Chromosome Location
Chr.9: 21802636-21865971 on Build GRCh38
Transcripts
NM_002451.3

SNP Detail

SNP ID Chromosome Location
rs150414950 Chr.9: 21862342-21862342 on Build GRCh38
rs140440868 Chr.9: 21862317-21862317 on Build GRCh38
rs556788598 Chr.9: 21861908-21861908 on Build GRCh38
rs78195856 Chr.9: 21862273-21862273 on Build GRCh38
rs529366343 Chr.9: 21861871-21861871 on Build GRCh38
rs983997486 Chr.9: 21862041-21862041 on Build GRCh38
rs749100975 Chr.9: 21862283-21862283 on Build GRCh38
rs767735914 Chr.9: 21862168-21862168 on Build GRCh38
rs368840669 Chr.9: 21862062-21862062 on Build GRCh38
rs567794387 Chr.9: 21861870-21861870 on Build GRCh38
rs1034877529 Chr.9: 21861916-21861916 on Build GRCh38
rs113189258 Chr.9: 21862295-21862295 on Build GRCh38
rs752812608 Chr.9: 21862259-21862259 on Build GRCh38
rs746554053 Chr.9: 21862058-21862058 on Build GRCh38
rs1029509814 Chr.9: 21861903-21861903 on Build GRCh38
rs1012071118 Chr.9: 21862272-21862273 on Build GRCh38
rs747905013 Chr.9: 21862040-21862040 on Build GRCh38
rs999293435 Chr.9: 21862233-21862234 on Build GRCh38
rs760196201 Chr.9: 21861958-21861958 on Build GRCh38
rs754086497 Chr.9: 21862029-21862029 on Build GRCh38
rs76352569 Chr.9: 21861878-21861878 on Build GRCh38
rs755040797 Chr.9: 21862116-21862116 on Build GRCh38
rs966493376 Chr.9: 21861904-21861905 on Build GRCh38
rs751850371 Chr.9: 21861987-21861987 on Build GRCh38
rs752127232 Chr.9: 21862070-21862070 on Build GRCh38
rs896322861 Chr.9: 21862265-21862265 on Build GRCh38
rs762318246 Chr.9: 21861997-21861997 on Build GRCh38
rs750498270 Chr.9: 21862010-21862010 on Build GRCh38
rs924313816 Chr.9: 21862166-21862166 on Build GRCh38
rs1039301225 Chr.9: 21862133-21862133 on Build GRCh38
rs775627882 Chr.9: 21861982-21861982 on Build GRCh38
rs553057304 Chr.9: 21862031-21862031 on Build GRCh38
rs548118434 Chr.9: 21861872-21861872 on Build GRCh38
rs561546447 Chr.9: 21862322-21862322 on Build GRCh38
rs948748414 Chr.9: 21861834-21861834 on Build GRCh38
rs367743939 Chr.9: 21862019-21862019 on Build GRCh38
rs772873530 Chr.9: 21861943-21861943 on Build GRCh38
rs374742223 Chr.9: 21861962-21861962 on Build GRCh38
rs1034758965 Chr.9: 21862264-21862264 on Build GRCh38
rs929426361 Chr.9: 21862036-21862036 on Build GRCh38
rs3208969 Chr.9: 21862271-21862271 on Build GRCh38
rs750844480 Chr.9: 21862032-21862032 on Build GRCh38
rs15735 Chr.9: 21862272-21862272 on Build GRCh38
rs1046656156 Chr.9: 21861828-21861828 on Build GRCh38
rs572882647 Chr.9: 21862119-21862119 on Build GRCh38
rs369244310 Chr.9: 21862095-21862095 on Build GRCh38
rs924241686 Chr.9: 21862341-21862341 on Build GRCh38
rs1039332205 Chr.9: 21861902-21861903 on Build GRCh38
rs796221993 Chr.9: 21861914-21861914 on Build GRCh38
rs67222036 Chr.9: 21861904-21861904 on Build GRCh38
rs897835622 Chr.9: 21862268-21862268 on Build GRCh38
rs1050644886 Chr.9: 21862235-21862235 on Build GRCh38
rs541896368 Chr.9: 21862128-21862128 on Build GRCh38
rs1022830835 Chr.9: 21862307-21862307 on Build GRCh38
rs772079978 Chr.9: 21861940-21861940 on Build GRCh38
rs766558829 Chr.9: 21862021-21862021 on Build GRCh38
rs770532013 Chr.9: 21861961-21861961 on Build GRCh38
rs763289310 Chr.9: 21861976-21861976 on Build GRCh38
rs772942966 Chr.9: 21861903-21861904 on Build GRCh38
rs1040343065 Chr.9: 21862264-21862264 on Build GRCh38
rs370160422 Chr.9: 21861898-21861898 on Build GRCh38
rs199655299 Chr.9: 21861935-21861935 on Build GRCh38
rs971166805 Chr.9: 21862072-21862072 on Build GRCh38
rs1010957926 Chr.9: 21861892-21861892 on Build GRCh38
rs564730053 Chr.9: 21862248-21862248 on Build GRCh38
rs946887125 Chr.9: 21862234-21862234 on Build GRCh38
rs11356405 Chr.9: 21861904-21861904 on Build GRCh38
rs949719864 Chr.9: 21861826-21861826 on Build GRCh38
rs971134192 Chr.9: 21862316-21862316 on Build GRCh38
rs951886861 Chr.9: 21862293-21862293 on Build GRCh38
rs555725472 Chr.9: 21862132-21862132 on Build GRCh38
rs375541397 Chr.9: 21862051-21862051 on Build GRCh38
rs988381158 Chr.9: 21862343-21862343 on Build GRCh38
rs370980390 Chr.9: 21862044-21862044 on Build GRCh38
rs765097494 Chr.9: 21861952-21861953 on Build GRCh38
rs1008839390 Chr.9: 21861914-21861914 on Build GRCh38
rs1052615203 Chr.9: 21861874-21861874 on Build GRCh38
rs886063782 Chr.9: 21862272-21862272 on Build GRCh38
rs973438374 Chr.9: 21862012-21862016 on Build GRCh38
rs763706581 Chr.9: 21861886-21861886 on Build GRCh38
rs201476509 Chr.9: 21861932-21861932 on Build GRCh38
rs370748314 Chr.9: 21861936-21861936 on Build GRCh38
rs915374568 Chr.9: 21862224-21862224 on Build GRCh38
rs575907260 Chr.9: 21862206-21862206 on Build GRCh38
rs146104023 Chr.9: 21861861-21861861 on Build GRCh38
rs544925331 Chr.9: 21862226-21862226 on Build GRCh38
rs951972481 Chr.9: 21861994-21861994 on Build GRCh38
rs778158868 Chr.9: 21861905-21861905 on Build GRCh38
rs375813031 Chr.9: 21862093-21862093 on Build GRCh38
rs974915284 Chr.9: 21862345-21862345 on Build GRCh38
rs770464905 Chr.9: 21862059-21862059 on Build GRCh38
rs538983344 Chr.9: 21862156-21862156 on Build GRCh38
rs767960835 Chr.9: 21862005-21862005 on Build GRCh38
rs111350457 Chr.9: 21862195-21862195 on Build GRCh38

View Full Product Details