Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15452824-15453338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAPC3
Entrez Gene ID
6619
Gene Name
small nuclear RNA activating complex polypeptide 3
Gene Aliases
PTFbeta, SNAP50
UniGene
Hs.546299
Gene Chromosome Location
Chr.9: 15422696-15466751 on Build GRCh38
Transcripts
NM_001039697.1 XM_017015056.1 XM_017015057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1107394 c.920T>A p.F307Y Chr.9: 15453145-15453145 on Build GRCh38
6516979 c.839C>A p.S280Y Chr.9: 15453064-15453064 on Build GRCh38
1107395 c.960C>A p.V320V Chr.9: 15453185-15453185 on Build GRCh38
1107393 c.893T>C p.F298S Chr.9: 15453118-15453118 on Build GRCh38
3669941 c.961A>T p.I321F Chr.9: 15453186-15453186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140869103 Chr.9: 15453064-15453064 on Build GRCh38
rs368876645 Chr.9: 15453011-15453011 on Build GRCh38
rs138841606 Chr.9: 15453180-15453180 on Build GRCh38
rs768721525 Chr.9: 15453019-15453020 on Build GRCh38
rs1022279883 Chr.9: 15452975-15452975 on Build GRCh38
rs138821543 Chr.9: 15453003-15453003 on Build GRCh38
rs775769321 Chr.9: 15453257-15453257 on Build GRCh38
rs1056544746 Chr.9: 15452936-15452936 on Build GRCh38
rs768134375 Chr.9: 15453060-15453060 on Build GRCh38
rs753775849 Chr.9: 15453203-15453203 on Build GRCh38
rs34346482 Chr.9: 15453140-15453140 on Build GRCh38
rs763885983 Chr.9: 15452887-15452887 on Build GRCh38
rs766942909 Chr.9: 15452994-15452994 on Build GRCh38
rs746199116 Chr.9: 15453246-15453246 on Build GRCh38
rs865777319 Chr.9: 15453277-15453277 on Build GRCh38
rs1022122060 Chr.9: 15453036-15453036 on Build GRCh38
rs138667984 Chr.9: 15453122-15453122 on Build GRCh38
rs755221061 Chr.9: 15452902-15452902 on Build GRCh38
rs1036644836 Chr.9: 15452950-15452950 on Build GRCh38
rs760117089 Chr.9: 15453081-15453081 on Build GRCh38
rs201761191 Chr.9: 15453126-15453126 on Build GRCh38
rs369622830 Chr.9: 15453090-15453090 on Build GRCh38
rs765616120 Chr.9: 15453156-15453156 on Build GRCh38
rs745779619 Chr.9: 15453035-15453035 on Build GRCh38
rs1004542777 Chr.9: 15452939-15452939 on Build GRCh38
rs751777117 Chr.9: 15453109-15453109 on Build GRCh38
rs1012147409 Chr.9: 15453303-15453303 on Build GRCh38
rs745726291 Chr.9: 15453121-15453121 on Build GRCh38
rs537642122 Chr.9: 15453211-15453211 on Build GRCh38
rs200063794 Chr.9: 15453217-15453217 on Build GRCh38
rs995274328 Chr.9: 15453268-15453268 on Build GRCh38
rs150161706 Chr.9: 15453098-15453098 on Build GRCh38
rs200969359 Chr.9: 15453031-15453031 on Build GRCh38
rs939662074 Chr.9: 15452848-15452848 on Build GRCh38
rs568919388 Chr.9: 15452984-15452984 on Build GRCh38
rs550436449 Chr.9: 15452897-15452897 on Build GRCh38
rs777195600 Chr.9: 15453079-15453079 on Build GRCh38
rs759781006 Chr.9: 15453238-15453238 on Build GRCh38
rs767727295 Chr.9: 15453187-15453187 on Build GRCh38
rs41268953 Chr.9: 15453283-15453283 on Build GRCh38
rs370068912 Chr.9: 15453255-15453255 on Build GRCh38
rs905362680 Chr.9: 15452884-15452884 on Build GRCh38
rs560454794 Chr.9: 15453275-15453275 on Build GRCh38
rs773772545 Chr.9: 15453146-15453146 on Build GRCh38
rs371241908 Chr.9: 15453192-15453192 on Build GRCh38
rs752890362 Chr.9: 15453017-15453017 on Build GRCh38
rs370662940 Chr.9: 15453267-15453267 on Build GRCh38
rs776099512 Chr.9: 15453158-15453158 on Build GRCh38
rs146075837 Chr.9: 15453150-15453150 on Build GRCh38
rs1036004619 Chr.9: 15453247-15453247 on Build GRCh38
rs776948840 Chr.9: 15453237-15453238 on Build GRCh38
rs770889788 Chr.9: 15453201-15453205 on Build GRCh38
rs747168827 Chr.9: 15453180-15453180 on Build GRCh38
rs758487086 Chr.9: 15453030-15453030 on Build GRCh38
rs150182630 Chr.9: 15452891-15452891 on Build GRCh38
rs926877012 Chr.9: 15453298-15453298 on Build GRCh38
rs978014786 Chr.9: 15452860-15452860 on Build GRCh38
rs778483914 Chr.9: 15453212-15453212 on Build GRCh38
rs781002347 Chr.9: 15453033-15453033 on Build GRCh38
rs199652997 Chr.9: 15453049-15453049 on Build GRCh38
rs1009503190 Chr.9: 15452958-15452958 on Build GRCh38
rs907434943 Chr.9: 15452969-15452969 on Build GRCh38
rs1034491640 Chr.9: 15453007-15453007 on Build GRCh38
rs765396635 Chr.9: 15453013-15453013 on Build GRCh38
rs762014901 Chr.9: 15453186-15453186 on Build GRCh38
rs752711361 Chr.9: 15453083-15453083 on Build GRCh38
rs371410944 Chr.9: 15452990-15452990 on Build GRCh38
rs779813812 Chr.9: 15453201-15453201 on Build GRCh38
rs766830567 Chr.9: 15453078-15453078 on Build GRCh38
rs373358593 Chr.9: 15453059-15453059 on Build GRCh38
rs763412909 Chr.9: 15453159-15453159 on Build GRCh38
rs757522607 Chr.9: 15453116-15453116 on Build GRCh38
rs1003091433 Chr.9: 15453266-15453266 on Build GRCh38
rs758277850 Chr.9: 15452871-15452871 on Build GRCh38
rs549707374 Chr.9: 15452867-15452867 on Build GRCh38
rs377748371 Chr.9: 15453189-15453189 on Build GRCh38
rs780949047 Chr.9: 15453117-15453117 on Build GRCh38
rs200945273 Chr.9: 15453155-15453155 on Build GRCh38
rs774154578 Chr.9: 15453030-15453031 on Build GRCh38
rs764209608 Chr.9: 15453167-15453167 on Build GRCh38
rs531938124 Chr.9: 15452895-15452895 on Build GRCh38
rs930860162 Chr.9: 15453130-15453130 on Build GRCh38
rs374095519 Chr.9: 15453152-15453152 on Build GRCh38
rs769777195 Chr.9: 15453037-15453037 on Build GRCh38
rs923849617 Chr.9: 15452865-15452865 on Build GRCh38
rs545184407 Chr.9: 15453273-15453273 on Build GRCh38
rs534467864 Chr.9: 15453276-15453277 on Build GRCh38
rs764185788 Chr.9: 15453107-15453107 on Build GRCh38
rs201978945 Chr.9: 15453220-15453220 on Build GRCh38
rs371211653 Chr.9: 15452872-15452872 on Build GRCh38
rs980214702 Chr.9: 15453290-15453290 on Build GRCh38

View Full Product Details