Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179847346-179847730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOR1AIP2
Entrez Gene ID
163590
Gene Name
torsin 1A interacting protein 2
Gene Aliases
IFRG15, LULL1, NET9
UniGene
Hs.571797
Gene Chromosome Location
Chr.1: 179839967-179878614 on Build GRCh38
Transcripts
NM_001199260.1 NM_145034.4 XM_005244939.4 XM_017000474.1 XM_017000475.1 XM_017000476.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
900320 c.612G>T p.Q204H Chr.1: 179847578-179847578 on Build GRCh38
424712 c.648G>T p.W216C Chr.1: 179847542-179847542 on Build GRCh38
6459713 c.637A>G p.K213E Chr.1: 179847553-179847553 on Build GRCh38
2095633 c.619A>G p.M207V Chr.1: 179847571-179847571 on Build GRCh38
6348257 c.655+8C>A p.? Chr.1: 179847527-179847527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049158487 Chr.1: 179847439-179847439 on Build GRCh38
rs543014518 Chr.1: 179847674-179847674 on Build GRCh38
rs201615031 Chr.1: 179847566-179847566 on Build GRCh38
rs182200636 Chr.1: 179847434-179847434 on Build GRCh38
rs556555981 Chr.1: 179847398-179847398 on Build GRCh38
rs534970796 Chr.1: 179847462-179847462 on Build GRCh38
rs572259962 Chr.1: 179847530-179847530 on Build GRCh38
rs774344789 Chr.1: 179847662-179847662 on Build GRCh38
rs762006431 Chr.1: 179847677-179847677 on Build GRCh38
rs1049421623 Chr.1: 179847457-179847457 on Build GRCh38
rs767971966 Chr.1: 179847521-179847521 on Build GRCh38
rs531836572 Chr.1: 179847705-179847705 on Build GRCh38
rs887636036 Chr.1: 179847474-179847474 on Build GRCh38
rs547038981 Chr.1: 179847472-179847472 on Build GRCh38
rs373687923 Chr.1: 179847511-179847511 on Build GRCh38
rs745619830 Chr.1: 179847614-179847614 on Build GRCh38
rs757912562 Chr.1: 179847560-179847560 on Build GRCh38
rs768753246 Chr.1: 179847657-179847657 on Build GRCh38
rs570800292 Chr.1: 179847401-179847401 on Build GRCh38
rs940488457 Chr.1: 179847534-179847534 on Build GRCh38
rs376083463 Chr.1: 179847650-179847650 on Build GRCh38
rs912947921 Chr.1: 179847397-179847397 on Build GRCh38
rs981124333 Chr.1: 179847419-179847419 on Build GRCh38
rs377364518 Chr.1: 179847527-179847527 on Build GRCh38
rs752303472 Chr.1: 179847550-179847550 on Build GRCh38
rs75169009 Chr.1: 179847629-179847629 on Build GRCh38
rs528670266 Chr.1: 179847549-179847549 on Build GRCh38
rs764140758 Chr.1: 179847484-179847484 on Build GRCh38
rs769593556 Chr.1: 179847618-179847618 on Build GRCh38
rs949482246 Chr.1: 179847396-179847396 on Build GRCh38
rs752107487 Chr.1: 179847510-179847510 on Build GRCh38
rs560798679 Chr.1: 179847571-179847571 on Build GRCh38
rs370462599 Chr.1: 179847577-179847577 on Build GRCh38
rs768100278 Chr.1: 179847678-179847678 on Build GRCh38
rs1037085242 Chr.1: 179847557-179847557 on Build GRCh38
rs191003966 Chr.1: 179847428-179847428 on Build GRCh38
rs7531195 Chr.1: 179847681-179847681 on Build GRCh38
rs770899401 Chr.1: 179847573-179847573 on Build GRCh38

View Full Product Details