Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186444271-186444869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724919
Entrez Gene ID
102724919
Gene Name
uncharacterized LOC102724919
Gene Aliases
-
UniGene
Hs.576493
Gene Chromosome Location
Chr.1: 186435161-186470291 on Build GRCh38
Transcripts
NR_126002.1 NM_002597.4 NM_022576.3 XM_011509603.2

Gene Symbol
PDC
Entrez Gene ID
5132
Gene Name
phosducin
Gene Aliases
MEKA, PHD, PhLOP, PhLP
UniGene
Hs.654381
Gene Chromosome Location
Chr.1: 186443566-186461108 on Build GRCh38
Transcripts
NR_126002.1 NM_002597.4 NM_022576.3 XM_011509603.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3400117 c.376A>C p.T126P Chr.1: 186444344-186444344 on Build GRCh38
241090 c.314T>G p.L105R Chr.1: 186444406-186444406 on Build GRCh38
6040106 c.271C>T p.R91C Chr.1: 186444449-186444449 on Build GRCh38
3480313 c.416A>C p.H139P Chr.1: 186444304-186444304 on Build GRCh38
6452927 c.272G>A p.R91H Chr.1: 186444448-186444448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027759868 Chr.1: 186444612-186444612 on Build GRCh38
rs776644840 Chr.1: 186444405-186444405 on Build GRCh38
rs772521962 Chr.1: 186444387-186444387 on Build GRCh38
rs1033295518 Chr.1: 186444783-186444783 on Build GRCh38
rs981716026 Chr.1: 186444778-186444778 on Build GRCh38
rs1016118644 Chr.1: 186444431-186444431 on Build GRCh38
rs534217507 Chr.1: 186444545-186444545 on Build GRCh38
rs759603122 Chr.1: 186444416-186444416 on Build GRCh38
rs777177713 Chr.1: 186444538-186444538 on Build GRCh38
rs1047456596 Chr.1: 186444649-186444649 on Build GRCh38
rs755673582 Chr.1: 186444358-186444358 on Build GRCh38
rs567786815 Chr.1: 186444440-186444440 on Build GRCh38
rs1030143841 Chr.1: 186444796-186444796 on Build GRCh38
rs779362996 Chr.1: 186444366-186444366 on Build GRCh38
rs374625049 Chr.1: 186444306-186444308 on Build GRCh38
rs368302911 Chr.1: 186444439-186444439 on Build GRCh38
rs574801745 Chr.1: 186444684-186444684 on Build GRCh38
rs954685791 Chr.1: 186444808-186444808 on Build GRCh38
rs755889902 Chr.1: 186444600-186444600 on Build GRCh38
rs754106832 Chr.1: 186444305-186444305 on Build GRCh38
rs778685072 Chr.1: 186444379-186444379 on Build GRCh38
rs376556470 Chr.1: 186444449-186444449 on Build GRCh38
rs748825871 Chr.1: 186444372-186444372 on Build GRCh38
rs781147574 Chr.1: 186444349-186444349 on Build GRCh38
rs970620869 Chr.1: 186444740-186444740 on Build GRCh38
rs560085874 Chr.1: 186444730-186444730 on Build GRCh38
rs549444291 Chr.1: 186444376-186444376 on Build GRCh38
rs986904651 Chr.1: 186444811-186444811 on Build GRCh38
rs959093941 Chr.1: 186444820-186444820 on Build GRCh38
rs750290018 Chr.1: 186444357-186444357 on Build GRCh38
rs950650474 Chr.1: 186444417-186444417 on Build GRCh38
rs1006123938 Chr.1: 186444427-186444427 on Build GRCh38
rs1015429794 Chr.1: 186444672-186444672 on Build GRCh38
rs147271545 Chr.1: 186444578-186444578 on Build GRCh38
rs986809948 Chr.1: 186444523-186444523 on Build GRCh38
rs771051369 Chr.1: 186444394-186444394 on Build GRCh38
rs766317270 Chr.1: 186444302-186444302 on Build GRCh38
rs554818827 Chr.1: 186444465-186444465 on Build GRCh38
rs540013359 Chr.1: 186444586-186444586 on Build GRCh38
rs752886840 Chr.1: 186444432-186444432 on Build GRCh38
rs996417210 Chr.1: 186444763-186444763 on Build GRCh38
rs573528658 Chr.1: 186444524-186444524 on Build GRCh38
rs527622804 Chr.1: 186444766-186444766 on Build GRCh38
rs914817885 Chr.1: 186444563-186444563 on Build GRCh38
rs542008768 Chr.1: 186444720-186444720 on Build GRCh38
rs761026786 Chr.1: 186444393-186444393 on Build GRCh38
rs780025844 Chr.1: 186444454-186444454 on Build GRCh38
rs763460137 Chr.1: 186444577-186444577 on Build GRCh38
rs990438208 Chr.1: 186444562-186444562 on Build GRCh38
rs900385200 Chr.1: 186444758-186444758 on Build GRCh38
rs747751839 Chr.1: 186444471-186444471 on Build GRCh38
rs938416198 Chr.1: 186444627-186444627 on Build GRCh38
rs559027814 Chr.1: 186444559-186444559 on Build GRCh38
rs143358201 Chr.1: 186444326-186444326 on Build GRCh38
rs377431091 Chr.1: 186444532-186444532 on Build GRCh38
rs763105074 Chr.1: 186444547-186444547 on Build GRCh38
rs1025912754 Chr.1: 186444396-186444396 on Build GRCh38
rs544591417 Chr.1: 186444581-186444581 on Build GRCh38
rs989633217 Chr.1: 186444837-186444837 on Build GRCh38
rs367961929 Chr.1: 186444474-186444474 on Build GRCh38
rs747397949 Chr.1: 186444525-186444525 on Build GRCh38
rs372016238 Chr.1: 186444448-186444448 on Build GRCh38
rs377504699 Chr.1: 186444841-186444841 on Build GRCh38
rs370382715 Chr.1: 186444544-186444544 on Build GRCh38
rs886131179 Chr.1: 186444687-186444687 on Build GRCh38
rs140036725 Chr.1: 186444327-186444327 on Build GRCh38
rs765495996 Chr.1: 186444422-186444422 on Build GRCh38
rs141951413 Chr.1: 186444462-186444462 on Build GRCh38
rs1002955548 Chr.1: 186444390-186444390 on Build GRCh38
rs199564768 Chr.1: 186444298-186444298 on Build GRCh38
rs753499101 Chr.1: 186444399-186444399 on Build GRCh38
rs764371279 Chr.1: 186444557-186444557 on Build GRCh38
rs748559830 Chr.1: 186444380-186444380 on Build GRCh38
rs751418186 Chr.1: 186444334-186444334 on Build GRCh38
rs757024258 Chr.1: 186444340-186444340 on Build GRCh38
rs745988803 Chr.1: 186444543-186444543 on Build GRCh38
rs995316100 Chr.1: 186444601-186444602 on Build GRCh38
rs11812050 Chr.1: 186444582-186444582 on Build GRCh38

View Full Product Details