Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113766228-113766827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHS1
Entrez Gene ID
79949
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C10orf81, HEL185, bA211N11.2
UniGene
Hs.228320
Gene Chromosome Location
Chr.10: 113751262-113782433 on Build GRCh38
Transcripts
NM_001193434.1 NM_001193435.1 NM_024889.4 NM_182601.1 XM_005270162.2 XM_005270163.2 XM_011540171.1 XM_011540172.1 XM_011540173.1 XM_011540174.2 XM_011540175.1 XM_011540176.1 XM_017016666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5675929 c.193A>G p.I65V Chr.10: 113766705-113766705 on Build GRCh38
3434238 c.82C>T p.Q28* Chr.10: 113766482-113766482 on Build GRCh38
6750063 c.11G>T p.G4V Chr.10: 113766411-113766411 on Build GRCh38
5527495 c.104C>T p.S35F Chr.10: 113766616-113766616 on Build GRCh38
3434242 c.135G>A p.K45K Chr.10: 113766647-113766647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777115891 Chr.10: 113766500-113766503 on Build GRCh38
rs771259353 Chr.10: 113766383-113766383 on Build GRCh38
rs201994714 Chr.10: 113766628-113766628 on Build GRCh38
rs751051793 Chr.10: 113766701-113766701 on Build GRCh38
rs959655245 Chr.10: 113766535-113766535 on Build GRCh38
rs530083408 Chr.10: 113766374-113766384 on Build GRCh38
rs771403432 Chr.10: 113766477-113766477 on Build GRCh38
rs770120866 Chr.10: 113766621-113766621 on Build GRCh38
rs554312868 Chr.10: 113766352-113766352 on Build GRCh38
rs776032199 Chr.10: 113766627-113766627 on Build GRCh38
rs866231177 Chr.10: 113766434-113766434 on Build GRCh38
rs1038733493 Chr.10: 113766374-113766374 on Build GRCh38
rs764284515 Chr.10: 113766401-113766401 on Build GRCh38
rs757952747 Chr.10: 113766597-113766597 on Build GRCh38
rs748213535 Chr.10: 113766750-113766750 on Build GRCh38
rs779871328 Chr.10: 113766413-113766413 on Build GRCh38
rs780889365 Chr.10: 113766706-113766706 on Build GRCh38
rs756881826 Chr.10: 113766702-113766702 on Build GRCh38
rs767668372 Chr.10: 113766662-113766662 on Build GRCh38
rs761109019 Chr.10: 113766365-113766365 on Build GRCh38
rs759372909 Chr.10: 113766687-113766687 on Build GRCh38
rs76119388 Chr.10: 113766634-113766634 on Build GRCh38
rs139556061 Chr.10: 113766681-113766681 on Build GRCh38
rs372022414 Chr.10: 113766689-113766689 on Build GRCh38
rs778898814 Chr.10: 113766590-113766590 on Build GRCh38
rs1051425429 Chr.10: 113766721-113766721 on Build GRCh38
rs368298647 Chr.10: 113766584-113766584 on Build GRCh38
rs760143019 Chr.10: 113766383-113766383 on Build GRCh38
rs992141510 Chr.10: 113766537-113766537 on Build GRCh38
rs917982207 Chr.10: 113766272-113766272 on Build GRCh38
rs761886088 Chr.10: 113766659-113766659 on Build GRCh38
rs539606658 Chr.10: 113766749-113766749 on Build GRCh38
rs780717276 Chr.10: 113766611-113766611 on Build GRCh38
rs769171705 Chr.10: 113766629-113766629 on Build GRCh38
rs755871581 Chr.10: 113766410-113766410 on Build GRCh38
rs35665302 Chr.10: 113766739-113766740 on Build GRCh38
rs374567672 Chr.10: 113766524-113766524 on Build GRCh38
rs777091294 Chr.10: 113766485-113766485 on Build GRCh38
rs925435476 Chr.10: 113766647-113766647 on Build GRCh38
rs10885500 Chr.10: 113766418-113766418 on Build GRCh38
rs780231244 Chr.10: 113766728-113766728 on Build GRCh38
rs376862334 Chr.10: 113766376-113766376 on Build GRCh38
rs992654584 Chr.10: 113766258-113766258 on Build GRCh38
rs758365422 Chr.10: 113766696-113766696 on Build GRCh38
rs759853886 Chr.10: 113766504-113766504 on Build GRCh38
rs777565644 Chr.10: 113766467-113766467 on Build GRCh38
rs771767388 Chr.10: 113766444-113766444 on Build GRCh38
rs751758019 Chr.10: 113766542-113766542 on Build GRCh38
rs201870731 Chr.10: 113766442-113766442 on Build GRCh38
rs752751763 Chr.10: 113766390-113766390 on Build GRCh38
rs778438704 Chr.10: 113766374-113766380 on Build GRCh38
rs763160661 Chr.10: 113766591-113766591 on Build GRCh38
rs764982919 Chr.10: 113766688-113766688 on Build GRCh38
rs774373925 Chr.10: 113766657-113766657 on Build GRCh38
rs77423862 Chr.10: 113766736-113766736 on Build GRCh38
rs753602913 Chr.10: 113766576-113766576 on Build GRCh38
rs966764781 Chr.10: 113766618-113766618 on Build GRCh38
rs772111794 Chr.10: 113766751-113766751 on Build GRCh38
rs777248966 Chr.10: 113766379-113766379 on Build GRCh38
rs925343783 Chr.10: 113766782-113766782 on Build GRCh38
rs936616438 Chr.10: 113766785-113766785 on Build GRCh38
rs925259481 Chr.10: 113766342-113766342 on Build GRCh38
rs1001865562 Chr.10: 113766501-113766501 on Build GRCh38
rs925548149 Chr.10: 113766340-113766340 on Build GRCh38
rs78924486 Chr.10: 113766294-113766294 on Build GRCh38
rs747832364 Chr.10: 113766383-113766387 on Build GRCh38
rs764196624 Chr.10: 113766538-113766538 on Build GRCh38
rs568995573 Chr.10: 113766725-113766725 on Build GRCh38
rs528751511 Chr.10: 113766421-113766421 on Build GRCh38
rs751843864 Chr.10: 113766406-113766406 on Build GRCh38
rs745544795 Chr.10: 113766720-113766720 on Build GRCh38
rs766388946 Chr.10: 113766761-113766761 on Build GRCh38
rs900176407 Chr.10: 113766387-113766387 on Build GRCh38
rs747636509 Chr.10: 113766443-113766443 on Build GRCh38
rs746544362 Chr.10: 113766603-113766603 on Build GRCh38
rs570061060 Chr.10: 113766472-113766472 on Build GRCh38
rs764162126 Chr.10: 113766699-113766699 on Build GRCh38
rs374743627 Chr.10: 113766562-113766562 on Build GRCh38
rs978241201 Chr.10: 113766319-113766319 on Build GRCh38
rs746227928 Chr.10: 113766619-113766619 on Build GRCh38
rs546633935 Chr.10: 113766474-113766474 on Build GRCh38
rs371539556 Chr.10: 113766526-113766526 on Build GRCh38
rs187837701 Chr.10: 113766697-113766697 on Build GRCh38
rs749683612 Chr.10: 113766729-113766729 on Build GRCh38
rs769083876 Chr.10: 113766746-113766746 on Build GRCh38
rs773454306 Chr.10: 113766758-113766758 on Build GRCh38
rs770671858 Chr.10: 113766607-113766607 on Build GRCh38
rs141560911 Chr.10: 113766786-113766786 on Build GRCh38
rs866785625 Chr.10: 113766716-113766716 on Build GRCh38
rs758417507 Chr.10: 113766400-113766400 on Build GRCh38
rs796870612 Chr.10: 113766374-113766387 on Build GRCh38

View Full Product Details