Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83458482-83459011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG2
Entrez Gene ID
1740
Gene Name
discs large MAGUK scaffold protein 2
Gene Aliases
PPP1R58, PSD-93, PSD93, chapsyn-110
UniGene
Hs.367656
Gene Chromosome Location
Chr.11: 83455009-85628534 on Build GRCh38
Transcripts
NM_001142699.1 NM_001142700.1 NM_001142702.1 NM_001206769.1 NM_001300983.1 NM_001364.3 XM_005273810.4 XM_005273811.4 XM_011544778.2 XM_011544780.2 XM_011544782.2 XM_011544788.2 XM_017017254.1 XM_017017255.1 XM_017017256.1 XM_017017257.1 XM_017017258.1 XM_017017259.1 XM_017017260.1 XM_017017261.1 XM_017017262.1 XM_017017263.1 XM_017017264.1 XM_017017265.1 XM_017017266.1 XM_017017267.1 XM_017017268.1 XM_017017269.1 XM_017017270.1 XM_017017271.1 XM_017017272.1 XM_017017273.1 XM_017017274.1 XM_017017275.1 XM_017017276.1 XM_017017277.1 XM_017017278.1 XM_017017279.1 XM_017017280.1 XM_017017281.1 XM_017017283.1 XM_017017284.1 XM_017017285.1 XM_017017286.1 XM_017017287.1 XM_017017288.1 XM_017017289.1 XM_017017290.1 XM_017017291.1 XM_017017292.1

SNP Detail

SNP ID Chromosome Location
rs761388635 Chr.11: 83458714-83458714 on Build GRCh38
rs1034006198 Chr.11: 83458518-83458518 on Build GRCh38
rs1056792137 Chr.11: 83458608-83458608 on Build GRCh38
rs553287928 Chr.11: 83458812-83458812 on Build GRCh38
rs376319642 Chr.11: 83458788-83458788 on Build GRCh38
rs181613797 Chr.11: 83458885-83458885 on Build GRCh38
rs937830412 Chr.11: 83458740-83458740 on Build GRCh38
rs779515451 Chr.11: 83458876-83458876 on Build GRCh38
rs543039665 Chr.11: 83458867-83458867 on Build GRCh38
rs910065305 Chr.11: 83458953-83458953 on Build GRCh38
rs144977727 Chr.11: 83458573-83458573 on Build GRCh38
rs980492015 Chr.11: 83458970-83458970 on Build GRCh38
rs377519016 Chr.11: 83458888-83458889 on Build GRCh38
rs902188542 Chr.11: 83458956-83458956 on Build GRCh38
rs1020595732 Chr.11: 83458546-83458546 on Build GRCh38
rs896432017 Chr.11: 83458540-83458540 on Build GRCh38
rs796155840 Chr.11: 83458676-83458676 on Build GRCh38
rs531787993 Chr.11: 83458894-83458894 on Build GRCh38
rs931347761 Chr.11: 83458712-83458712 on Build GRCh38
rs964401577 Chr.11: 83458783-83458783 on Build GRCh38
rs547165938 Chr.11: 83458885-83458886 on Build GRCh38
rs962411858 Chr.11: 83458954-83458954 on Build GRCh38
rs912791921 Chr.11: 83458662-83458662 on Build GRCh38
rs892422707 Chr.11: 83458619-83458619 on Build GRCh38
rs1002083989 Chr.11: 83458509-83458509 on Build GRCh38
rs753812991 Chr.11: 83458891-83458891 on Build GRCh38
rs550969698 Chr.11: 83458541-83458541 on Build GRCh38
rs185928380 Chr.11: 83458895-83458895 on Build GRCh38
rs926275052 Chr.11: 83458974-83458974 on Build GRCh38
rs998233591 Chr.11: 83458811-83458811 on Build GRCh38
rs765591215 Chr.11: 83458630-83458630 on Build GRCh38
rs1043609628 Chr.11: 83458663-83458663 on Build GRCh38
rs572512701 Chr.11: 83458817-83458817 on Build GRCh38
rs918601731 Chr.11: 83458705-83458705 on Build GRCh38
rs1032432453 Chr.11: 83458616-83458616 on Build GRCh38
rs1044528190 Chr.11: 83458621-83458621 on Build GRCh38
rs982897984 Chr.11: 83458913-83458913 on Build GRCh38
rs1048507512 Chr.11: 83458726-83458726 on Build GRCh38
rs753095044 Chr.11: 83458704-83458704 on Build GRCh38
rs569688307 Chr.11: 83458520-83458520 on Build GRCh38
rs115255322 Chr.11: 83458566-83458566 on Build GRCh38
rs954350745 Chr.11: 83458623-83458623 on Build GRCh38
rs1032042859 Chr.11: 83458982-83458982 on Build GRCh38
rs189266356 Chr.11: 83458729-83458729 on Build GRCh38
rs939242749 Chr.11: 83458607-83458607 on Build GRCh38
rs541346503 Chr.11: 83458735-83458735 on Build GRCh38
rs555503313 Chr.11: 83458756-83458756 on Build GRCh38
rs1018251457 Chr.11: 83458772-83458772 on Build GRCh38

View Full Product Details