Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101916145-101916684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL5
Entrez Gene ID
253935
Gene Name
angiopoietin like 5
Gene Aliases
-
UniGene
Hs.318370
Gene Chromosome Location
Chr.11: 101890674-101916522 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1 NM_020802.3 XM_005271627.4

Gene Symbol
CEP126
Entrez Gene ID
57562
Gene Name
centrosomal protein 126
Gene Aliases
KIAA1377
UniGene
Hs.156352
Gene Chromosome Location
Chr.11: 101915015-102001065 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1 NM_020802.3 XM_005271627.4

SNP Detail

SNP ID Chromosome Location
rs575707572 Chr.11: 101916338-101916339 on Build GRCh38
rs543612945 Chr.11: 101916545-101916545 on Build GRCh38
rs563531977 Chr.11: 101916551-101916551 on Build GRCh38
rs1026956824 Chr.11: 101916484-101916484 on Build GRCh38
rs1018702024 Chr.11: 101916230-101916230 on Build GRCh38
rs912497646 Chr.11: 101916594-101916594 on Build GRCh38
rs750275259 Chr.11: 101916233-101916233 on Build GRCh38
rs552052657 Chr.11: 101916535-101916535 on Build GRCh38
rs1057376830 Chr.11: 101916501-101916501 on Build GRCh38
rs189826086 Chr.11: 101916253-101916253 on Build GRCh38
rs988042558 Chr.11: 101916583-101916583 on Build GRCh38
rs550797004 Chr.11: 101916280-101916280 on Build GRCh38
rs1012933571 Chr.11: 101916522-101916522 on Build GRCh38
rs747141141 Chr.11: 101916345-101916345 on Build GRCh38
rs981346873 Chr.11: 101916326-101916326 on Build GRCh38
rs1049076566 Chr.11: 101916444-101916444 on Build GRCh38
rs767912607 Chr.11: 101916324-101916324 on Build GRCh38
rs931037827 Chr.11: 101916439-101916439 on Build GRCh38
rs1027051851 Chr.11: 101916263-101916263 on Build GRCh38
rs983701402 Chr.11: 101916300-101916300 on Build GRCh38
rs1008931467 Chr.11: 101916418-101916418 on Build GRCh38
rs927194681 Chr.11: 101916545-101916545 on Build GRCh38
rs921027337 Chr.11: 101916434-101916434 on Build GRCh38
rs146735388 Chr.11: 101916401-101916401 on Build GRCh38
rs975058003 Chr.11: 101916650-101916650 on Build GRCh38
rs557027338 Chr.11: 101916281-101916282 on Build GRCh38
rs765064659 Chr.11: 101916164-101916164 on Build GRCh38
rs988013203 Chr.11: 101916394-101916394 on Build GRCh38
rs140601271 Chr.11: 101916412-101916412 on Build GRCh38
rs187787609 Chr.11: 101916502-101916502 on Build GRCh38
rs991592784 Chr.11: 101916178-101916178 on Build GRCh38
rs965238403 Chr.11: 101916622-101916622 on Build GRCh38
rs971246608 Chr.11: 101916503-101916503 on Build GRCh38
rs1032627120 Chr.11: 101916576-101916576 on Build GRCh38
rs974195535 Chr.11: 101916255-101916255 on Build GRCh38
rs887275162 Chr.11: 101916483-101916483 on Build GRCh38
rs7105709 Chr.11: 101916381-101916381 on Build GRCh38
rs144341338 Chr.11: 101916566-101916566 on Build GRCh38
rs1008443164 Chr.11: 101916191-101916191 on Build GRCh38
rs571334534 Chr.11: 101916223-101916223 on Build GRCh38
rs899858488 Chr.11: 101916423-101916423 on Build GRCh38
rs185020257 Chr.11: 101916206-101916206 on Build GRCh38
rs907674545 Chr.11: 101916310-101916310 on Build GRCh38
rs995491871 Chr.11: 101916451-101916451 on Build GRCh38
rs891390573 Chr.11: 101916170-101916170 on Build GRCh38
rs925103865 Chr.11: 101916369-101916369 on Build GRCh38
rs551173423 Chr.11: 101916175-101916175 on Build GRCh38
rs926994597 Chr.11: 101916327-101916327 on Build GRCh38
rs183114499 Chr.11: 101916498-101916498 on Build GRCh38
rs762655451 Chr.11: 101916257-101916257 on Build GRCh38
rs1054162795 Chr.11: 101916364-101916364 on Build GRCh38
rs1039978523 Chr.11: 101916421-101916421 on Build GRCh38
rs567525194 Chr.11: 101916294-101916294 on Build GRCh38
rs956527658 Chr.11: 101916579-101916579 on Build GRCh38
rs914200084 Chr.11: 101916647-101916647 on Build GRCh38
rs542891009 Chr.11: 101916641-101916641 on Build GRCh38
rs912261271 Chr.11: 101916399-101916399 on Build GRCh38
rs554606250 Chr.11: 101916504-101916504 on Build GRCh38

View Full Product Details