Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94828328-94828764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
UniGene
Hs.483423
Gene Chromosome Location
Chr.1: 94820342-94895247 on Build GRCh38
Transcripts
NM_001114106.2 NM_001258340.1 NM_001258341.1 NM_001258342.1 NM_001258343.1 NM_001301079.1 NM_152369.4 XM_005270440.2 XM_005270441.2 XM_005270442.1 XM_011540632.2 XM_011540633.2 XM_011540635.2 XM_011540636.2 XM_017000262.1 XM_017000263.1 XM_017000264.1 XM_017000265.1 XM_017000266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7417958 c.290A>T p.Y97F Chr.1: 94828511-94828511 on Build GRCh38
7423166 c.292A>T p.S98C Chr.1: 94828513-94828513 on Build GRCh38
5922107 c.467C>T p.P156L Chr.1: 94828544-94828544 on Build GRCh38
682632 c.355G>T p.V119F Chr.1: 94828576-94828576 on Build GRCh38
6782336 c.473C>A p.A158E Chr.1: 94828550-94828550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991611469 Chr.1: 94828453-94828453 on Build GRCh38
rs764500400 Chr.1: 94828600-94828600 on Build GRCh38
rs993305144 Chr.1: 94828708-94828708 on Build GRCh38
rs958687734 Chr.1: 94828441-94828441 on Build GRCh38
rs763372073 Chr.1: 94828492-94828492 on Build GRCh38
rs749853879 Chr.1: 94828381-94828381 on Build GRCh38
rs2640065 Chr.1: 94828533-94828533 on Build GRCh38
rs749192614 Chr.1: 94828540-94828540 on Build GRCh38
rs952028423 Chr.1: 94828409-94828409 on Build GRCh38
rs757758535 Chr.1: 94828507-94828507 on Build GRCh38
rs780974110 Chr.1: 94828508-94828508 on Build GRCh38
rs771116283 Chr.1: 94828622-94828622 on Build GRCh38
rs142252804 Chr.1: 94828434-94828434 on Build GRCh38
rs1054133355 Chr.1: 94828554-94828554 on Build GRCh38
rs1035286843 Chr.1: 94828715-94828715 on Build GRCh38
rs1054182141 Chr.1: 94828633-94828633 on Build GRCh38
rs1005570271 Chr.1: 94828417-94828417 on Build GRCh38
rs763147379 Chr.1: 94828594-94828594 on Build GRCh38
rs1040124980 Chr.1: 94828655-94828655 on Build GRCh38
rs918586202 Chr.1: 94828527-94828527 on Build GRCh38
rs750031769 Chr.1: 94828635-94828635 on Build GRCh38
rs1035211187 Chr.1: 94828396-94828396 on Build GRCh38
rs768465394 Chr.1: 94828555-94828555 on Build GRCh38
rs759588872 Chr.1: 94828488-94828488 on Build GRCh38
rs1019947411 Chr.1: 94828380-94828380 on Build GRCh38
rs147302745 Chr.1: 94828564-94828564 on Build GRCh38
rs762223039 Chr.1: 94828626-94828626 on Build GRCh38
rs375150243 Chr.1: 94828720-94828720 on Build GRCh38
rs1041804814 Chr.1: 94828693-94828693 on Build GRCh38
rs1012425175 Chr.1: 94828728-94828728 on Build GRCh38
rs768028821 Chr.1: 94828629-94828629 on Build GRCh38
rs189869104 Chr.1: 94828701-94828701 on Build GRCh38
rs867273011 Chr.1: 94828385-94828385 on Build GRCh38
rs201060535 Chr.1: 94828524-94828524 on Build GRCh38
rs370619931 Chr.1: 94828694-94828694 on Build GRCh38
rs1010324415 Chr.1: 94828359-94828359 on Build GRCh38
rs567925128 Chr.1: 94828703-94828703 on Build GRCh38
rs1003786167 Chr.1: 94828394-94828394 on Build GRCh38
rs750125599 Chr.1: 94828511-94828511 on Build GRCh38
rs182030456 Chr.1: 94828427-94828427 on Build GRCh38
rs759706729 Chr.1: 94828575-94828575 on Build GRCh38
rs774031932 Chr.1: 94828467-94828467 on Build GRCh38
rs368126326 Chr.1: 94828489-94828489 on Build GRCh38
rs544904106 Chr.1: 94828389-94828389 on Build GRCh38
rs564808523 Chr.1: 94828418-94828418 on Build GRCh38
rs887704541 Chr.1: 94828721-94828721 on Build GRCh38
rs986662069 Chr.1: 94828549-94828549 on Build GRCh38
rs1033598605 Chr.1: 94828730-94828730 on Build GRCh38
rs751955532 Chr.1: 94828504-94828504 on Build GRCh38
rs747906373 Chr.1: 94828567-94828567 on Build GRCh38
rs137975014 Chr.1: 94828491-94828491 on Build GRCh38
rs375424161 Chr.1: 94828664-94828664 on Build GRCh38
rs901725660 Chr.1: 94828354-94828354 on Build GRCh38
rs752165773 Chr.1: 94828609-94828609 on Build GRCh38
rs374271106 Chr.1: 94828477-94828477 on Build GRCh38
rs761655206 Chr.1: 94828470-94828470 on Build GRCh38
rs72958566 Chr.1: 94828709-94828709 on Build GRCh38
rs764458954 Chr.1: 94828493-94828493 on Build GRCh38
rs993274200 Chr.1: 94828387-94828387 on Build GRCh38
rs115641784 Chr.1: 94828731-94828731 on Build GRCh38
rs945809522 Chr.1: 94828691-94828691 on Build GRCh38
rs749217419 Chr.1: 94828449-94828449 on Build GRCh38
rs113953918 Chr.1: 94828593-94828593 on Build GRCh38
rs530127822 Chr.1: 94828569-94828569 on Build GRCh38
rs368445194 Chr.1: 94828486-94828486 on Build GRCh38
rs373005954 Chr.1: 94828579-94828579 on Build GRCh38
rs768432364 Chr.1: 94828452-94828452 on Build GRCh38
rs776783466 Chr.1: 94828570-94828570 on Build GRCh38
rs150374603 Chr.1: 94828724-94828724 on Build GRCh38

View Full Product Details