Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 163336505-163336971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUF2
Entrez Gene ID
83540
Gene Name
NUF2, NDC80 kinetochore complex component
Gene Aliases
CDCA1, CT106, NUF2R
UniGene
Hs.651950
Gene Chromosome Location
Chr.1: 163321933-163355764 on Build GRCh38
Transcripts
NM_031423.3 NM_145697.2 XM_011510036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6427567 c.430C>T p.Q144* Chr.1: 163336843-163336843 on Build GRCh38
7380675 c.409A>T p.T137S Chr.1: 163336822-163336822 on Build GRCh38
1668172 c.347G>T p.R116L Chr.1: 163336760-163336760 on Build GRCh38
7094108 c.360T>C p.F120F Chr.1: 163336773-163336773 on Build GRCh38
1335830 c.342delA p.K115fs*7 Chr.1: 163336755-163336755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201079271 Chr.1: 163336760-163336760 on Build GRCh38
rs201463150 Chr.1: 163336817-163336817 on Build GRCh38
rs781627921 Chr.1: 163336769-163336769 on Build GRCh38
rs148215962 Chr.1: 163336824-163336824 on Build GRCh38
rs373198630 Chr.1: 163336816-163336816 on Build GRCh38
rs748734566 Chr.1: 163336656-163336656 on Build GRCh38
rs767765876 Chr.1: 163336874-163336874 on Build GRCh38
rs1013633046 Chr.1: 163336887-163336887 on Build GRCh38
rs200729694 Chr.1: 163336872-163336872 on Build GRCh38
rs7541849 Chr.1: 163336641-163336641 on Build GRCh38
rs541211806 Chr.1: 163336675-163336675 on Build GRCh38
rs2055747 Chr.1: 163336580-163336580 on Build GRCh38
rs777922553 Chr.1: 163336900-163336900 on Build GRCh38
rs777797338 Chr.1: 163336792-163336792 on Build GRCh38
rs111965879 Chr.1: 163336606-163336606 on Build GRCh38
rs753099534 Chr.1: 163336877-163336877 on Build GRCh38
rs57753183 Chr.1: 163336733-163336733 on Build GRCh38
rs147917145 Chr.1: 163336804-163336804 on Build GRCh38
rs568246569 Chr.1: 163336907-163336907 on Build GRCh38
rs928429826 Chr.1: 163336558-163336558 on Build GRCh38
rs752225577 Chr.1: 163336714-163336714 on Build GRCh38
rs534017905 Chr.1: 163336935-163336935 on Build GRCh38
rs752134080 Chr.1: 163336765-163336765 on Build GRCh38
rs753469318 Chr.1: 163336759-163336759 on Build GRCh38
rs776262779 Chr.1: 163336849-163336849 on Build GRCh38
rs1042986437 Chr.1: 163336623-163336623 on Build GRCh38
rs527852224 Chr.1: 163336674-163336674 on Build GRCh38
rs771708937 Chr.1: 163336821-163336821 on Build GRCh38
rs756305059 Chr.1: 163336878-163336878 on Build GRCh38
rs533250760 Chr.1: 163336738-163336738 on Build GRCh38
rs763685435 Chr.1: 163336739-163336739 on Build GRCh38
rs369947700 Chr.1: 163336830-163336830 on Build GRCh38
rs755226490 Chr.1: 163336768-163336768 on Build GRCh38
rs866674743 Chr.1: 163336842-163336842 on Build GRCh38
rs748354238 Chr.1: 163336770-163336770 on Build GRCh38
rs933053892 Chr.1: 163336660-163336660 on Build GRCh38
rs996215572 Chr.1: 163336721-163336721 on Build GRCh38
rs1052837178 Chr.1: 163336596-163336596 on Build GRCh38
rs142235785 Chr.1: 163336848-163336848 on Build GRCh38
rs749418049 Chr.1: 163336796-163336796 on Build GRCh38
rs369351164 Chr.1: 163336783-163336783 on Build GRCh38
rs573999775 Chr.1: 163336886-163336886 on Build GRCh38
rs141744491 Chr.1: 163336812-163336812 on Build GRCh38
rs759201924 Chr.1: 163336702-163336702 on Build GRCh38
rs374419828 Chr.1: 163336859-163336859 on Build GRCh38
rs867437216 Chr.1: 163336834-163336834 on Build GRCh38
rs751949686 Chr.1: 163336871-163336871 on Build GRCh38
rs760403092 Chr.1: 163336843-163336843 on Build GRCh38
rs767377057 Chr.1: 163336712-163336712 on Build GRCh38
rs891630456 Chr.1: 163336598-163336598 on Build GRCh38
rs764804886 Chr.1: 163336863-163336863 on Build GRCh38
rs556184314 Chr.1: 163336567-163336567 on Build GRCh38
rs554003740 Chr.1: 163336940-163336940 on Build GRCh38

View Full Product Details