Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 164508465-164508851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRB14
Entrez Gene ID
2888
Gene Name
growth factor receptor bound protein 14
Gene Aliases
-
UniGene
Hs.411881
Gene Chromosome Location
Chr.2: 164492406-164621850 on Build GRCh38
Transcripts
NM_001303422.1 NM_004490.2 XM_011511022.2 XM_017003899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4304882 c.641C>T p.P214L Chr.2: 164508767-164508767 on Build GRCh38
6674844 c.821C>T p.P274L Chr.2: 164508848-164508848 on Build GRCh38
4613543 c.891_892insAA p.H298fs*23 Chr.2: 164508777-164508778 on Build GRCh38
4642529 c.906T>A p.T302T Chr.2: 164508763-164508763 on Build GRCh38
7237953 c.635G>T p.G212V Chr.2: 164508773-164508773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775494148 Chr.2: 164508808-164508808 on Build GRCh38
rs199902676 Chr.2: 164508485-164508485 on Build GRCh38
rs755757465 Chr.2: 164508498-164508500 on Build GRCh38
rs200037089 Chr.2: 164508716-164508716 on Build GRCh38
rs760825186 Chr.2: 164508766-164508766 on Build GRCh38
rs376346366 Chr.2: 164508507-164508507 on Build GRCh38
rs987265941 Chr.2: 164508781-164508781 on Build GRCh38
rs776596903 Chr.2: 164508502-164508502 on Build GRCh38
rs781278687 Chr.2: 164508668-164508668 on Build GRCh38
rs1047973847 Chr.2: 164508567-164508567 on Build GRCh38
rs781054019 Chr.2: 164508786-164508786 on Build GRCh38
rs768520940 Chr.2: 164508498-164508498 on Build GRCh38
rs144729984 Chr.2: 164508758-164508758 on Build GRCh38
rs111945269 Chr.2: 164508525-164508525 on Build GRCh38
rs754663674 Chr.2: 164508782-164508782 on Build GRCh38
rs138627056 Chr.2: 164508768-164508768 on Build GRCh38
rs1035151378 Chr.2: 164508589-164508589 on Build GRCh38
rs771034359 Chr.2: 164508596-164508596 on Build GRCh38
rs374915204 Chr.2: 164508571-164508571 on Build GRCh38
rs750943071 Chr.2: 164508776-164508776 on Build GRCh38
rs762151159 Chr.2: 164508771-164508771 on Build GRCh38
rs574967446 Chr.2: 164508512-164508512 on Build GRCh38
rs761775375 Chr.2: 164508506-164508506 on Build GRCh38
rs869309375 Chr.2: 164508657-164508657 on Build GRCh38
rs1019727721 Chr.2: 164508670-164508670 on Build GRCh38
rs762217333 Chr.2: 164508826-164508826 on Build GRCh38
rs769761882 Chr.2: 164508710-164508710 on Build GRCh38
rs746771197 Chr.2: 164508494-164508494 on Build GRCh38
rs777473668 Chr.2: 164508517-164508517 on Build GRCh38
rs370149376 Chr.2: 164508541-164508541 on Build GRCh38
rs756354070 Chr.2: 164508528-164508528 on Build GRCh38
rs41268657 Chr.2: 164508539-164508539 on Build GRCh38
rs751127690 Chr.2: 164508779-164508779 on Build GRCh38
rs148544083 Chr.2: 164508796-164508796 on Build GRCh38
rs764406880 Chr.2: 164508531-164508531 on Build GRCh38
rs767745087 Chr.2: 164508515-164508515 on Build GRCh38
rs775580138 Chr.2: 164508760-164508760 on Build GRCh38
rs749142871 Chr.2: 164508801-164508801 on Build GRCh38
rs532552011 Chr.2: 164508698-164508698 on Build GRCh38
rs759636661 Chr.2: 164508718-164508718 on Build GRCh38
rs745896056 Chr.2: 164508817-164508817 on Build GRCh38
rs770864299 Chr.2: 164508803-164508803 on Build GRCh38
rs978110977 Chr.2: 164508711-164508711 on Build GRCh38
rs896387866 Chr.2: 164508510-164508510 on Build GRCh38
rs758974140 Chr.2: 164508777-164508777 on Build GRCh38
rs768427592 Chr.2: 164508577-164508577 on Build GRCh38
rs765746221 Chr.2: 164508772-164508772 on Build GRCh38
rs1004103090 Chr.2: 164508529-164508529 on Build GRCh38
rs779638211 Chr.2: 164508778-164508778 on Build GRCh38
rs1012484387 Chr.2: 164508635-164508635 on Build GRCh38
rs372525023 Chr.2: 164508486-164508486 on Build GRCh38
rs747940797 Chr.2: 164508787-164508787 on Build GRCh38
rs757905410 Chr.2: 164508778-164508778 on Build GRCh38
rs144801972 Chr.2: 164508540-164508540 on Build GRCh38
rs538731091 Chr.2: 164508680-164508680 on Build GRCh38
rs768862669 Chr.2: 164508815-164508815 on Build GRCh38
rs759596593 Chr.2: 164508513-164508513 on Build GRCh38
rs149516805 Chr.2: 164508587-164508587 on Build GRCh38
rs1022487318 Chr.2: 164508662-164508662 on Build GRCh38
rs746618166 Chr.2: 164508576-164508576 on Build GRCh38
rs869240678 Chr.2: 164508614-164508615 on Build GRCh38
rs760802934 Chr.2: 164508809-164508809 on Build GRCh38
rs568671902 Chr.2: 164508830-164508830 on Build GRCh38
rs753383699 Chr.2: 164508777-164508778 on Build GRCh38
rs373508476 Chr.2: 164508530-164508530 on Build GRCh38
rs764317240 Chr.2: 164508767-164508767 on Build GRCh38
rs748010252 Chr.2: 164508601-164508601 on Build GRCh38

View Full Product Details