Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 81474320-81474892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEL1L
Entrez Gene ID
6400
Gene Name
SEL1L ERAD E3 ligase adaptor subunit
Gene Aliases
PRO1063, SEL1-LIKE, SEL1L1
UniGene
Hs.181300
Gene Chromosome Location
Chr.14: 81471547-81533864 on Build GRCh38
Transcripts
NM_005065.5 XM_005267988.2 XM_005267989.3

SNP Detail

SNP ID Chromosome Location
rs11848933 Chr.14: 81474792-81474792 on Build GRCh38
rs756451539 Chr.14: 81474839-81474839 on Build GRCh38
rs929224894 Chr.14: 81474458-81474458 on Build GRCh38
rs942997769 Chr.14: 81474384-81474384 on Build GRCh38
rs910416494 Chr.14: 81474833-81474833 on Build GRCh38
rs11848973 Chr.14: 81474870-81474870 on Build GRCh38
rs999765108 Chr.14: 81474864-81474864 on Build GRCh38
rs910469373 Chr.14: 81474481-81474481 on Build GRCh38
rs148127930 Chr.14: 81474869-81474869 on Build GRCh38
rs887596298 Chr.14: 81474578-81474578 on Build GRCh38
rs930464461 Chr.14: 81474421-81474421 on Build GRCh38
rs1013559056 Chr.14: 81474697-81474697 on Build GRCh38
rs1026076237 Chr.14: 81474713-81474713 on Build GRCh38
rs373925647 Chr.14: 81474514-81474514 on Build GRCh38
rs528945786 Chr.14: 81474613-81474613 on Build GRCh38
rs773507622 Chr.14: 81474773-81474773 on Build GRCh38
rs202218166 Chr.14: 81474725-81474725 on Build GRCh38
rs559517786 Chr.14: 81474843-81474843 on Build GRCh38
rs189570996 Chr.14: 81474499-81474499 on Build GRCh38
rs1055596089 Chr.14: 81474642-81474642 on Build GRCh38
rs779992355 Chr.14: 81474346-81474346 on Build GRCh38
rs551649169 Chr.14: 81474774-81474774 on Build GRCh38
rs966975646 Chr.14: 81474803-81474803 on Build GRCh38
rs373604202 Chr.14: 81474657-81474659 on Build GRCh38
rs943271566 Chr.14: 81474844-81474844 on Build GRCh38
rs369780216 Chr.14: 81474662-81474662 on Build GRCh38
rs543602876 Chr.14: 81474484-81474484 on Build GRCh38
rs943305998 Chr.14: 81474491-81474491 on Build GRCh38
rs116642104 Chr.14: 81474576-81474576 on Build GRCh38
rs971799701 Chr.14: 81474705-81474705 on Build GRCh38
rs910236041 Chr.14: 81474381-81474381 on Build GRCh38
rs538432114 Chr.14: 81474650-81474652 on Build GRCh38
rs895151822 Chr.14: 81474656-81474656 on Build GRCh38
rs984658367 Chr.14: 81474422-81474422 on Build GRCh38
rs1034662829 Chr.14: 81474674-81474674 on Build GRCh38
rs951816623 Chr.14: 81474747-81474747 on Build GRCh38
rs55735981 Chr.14: 81474347-81474347 on Build GRCh38
rs1040273657 Chr.14: 81474410-81474410 on Build GRCh38
rs901799392 Chr.14: 81474424-81474424 on Build GRCh38
rs34755339 Chr.14: 81474604-81474605 on Build GRCh38
rs960670576 Chr.14: 81474686-81474686 on Build GRCh38
rs934069617 Chr.14: 81474524-81474524 on Build GRCh38
rs746844816 Chr.14: 81474579-81474579 on Build GRCh38
rs1011020993 Chr.14: 81474610-81474610 on Build GRCh38

View Full Product Details