Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65624433-65624996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC24A1
Entrez Gene ID
9187
Gene Name
solute carrier family 24 member 1
Gene Aliases
CSNB1D, HsT17412, NCKX, NCKX1, RODX
UniGene
Hs.173092
Gene Chromosome Location
Chr.15: 65611405-65660995 on Build GRCh38
Transcripts
NM_001301031.1 NM_001301032.1 NM_001301033.1 NM_004727.2 XM_005254778.3 XM_005254781.3 XM_006720768.3 XM_011522219.1 XM_011522220.2 XM_011522221.2 XM_011522222.2 XM_011522226.2 XM_017022724.1 XM_017022725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6222874 c.844T>G p.F282V Chr.15: 65624924-65624924 on Build GRCh38
4518891 c.849_850CC>TT p.P284S Chr.15: 65624929-65624930 on Build GRCh38
5437639 c.711T>G p.L237L Chr.15: 65624791-65624791 on Build GRCh38
4557324 c.723G>A p.M241I Chr.15: 65624803-65624803 on Build GRCh38
6225118 c.600C>T p.Y200Y Chr.15: 65624680-65624680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374754108 Chr.15: 65624685-65624685 on Build GRCh38
rs956792845 Chr.15: 65624927-65624927 on Build GRCh38
rs761718583 Chr.15: 65624944-65624947 on Build GRCh38
rs760045363 Chr.15: 65624976-65624976 on Build GRCh38
rs767891989 Chr.15: 65624819-65624819 on Build GRCh38
rs548862563 Chr.15: 65624932-65624932 on Build GRCh38
rs528969623 Chr.15: 65624942-65624942 on Build GRCh38
rs565538631 Chr.15: 65624785-65624785 on Build GRCh38
rs981823607 Chr.15: 65624876-65624876 on Build GRCh38
rs773119489 Chr.15: 65624623-65624623 on Build GRCh38
rs750598986 Chr.15: 65624563-65624563 on Build GRCh38
rs200047855 Chr.15: 65624561-65624561 on Build GRCh38
rs375544210 Chr.15: 65624718-65624718 on Build GRCh38
rs549873048 Chr.15: 65624916-65624916 on Build GRCh38
rs745538799 Chr.15: 65624760-65624760 on Build GRCh38
rs566014454 Chr.15: 65624508-65624508 on Build GRCh38
rs887605284 Chr.15: 65624920-65624920 on Build GRCh38
rs779702931 Chr.15: 65624665-65624665 on Build GRCh38
rs770611934 Chr.15: 65624659-65624659 on Build GRCh38
rs148676448 Chr.15: 65624611-65624611 on Build GRCh38
rs749097040 Chr.15: 65624747-65624747 on Build GRCh38
rs757876905 Chr.15: 65624496-65624499 on Build GRCh38
rs759809092 Chr.15: 65624807-65624807 on Build GRCh38
rs370009091 Chr.15: 65624769-65624769 on Build GRCh38
rs887615107 Chr.15: 65624524-65624524 on Build GRCh38
rs794727276 Chr.15: 65624966-65624966 on Build GRCh38
rs759402955 Chr.15: 65624971-65624971 on Build GRCh38
rs776452871 Chr.15: 65624915-65624917 on Build GRCh38
rs747889129 Chr.15: 65624458-65624458 on Build GRCh38
rs774546352 Chr.15: 65624973-65624973 on Build GRCh38
rs142214677 Chr.15: 65624657-65624657 on Build GRCh38
rs770284123 Chr.15: 65624771-65624771 on Build GRCh38
rs901728428 Chr.15: 65624943-65624943 on Build GRCh38
rs559502671 Chr.15: 65624682-65624682 on Build GRCh38
rs769783064 Chr.15: 65624958-65624958 on Build GRCh38
rs768402643 Chr.15: 65624840-65624840 on Build GRCh38
rs777989874 Chr.15: 65624834-65624835 on Build GRCh38
rs762689432 Chr.15: 65624506-65624506 on Build GRCh38
rs781205450 Chr.15: 65624588-65624588 on Build GRCh38
rs769315436 Chr.15: 65624716-65624716 on Build GRCh38
rs773726222 Chr.15: 65624950-65624950 on Build GRCh38
rs745729283 Chr.15: 65624903-65624903 on Build GRCh38
rs748124445 Chr.15: 65624589-65624589 on Build GRCh38
rs750772375 Chr.15: 65624692-65624692 on Build GRCh38
rs773602954 Chr.15: 65624776-65624776 on Build GRCh38
rs755024080 Chr.15: 65624579-65624579 on Build GRCh38
rs777796785 Chr.15: 65624847-65624847 on Build GRCh38
rs202245263 Chr.15: 65624883-65624883 on Build GRCh38
rs370617242 Chr.15: 65624856-65624856 on Build GRCh38
rs781442429 Chr.15: 65624719-65624719 on Build GRCh38
rs780182568 Chr.15: 65624575-65624575 on Build GRCh38
rs778541607 Chr.15: 65624575-65624575 on Build GRCh38
rs759036111 Chr.15: 65624523-65624523 on Build GRCh38
rs775927007 Chr.15: 65624680-65624680 on Build GRCh38
rs989047281 Chr.15: 65624787-65624787 on Build GRCh38
rs574198179 Chr.15: 65624663-65624663 on Build GRCh38
rs1001642304 Chr.15: 65624496-65624496 on Build GRCh38
rs367903341 Chr.15: 65624686-65624686 on Build GRCh38
rs765829105 Chr.15: 65624881-65624881 on Build GRCh38
rs774962423 Chr.15: 65624766-65624766 on Build GRCh38
rs371758439 Chr.15: 65624694-65624694 on Build GRCh38
rs749076357 Chr.15: 65624631-65624631 on Build GRCh38
rs779750981 Chr.15: 65624917-65624917 on Build GRCh38
rs1013936971 Chr.15: 65624855-65624855 on Build GRCh38
rs763281492 Chr.15: 65624700-65624700 on Build GRCh38
rs1023531965 Chr.15: 65624871-65624871 on Build GRCh38
rs201525281 Chr.15: 65624509-65624509 on Build GRCh38
rs754357494 Chr.15: 65624541-65624541 on Build GRCh38
rs747095432 Chr.15: 65624577-65624577 on Build GRCh38
rs373003119 Chr.15: 65624777-65624777 on Build GRCh38
rs756319716 Chr.15: 65624830-65624830 on Build GRCh38
rs771686207 Chr.15: 65624912-65624912 on Build GRCh38
rs558229888 Chr.15: 65624511-65624511 on Build GRCh38
rs532760023 Chr.15: 65624902-65624902 on Build GRCh38
rs766681351 Chr.15: 65624789-65624789 on Build GRCh38
rs756085069 Chr.15: 65624730-65624730 on Build GRCh38
rs566399857 Chr.15: 65624832-65624832 on Build GRCh38
rs745437732 Chr.15: 65624581-65624587 on Build GRCh38
rs771524909 Chr.15: 65624764-65624764 on Build GRCh38
rs201188098 Chr.15: 65624744-65624744 on Build GRCh38
rs770646453 Chr.15: 65624750-65624750 on Build GRCh38
rs757471063 Chr.15: 65624909-65624910 on Build GRCh38
rs545291420 Chr.15: 65624797-65624797 on Build GRCh38
rs752818074 Chr.15: 65624827-65624827 on Build GRCh38
rs769713196 Chr.15: 65624459-65624459 on Build GRCh38
rs376692252 Chr.15: 65624921-65624921 on Build GRCh38
rs771615174 Chr.15: 65624592-65624593 on Build GRCh38
rs933577086 Chr.15: 65624652-65624652 on Build GRCh38
rs563752295 Chr.15: 65624887-65624887 on Build GRCh38
rs766563238 Chr.15: 65624709-65624709 on Build GRCh38
rs772958253 Chr.15: 65624478-65624478 on Build GRCh38
rs752873180 Chr.15: 65624727-65624727 on Build GRCh38
rs1004755801 Chr.15: 65624583-65624583 on Build GRCh38
rs754104796 Chr.15: 65624941-65624941 on Build GRCh38
rs896122807 Chr.15: 65624836-65624836 on Build GRCh38
rs757037005 Chr.15: 65624882-65624882 on Build GRCh38
rs886051349 Chr.15: 65624505-65624508 on Build GRCh38
rs373383445 Chr.15: 65624753-65624753 on Build GRCh38
rs763388811 Chr.15: 65624953-65624953 on Build GRCh38
rs200714082 Chr.15: 65624681-65624681 on Build GRCh38
rs189152503 Chr.15: 65624530-65624530 on Build GRCh38
rs771517323 Chr.15: 65624969-65624969 on Build GRCh38
rs775750975 Chr.15: 65624538-65624538 on Build GRCh38
rs751730680 Chr.15: 65624712-65624712 on Build GRCh38

View Full Product Details