Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15643182-15643758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR484
Entrez Gene ID
619553
Gene Name
microRNA 484
Gene Aliases
MIRN484, hsa-mir-484, mir-484
UniGene
Gene Chromosome Location
Chr.16: 15643294-15643372 on Build GRCh38
Transcripts
NR_030159.1 NM_001143979.1 XM_006720897.3 XM_011522553.2

Gene Symbol
NDE1
Entrez Gene ID
54820
Gene Name
nudE neurodevelopment protein 1
Gene Aliases
HOM-TES-87, LIS4, MHAC, NDE, NUDE, NUDE1
UniGene
Hs.655378
Gene Chromosome Location
Chr.16: 15643267-15726353 on Build GRCh38
Transcripts
NR_030159.1 NM_001143979.1 XM_006720897.3 XM_011522553.2

SNP Detail

SNP ID Chromosome Location
rs758508515 Chr.16: 15643322-15643322 on Build GRCh38
rs969536601 Chr.16: 15643258-15643258 on Build GRCh38
rs773432152 Chr.16: 15643393-15643393 on Build GRCh38
rs768896489 Chr.16: 15643357-15643357 on Build GRCh38
rs373853748 Chr.16: 15643403-15643403 on Build GRCh38
rs978398324 Chr.16: 15643530-15643530 on Build GRCh38
rs772657269 Chr.16: 15643290-15643290 on Build GRCh38
rs920716102 Chr.16: 15643591-15643591 on Build GRCh38
rs780092467 Chr.16: 15643363-15643363 on Build GRCh38
rs886051711 Chr.16: 15643352-15643353 on Build GRCh38
rs752629218 Chr.16: 15643399-15643399 on Build GRCh38
rs368690836 Chr.16: 15643329-15643329 on Build GRCh38
rs747902225 Chr.16: 15643255-15643255 on Build GRCh38
rs566801193 Chr.16: 15643405-15643405 on Build GRCh38
rs146418251 Chr.16: 15643556-15643556 on Build GRCh38
rs911986645 Chr.16: 15643306-15643306 on Build GRCh38
rs908097165 Chr.16: 15643649-15643649 on Build GRCh38
rs777106128 Chr.16: 15643376-15643376 on Build GRCh38
rs886051715 Chr.16: 15643614-15643614 on Build GRCh38
rs557029026 Chr.16: 15643538-15643538 on Build GRCh38
rs1026622893 Chr.16: 15643226-15643226 on Build GRCh38
rs970506650 Chr.16: 15643343-15643343 on Build GRCh38
rs886051712 Chr.16: 15643558-15643558 on Build GRCh38
rs957880160 Chr.16: 15643348-15643348 on Build GRCh38
rs373275027 Chr.16: 15643406-15643406 on Build GRCh38
rs930779079 Chr.16: 15643607-15643607 on Build GRCh38
rs796579453 Chr.16: 15643571-15643571 on Build GRCh38
rs957216532 Chr.16: 15643489-15643489 on Build GRCh38
rs765211962 Chr.16: 15643319-15643319 on Build GRCh38
rs950343522 Chr.16: 15643713-15643713 on Build GRCh38
rs1042834502 Chr.16: 15643295-15643295 on Build GRCh38
rs551758626 Chr.16: 15643398-15643398 on Build GRCh38
rs561902034 Chr.16: 15643294-15643294 on Build GRCh38
rs540409907 Chr.16: 15643238-15643238 on Build GRCh38
rs1046974765 Chr.16: 15643695-15643695 on Build GRCh38
rs540576709 Chr.16: 15643722-15643722 on Build GRCh38
rs770021443 Chr.16: 15643384-15643384 on Build GRCh38
rs187630586 Chr.16: 15643558-15643558 on Build GRCh38
rs35814370 Chr.16: 15643285-15643286 on Build GRCh38
rs957953716 Chr.16: 15643277-15643277 on Build GRCh38
rs1045031275 Chr.16: 15643735-15643735 on Build GRCh38
rs1010750700 Chr.16: 15643549-15643549 on Build GRCh38
rs777234206 Chr.16: 15643270-15643270 on Build GRCh38
rs749030245 Chr.16: 15643275-15643275 on Build GRCh38
rs767375940 Chr.16: 15643326-15643326 on Build GRCh38
rs757090627 Chr.16: 15643346-15643347 on Build GRCh38
rs745331642 Chr.16: 15643346-15643346 on Build GRCh38
rs762110135 Chr.16: 15643380-15643380 on Build GRCh38
rs1046427580 Chr.16: 15643731-15643738 on Build GRCh38
rs753513445 Chr.16: 15643311-15643312 on Build GRCh38
rs371097200 Chr.16: 15643395-15643395 on Build GRCh38
rs760705591 Chr.16: 15643402-15643402 on Build GRCh38
rs756111511 Chr.16: 15643267-15643267 on Build GRCh38
rs974798346 Chr.16: 15643581-15643581 on Build GRCh38
rs867184365 Chr.16: 15643299-15643299 on Build GRCh38
rs112103086 Chr.16: 15643360-15643360 on Build GRCh38
rs1025632262 Chr.16: 15643208-15643208 on Build GRCh38
rs34560499 Chr.16: 15643418-15643418 on Build GRCh38
rs750701617 Chr.16: 15643303-15643303 on Build GRCh38
rs866065662 Chr.16: 15643691-15643691 on Build GRCh38
rs951082490 Chr.16: 15643264-15643264 on Build GRCh38
rs988412183 Chr.16: 15643505-15643505 on Build GRCh38
rs886051714 Chr.16: 15643581-15643581 on Build GRCh38
rs983586597 Chr.16: 15643633-15643633 on Build GRCh38
rs182957892 Chr.16: 15643444-15643444 on Build GRCh38
rs1004148539 Chr.16: 15643480-15643480 on Build GRCh38
rs773950878 Chr.16: 15643283-15643283 on Build GRCh38
rs374184031 Chr.16: 15643328-15643328 on Build GRCh38
rs77774475 Chr.16: 15643548-15643548 on Build GRCh38
rs533376652 Chr.16: 15643349-15643349 on Build GRCh38
rs929760532 Chr.16: 15643661-15643661 on Build GRCh38
rs868335600 Chr.16: 15643202-15643202 on Build GRCh38
rs943491074 Chr.16: 15643307-15643307 on Build GRCh38
rs915460737 Chr.16: 15643513-15643513 on Build GRCh38
rs988915860 Chr.16: 15643280-15643280 on Build GRCh38
rs762137640 Chr.16: 15643317-15643317 on Build GRCh38
rs1035186302 Chr.16: 15643486-15643486 on Build GRCh38
rs987847102 Chr.16: 15643569-15643569 on Build GRCh38
rs778729812 Chr.16: 15643335-15643335 on Build GRCh38
rs746354381 Chr.16: 15643289-15643289 on Build GRCh38
rs376603273 Chr.16: 15643320-15643320 on Build GRCh38
rs771708381 Chr.16: 15643347-15643347 on Build GRCh38
rs886051716 Chr.16: 15643695-15643695 on Build GRCh38
rs1018935228 Chr.16: 15643570-15643571 on Build GRCh38
rs1023637555 Chr.16: 15643228-15643228 on Build GRCh38
rs886051710 Chr.16: 15643281-15643281 on Build GRCh38
rs573161303 Chr.16: 15643652-15643652 on Build GRCh38
rs888335379 Chr.16: 15643702-15643702 on Build GRCh38
rs747574561 Chr.16: 15643356-15643356 on Build GRCh38
rs886051713 Chr.16: 15643581-15643582 on Build GRCh38
rs1047961436 Chr.16: 15643422-15643422 on Build GRCh38
rs1041311933 Chr.16: 15643583-15643583 on Build GRCh38
rs557379176 Chr.16: 15643266-15643267 on Build GRCh38
rs1046083638 Chr.16: 15643490-15643490 on Build GRCh38
rs1052272611 Chr.16: 15643300-15643300 on Build GRCh38
rs1039193760 Chr.16: 15643309-15643309 on Build GRCh38
rs912898761 Chr.16: 15643285-15643285 on Build GRCh38
rs770341582 Chr.16: 15643282-15643282 on Build GRCh38
rs1025111704 Chr.16: 15643337-15643337 on Build GRCh38
rs969422378 Chr.16: 15643452-15643452 on Build GRCh38
rs982274723 Chr.16: 15643263-15643263 on Build GRCh38
rs1005590783 Chr.16: 15643224-15643224 on Build GRCh38
rs112906136 Chr.16: 15643361-15643361 on Build GRCh38
rs776702047 Chr.16: 15643314-15643314 on Build GRCh38
rs913853637 Chr.16: 15643372-15643372 on Build GRCh38
rs952834573 Chr.16: 15643219-15643219 on Build GRCh38
rs941384581 Chr.16: 15643708-15643708 on Build GRCh38
rs79063947 Chr.16: 15643529-15643529 on Build GRCh38
rs776168097 Chr.16: 15643291-15643291 on Build GRCh38
rs555304619 Chr.16: 15643623-15643623 on Build GRCh38
rs928229223 Chr.16: 15643265-15643265 on Build GRCh38
rs755941970 Chr.16: 15643267-15643267 on Build GRCh38

View Full Product Details