Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50328568-50328999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD7
Entrez Gene ID
29117
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, NAG4
UniGene
Hs.437894
Gene Chromosome Location
Chr.16: 50317563-50368969 on Build GRCh38
Transcripts
NM_001173984.2 NM_013263.4 XM_011523046.2 XM_011523047.2 XM_011523048.2 XM_011523049.2 XM_011523050.2 XM_017023179.1 XM_017023180.1 XM_017023181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1378189 c.1015G>A p.E339K Chr.16: 50328741-50328741 on Build GRCh38
6016184 c.1047G>A p.T349T Chr.16: 50328709-50328709 on Build GRCh38
6016185 c.1025G>C p.R342T Chr.16: 50328731-50328731 on Build GRCh38
6421361 c.1015G>T p.E339* Chr.16: 50328741-50328741 on Build GRCh38
3387465 c.1012-1G>C p.? Chr.16: 50328745-50328745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781130848 Chr.16: 50328645-50328645 on Build GRCh38
rs779831987 Chr.16: 50328786-50328786 on Build GRCh38
rs1033821059 Chr.16: 50328887-50328887 on Build GRCh38
rs775705058 Chr.16: 50328651-50328651 on Build GRCh38
rs377360131 Chr.16: 50328935-50328935 on Build GRCh38
rs745659123 Chr.16: 50328771-50328771 on Build GRCh38
rs780195350 Chr.16: 50328657-50328657 on Build GRCh38
rs747258100 Chr.16: 50328897-50328897 on Build GRCh38
rs751164926 Chr.16: 50328761-50328761 on Build GRCh38
rs762927474 Chr.16: 50328671-50328671 on Build GRCh38
rs1001530107 Chr.16: 50328855-50328855 on Build GRCh38
rs745750643 Chr.16: 50328647-50328647 on Build GRCh38
rs904142682 Chr.16: 50328845-50328845 on Build GRCh38
rs1013588706 Chr.16: 50328826-50328826 on Build GRCh38
rs749424591 Chr.16: 50328789-50328789 on Build GRCh38
rs751947061 Chr.16: 50328647-50328648 on Build GRCh38
rs116313086 Chr.16: 50328712-50328712 on Build GRCh38
rs774732703 Chr.16: 50328796-50328796 on Build GRCh38
rs568449209 Chr.16: 50328780-50328780 on Build GRCh38
rs780900090 Chr.16: 50328770-50328770 on Build GRCh38
rs991901357 Chr.16: 50328914-50328914 on Build GRCh38
rs752303750 Chr.16: 50328742-50328742 on Build GRCh38
rs983314905 Chr.16: 50328617-50328617 on Build GRCh38
rs1021207818 Chr.16: 50328817-50328817 on Build GRCh38
rs538440586 Chr.16: 50328741-50328741 on Build GRCh38
rs188183810 Chr.16: 50328679-50328679 on Build GRCh38
rs768971711 Chr.16: 50328672-50328672 on Build GRCh38
rs769770222 Chr.16: 50328650-50328650 on Build GRCh38
rs866591032 Chr.16: 50328681-50328681 on Build GRCh38
rs761173561 Chr.16: 50328728-50328728 on Build GRCh38
rs950739997 Chr.16: 50328937-50328937 on Build GRCh38
rs569575592 Chr.16: 50328722-50328722 on Build GRCh38
rs374524830 Chr.16: 50328756-50328756 on Build GRCh38
rs566489258 Chr.16: 50328608-50328608 on Build GRCh38
rs891112321 Chr.16: 50328777-50328777 on Build GRCh38
rs756132589 Chr.16: 50328774-50328774 on Build GRCh38
rs1042806740 Chr.16: 50328640-50328640 on Build GRCh38
rs757820106 Chr.16: 50328751-50328751 on Build GRCh38
rs147728435 Chr.16: 50328926-50328926 on Build GRCh38
rs1002358438 Chr.16: 50328673-50328673 on Build GRCh38
rs770674241 Chr.16: 50328636-50328636 on Build GRCh38
rs768575578 Chr.16: 50328795-50328795 on Build GRCh38
rs746850124 Chr.16: 50328631-50328631 on Build GRCh38
rs534329734 Chr.16: 50328812-50328812 on Build GRCh38
rs555187356 Chr.16: 50328763-50328763 on Build GRCh38
rs745648261 Chr.16: 50328683-50328683 on Build GRCh38
rs762296439 Chr.16: 50328687-50328687 on Build GRCh38

View Full Product Details