Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28358727-28359260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM199
Entrez Gene ID
147007
Gene Name
transmembrane protein 199
Gene Aliases
C17orf32, CDG2P, VMA12, VPH2
UniGene
Hs.594696
Gene Chromosome Location
Chr.17: 28357581-28363683 on Build GRCh38
Transcripts
NM_152464.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749918 c.283C>T p.R95W Chr.17: 28358987-28358987 on Build GRCh38
3515321 c.281C>T p.P94L Chr.17: 28358985-28358985 on Build GRCh38
3717244 c.285+10G>A p.? Chr.17: 28358999-28358999 on Build GRCh38
2800045 c.269T>C p.V90A Chr.17: 28358973-28358973 on Build GRCh38
7394217 c.256T>C p.Y86H Chr.17: 28358960-28358960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201057584 Chr.17: 28358929-28358929 on Build GRCh38
rs782430196 Chr.17: 28358919-28358919 on Build GRCh38
rs782799715 Chr.17: 28358965-28358965 on Build GRCh38
rs567566543 Chr.17: 28358838-28358838 on Build GRCh38
rs762621199 Chr.17: 28358982-28358982 on Build GRCh38
rs186521941 Chr.17: 28359042-28359042 on Build GRCh38
rs377332220 Chr.17: 28359158-28359159 on Build GRCh38
rs995773483 Chr.17: 28359068-28359068 on Build GRCh38
rs946663749 Chr.17: 28358963-28358963 on Build GRCh38
rs887266320 Chr.17: 28359196-28359196 on Build GRCh38
rs782174966 Chr.17: 28359034-28359034 on Build GRCh38
rs35276012 Chr.17: 28358960-28358960 on Build GRCh38
rs782792993 Chr.17: 28358961-28358961 on Build GRCh38
rs7210719 Chr.17: 28359195-28359195 on Build GRCh38
rs371844645 Chr.17: 28358955-28358955 on Build GRCh38
rs781938404 Chr.17: 28358909-28358909 on Build GRCh38
rs896259772 Chr.17: 28358984-28358984 on Build GRCh38
rs767804545 Chr.17: 28359214-28359214 on Build GRCh38
rs782307628 Chr.17: 28359017-28359017 on Build GRCh38
rs979221515 Chr.17: 28358967-28358967 on Build GRCh38
rs782569126 Chr.17: 28358975-28358975 on Build GRCh38
rs782544541 Chr.17: 28358989-28358989 on Build GRCh38
rs181153769 Chr.17: 28359035-28359035 on Build GRCh38
rs964197022 Chr.17: 28359237-28359237 on Build GRCh38
rs782623073 Chr.17: 28359159-28359159 on Build GRCh38
rs547014476 Chr.17: 28358887-28358887 on Build GRCh38
rs535316912 Chr.17: 28358879-28358879 on Build GRCh38
rs782754708 Chr.17: 28358871-28358871 on Build GRCh38
rs781823703 Chr.17: 28359099-28359099 on Build GRCh38
rs782238201 Chr.17: 28359190-28359190 on Build GRCh38
rs545031531 Chr.17: 28359176-28359176 on Build GRCh38
rs549119839 Chr.17: 28358806-28358806 on Build GRCh38
rs373879073 Chr.17: 28358823-28358823 on Build GRCh38
rs782128570 Chr.17: 28358932-28358932 on Build GRCh38
rs189741454 Chr.17: 28358902-28358902 on Build GRCh38
rs1019655653 Chr.17: 28359228-28359228 on Build GRCh38
rs781812829 Chr.17: 28358872-28358872 on Build GRCh38
rs782554903 Chr.17: 28358878-28358878 on Build GRCh38
rs1044769906 Chr.17: 28359033-28359033 on Build GRCh38
rs377231664 Chr.17: 28358959-28358959 on Build GRCh38
rs964374455 Chr.17: 28358950-28358950 on Build GRCh38
rs765960166 Chr.17: 28359074-28359074 on Build GRCh38
rs36063495 Chr.17: 28359154-28359155 on Build GRCh38
rs139555380 Chr.17: 28359075-28359075 on Build GRCh38
rs782240691 Chr.17: 28358999-28358999 on Build GRCh38
rs75296655 Chr.17: 28358864-28358864 on Build GRCh38
rs1019688135 Chr.17: 28358893-28358893 on Build GRCh38
rs143100512 Chr.17: 28358878-28358878 on Build GRCh38
rs369451173 Chr.17: 28359030-28359030 on Build GRCh38
rs565003133 Chr.17: 28359073-28359073 on Build GRCh38
rs782623668 Chr.17: 28358839-28358839 on Build GRCh38
rs781995281 Chr.17: 28359026-28359027 on Build GRCh38
rs782379269 Chr.17: 28359004-28359004 on Build GRCh38
rs201975015 Chr.17: 28358988-28358988 on Build GRCh38
rs200616117 Chr.17: 28358972-28358972 on Build GRCh38
rs781910697 Chr.17: 28358962-28358962 on Build GRCh38
rs782501471 Chr.17: 28358964-28358964 on Build GRCh38
rs145031553 Chr.17: 28359215-28359215 on Build GRCh38
rs367645620 Chr.17: 28358934-28358934 on Build GRCh38
rs782673839 Chr.17: 28358993-28358993 on Build GRCh38
rs199512818 Chr.17: 28358974-28358974 on Build GRCh38

View Full Product Details