Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 55393515-55394114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMD
Entrez Gene ID
23531
Gene Name
monocyte to macrophage differentiation associated
Gene Aliases
MMA, MMD1, PAQR11
UniGene
Hs.463483
Gene Chromosome Location
Chr.17: 55392613-55421993 on Build GRCh38
Transcripts
NM_012329.2 XM_006721795.2

SNP Detail

SNP ID Chromosome Location
rs903757617 Chr.17: 55394007-55394007 on Build GRCh38
rs922679677 Chr.17: 55393824-55393824 on Build GRCh38
rs941635751 Chr.17: 55393862-55393862 on Build GRCh38
rs758961950 Chr.17: 55393662-55393662 on Build GRCh38
rs1023354508 Chr.17: 55393810-55393810 on Build GRCh38
rs889875543 Chr.17: 55393951-55393951 on Build GRCh38
rs942673791 Chr.17: 55393984-55393984 on Build GRCh38
rs939984153 Chr.17: 55393739-55393739 on Build GRCh38
rs904193460 Chr.17: 55394033-55394033 on Build GRCh38
rs565406252 Chr.17: 55393745-55393745 on Build GRCh38
rs1022179927 Chr.17: 55393653-55393653 on Build GRCh38
rs542953352 Chr.17: 55393564-55393564 on Build GRCh38
rs114158015 Chr.17: 55393897-55393897 on Build GRCh38
rs1007517307 Chr.17: 55393975-55393975 on Build GRCh38
rs530686616 Chr.17: 55394004-55394004 on Build GRCh38
rs999360403 Chr.17: 55393696-55393696 on Build GRCh38
rs765835891 Chr.17: 55393597-55393597 on Build GRCh38
rs1051145035 Chr.17: 55393948-55393948 on Build GRCh38
rs4476230 Chr.17: 55393744-55393744 on Build GRCh38
rs992982607 Chr.17: 55393613-55393613 on Build GRCh38
rs761473678 Chr.17: 55393630-55393630 on Build GRCh38
rs889760031 Chr.17: 55393606-55393606 on Build GRCh38
rs527996194 Chr.17: 55393952-55393952 on Build GRCh38
rs960471170 Chr.17: 55393740-55393740 on Build GRCh38
rs972906499 Chr.17: 55393882-55393882 on Build GRCh38
rs751532494 Chr.17: 55393921-55393921 on Build GRCh38
rs998644886 Chr.17: 55393588-55393588 on Build GRCh38
rs118033564 Chr.17: 55393815-55393815 on Build GRCh38
rs4570925 Chr.17: 55393830-55393830 on Build GRCh38
rs751989522 Chr.17: 55393814-55393814 on Build GRCh38
rs1042992694 Chr.17: 55393989-55393989 on Build GRCh38
rs1031282941 Chr.17: 55393706-55393706 on Build GRCh38
rs75304510 Chr.17: 55393658-55393658 on Build GRCh38
rs560791394 Chr.17: 55393784-55393784 on Build GRCh38
rs888937272 Chr.17: 55393909-55393910 on Build GRCh38
rs1051206983 Chr.17: 55393576-55393576 on Build GRCh38
rs113567733 Chr.17: 55393646-55393646 on Build GRCh38
rs910126176 Chr.17: 55393860-55393860 on Build GRCh38
rs560381509 Chr.17: 55393547-55393547 on Build GRCh38
rs984424365 Chr.17: 55393857-55393857 on Build GRCh38

View Full Product Details