Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8810739-8811274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF558
Entrez Gene ID
148156
Gene Name
zinc finger protein 558
Gene Aliases
-
UniGene
Hs.349444
Gene Chromosome Location
Chr.19: 8809576-8832304 on Build GRCh38
Transcripts
NM_001304350.1 NM_144693.2 XM_011527703.2 XM_017026370.1 XM_017026371.1 XM_017026372.1

SNP Detail

SNP ID Chromosome Location
rs1023633555 Chr.19: 8810866-8810866 on Build GRCh38
rs567007993 Chr.19: 8811215-8811215 on Build GRCh38
rs369989137 Chr.19: 8811247-8811247 on Build GRCh38
rs992687306 Chr.19: 8810835-8810835 on Build GRCh38
rs533737242 Chr.19: 8810952-8810952 on Build GRCh38
rs35450309 Chr.19: 8810993-8810994 on Build GRCh38
rs547176748 Chr.19: 8811143-8811143 on Build GRCh38
rs532241820 Chr.19: 8810818-8810818 on Build GRCh38
rs966293263 Chr.19: 8811195-8811195 on Build GRCh38
rs112409351 Chr.19: 8811251-8811251 on Build GRCh38
rs564355717 Chr.19: 8811069-8811069 on Build GRCh38
rs540775196 Chr.19: 8811144-8811144 on Build GRCh38
rs138035537 Chr.19: 8811040-8811040 on Build GRCh38
rs960786885 Chr.19: 8810824-8810824 on Build GRCh38
rs540465283 Chr.19: 8810776-8810776 on Build GRCh38
rs913064101 Chr.19: 8811191-8811191 on Build GRCh38
rs150312342 Chr.19: 8810865-8810865 on Build GRCh38
rs560000577 Chr.19: 8811235-8811235 on Build GRCh38
rs1027884933 Chr.19: 8810804-8810804 on Build GRCh38
rs1004967032 Chr.19: 8811051-8811051 on Build GRCh38
rs566913700 Chr.19: 8811158-8811158 on Build GRCh38
rs912950896 Chr.19: 8810935-8810935 on Build GRCh38
rs1024169137 Chr.19: 8811112-8811112 on Build GRCh38
rs112816154 Chr.19: 8811229-8811229 on Build GRCh38
rs1040564322 Chr.19: 8810946-8810946 on Build GRCh38
rs1582532 Chr.19: 8810810-8810810 on Build GRCh38
rs975842707 Chr.19: 8811225-8811225 on Build GRCh38
rs782310116 Chr.19: 8811121-8811121 on Build GRCh38
rs559120167 Chr.19: 8811200-8811200 on Build GRCh38
rs782819564 Chr.19: 8811244-8811244 on Build GRCh38
rs566372066 Chr.19: 8810875-8810875 on Build GRCh38
rs10407106 Chr.19: 8811167-8811167 on Build GRCh38
rs1049273642 Chr.19: 8810960-8810960 on Build GRCh38
rs944463532 Chr.19: 8810940-8810940 on Build GRCh38
rs935798576 Chr.19: 8810889-8810889 on Build GRCh38
rs538281313 Chr.19: 8811206-8811206 on Build GRCh38
rs531235444 Chr.19: 8811046-8811046 on Build GRCh38
rs386028 Chr.19: 8811173-8811173 on Build GRCh38
rs896687498 Chr.19: 8811079-8811079 on Build GRCh38
rs544611520 Chr.19: 8810907-8810907 on Build GRCh38
rs996437844 Chr.19: 8810799-8810799 on Build GRCh38
rs190935602 Chr.19: 8810937-8810937 on Build GRCh38
rs988647603 Chr.19: 8810917-8810917 on Build GRCh38
rs569354812 Chr.19: 8811202-8811202 on Build GRCh38
rs925875563 Chr.19: 8810888-8810888 on Build GRCh38
rs149503399 Chr.19: 8811090-8811090 on Build GRCh38
rs957280775 Chr.19: 8811166-8811166 on Build GRCh38
rs782142004 Chr.19: 8811238-8811238 on Build GRCh38
rs200950028 Chr.19: 8811248-8811248 on Build GRCh38
rs547768775 Chr.19: 8811086-8811086 on Build GRCh38
rs782222908 Chr.19: 8811230-8811230 on Build GRCh38
rs980032046 Chr.19: 8810885-8810885 on Build GRCh38
rs782387678 Chr.19: 8811017-8811017 on Build GRCh38
rs34195257 Chr.19: 8811165-8811166 on Build GRCh38
rs921594732 Chr.19: 8810947-8810947 on Build GRCh38
rs1005019796 Chr.19: 8810813-8810813 on Build GRCh38
rs753781497 Chr.19: 8811060-8811060 on Build GRCh38

View Full Product Details