Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 171602242-171602766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLD1
Entrez Gene ID
5337
Gene Name
phospholipase D1
Gene Aliases
-
UniGene
Hs.382865
Gene Chromosome Location
Chr.3: 171600404-171810494 on Build GRCh38
Transcripts
NM_001130081.2 NM_002662.4 XM_005247533.2 XM_005247534.2 XM_011512899.2 XM_017006624.1

SNP Detail

SNP ID Chromosome Location
rs546412109 Chr.3: 171602583-171602583 on Build GRCh38
rs983589073 Chr.3: 171602630-171602630 on Build GRCh38
rs147685486 Chr.3: 171602343-171602343 on Build GRCh38
rs547579727 Chr.3: 171602669-171602669 on Build GRCh38
rs761432990 Chr.3: 171602495-171602495 on Build GRCh38
rs568738156 Chr.3: 171602494-171602494 on Build GRCh38
rs993018263 Chr.3: 171602492-171602492 on Build GRCh38
rs948889179 Chr.3: 171602500-171602500 on Build GRCh38
rs899699258 Chr.3: 171602591-171602591 on Build GRCh38
rs766166991 Chr.3: 171602691-171602691 on Build GRCh38
rs571080728 Chr.3: 171602536-171602536 on Build GRCh38
rs968878498 Chr.3: 171602678-171602678 on Build GRCh38
rs928181857 Chr.3: 171602349-171602349 on Build GRCh38
rs1025358269 Chr.3: 171602579-171602579 on Build GRCh38
rs772769559 Chr.3: 171602372-171602372 on Build GRCh38
rs902365670 Chr.3: 171602455-171602455 on Build GRCh38
rs375345526 Chr.3: 171602501-171602501 on Build GRCh38
rs924244736 Chr.3: 171602451-171602451 on Build GRCh38
rs534506998 Chr.3: 171602511-171602511 on Build GRCh38
rs997079114 Chr.3: 171602611-171602611 on Build GRCh38
rs916568469 Chr.3: 171602424-171602424 on Build GRCh38
rs549391388 Chr.3: 171602280-171602280 on Build GRCh38
rs973813600 Chr.3: 171602285-171602285 on Build GRCh38
rs73038021 Chr.3: 171602284-171602284 on Build GRCh38
rs570994078 Chr.3: 171602413-171602413 on Build GRCh38
rs774937090 Chr.3: 171602298-171602298 on Build GRCh38
rs187747806 Chr.3: 171602542-171602542 on Build GRCh38
rs547526907 Chr.3: 171602323-171602323 on Build GRCh38
rs995260992 Chr.3: 171602547-171602547 on Build GRCh38
rs183477797 Chr.3: 171602369-171602369 on Build GRCh38
rs554495303 Chr.3: 171602507-171602507 on Build GRCh38
rs964201078 Chr.3: 171602399-171602399 on Build GRCh38
rs961272490 Chr.3: 171602665-171602665 on Build GRCh38
rs1031318520 Chr.3: 171602694-171602694 on Build GRCh38
rs112775585 Chr.3: 171602416-171602416 on Build GRCh38
rs760123028 Chr.3: 171602317-171602317 on Build GRCh38
rs938948936 Chr.3: 171602491-171602491 on Build GRCh38
rs529537174 Chr.3: 171602695-171602695 on Build GRCh38
rs9822322 Chr.3: 171602437-171602437 on Build GRCh38
rs935128763 Chr.3: 171602459-171602459 on Build GRCh38
rs556617161 Chr.3: 171602446-171602446 on Build GRCh38
rs1021416800 Chr.3: 171602672-171602672 on Build GRCh38
rs554375221 Chr.3: 171602517-171602517 on Build GRCh38
rs950826898 Chr.3: 171602308-171602308 on Build GRCh38
rs959683584 Chr.3: 171602709-171602709 on Build GRCh38
rs1021785347 Chr.3: 171602425-171602425 on Build GRCh38
rs1047769627 Chr.3: 171602532-171602532 on Build GRCh38
rs991085434 Chr.3: 171602675-171602675 on Build GRCh38
rs539668511 Chr.3: 171602357-171602357 on Build GRCh38
rs147062338 Chr.3: 171602304-171602304 on Build GRCh38
rs968451041 Chr.3: 171602428-171602428 on Build GRCh38

View Full Product Details