Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113273135-113273639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOC
Entrez Gene ID
91653
Gene Name
BOC cell adhesion associated, oncogene regulated
Gene Aliases
CDON2
UniGene
Hs.591318
Gene Chromosome Location
Chr.3: 113211003-113287463 on Build GRCh38
Transcripts
NM_001301861.1 NM_033254.3 XM_005247891.2 XM_005247892.2 XM_005247897.2 XM_011513305.2 XM_017007441.1 XM_017007442.1 XM_017007443.1 XM_017007444.1 XM_017007445.1 XM_017007446.1 XM_017007447.1 XM_017007448.1 XM_017007449.1 XM_017007450.1 XM_017007451.1 XM_017007452.1 XM_017007453.1 XM_017007454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453836 c.1039C>T p.R347C Chr.3: 113273146-113273146 on Build GRCh38
5141421 c.1162G>A p.V388I Chr.3: 113273269-113273269 on Build GRCh38
294169 c.1161C>A p.G387G Chr.3: 113273268-113273268 on Build GRCh38
397221 c.1071G>A p.L357L Chr.3: 113273178-113273178 on Build GRCh38
6612227 c.1055C>T p.P352L Chr.3: 113273162-113273162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998774925 Chr.3: 113273224-113273224 on Build GRCh38
rs777077077 Chr.3: 113273157-113273157 on Build GRCh38
rs779374530 Chr.3: 113273377-113273377 on Build GRCh38
rs750517961 Chr.3: 113273173-113273173 on Build GRCh38
rs747247386 Chr.3: 113273216-113273216 on Build GRCh38
rs765657069 Chr.3: 113273160-113273160 on Build GRCh38
rs767660512 Chr.3: 113273352-113273352 on Build GRCh38
rs1018129619 Chr.3: 113273259-113273259 on Build GRCh38
rs772662226 Chr.3: 113273391-113273391 on Build GRCh38
rs757037428 Chr.3: 113273467-113273467 on Build GRCh38
rs772821398 Chr.3: 113273313-113273313 on Build GRCh38
rs369203280 Chr.3: 113273214-113273214 on Build GRCh38
rs112165738 Chr.3: 113273229-113273229 on Build GRCh38
rs374518460 Chr.3: 113273356-113273356 on Build GRCh38
rs765251861 Chr.3: 113273328-113273328 on Build GRCh38
rs750153821 Chr.3: 113273198-113273198 on Build GRCh38
rs761403195 Chr.3: 113273179-113273179 on Build GRCh38
rs547676544 Chr.3: 113273589-113273589 on Build GRCh38
rs749770235 Chr.3: 113273292-113273292 on Build GRCh38
rs749662456 Chr.3: 113273359-113273359 on Build GRCh38
rs767316555 Chr.3: 113273180-113273180 on Build GRCh38
rs751004098 Chr.3: 113273434-113273434 on Build GRCh38
rs781732471 Chr.3: 113273219-113273219 on Build GRCh38
rs777320137 Chr.3: 113273314-113273314 on Build GRCh38
rs771454767 Chr.3: 113273154-113273154 on Build GRCh38
rs145076589 Chr.3: 113273293-113273293 on Build GRCh38
rs777871713 Chr.3: 113273215-113273215 on Build GRCh38
rs745375107 Chr.3: 113273542-113273542 on Build GRCh38
rs997310597 Chr.3: 113273586-113273586 on Build GRCh38
rs527592727 Chr.3: 113273248-113273248 on Build GRCh38
rs770082083 Chr.3: 113273286-113273286 on Build GRCh38
rs766110759 Chr.3: 113273322-113273322 on Build GRCh38
rs768070815 Chr.3: 113273236-113273236 on Build GRCh38
rs901153043 Chr.3: 113273197-113273197 on Build GRCh38
rs755673625 Chr.3: 113273367-113273367 on Build GRCh38
rs934907488 Chr.3: 113273472-113273472 on Build GRCh38
rs762343234 Chr.3: 113273167-113273167 on Build GRCh38
rs7612497 Chr.3: 113273378-113273378 on Build GRCh38
rs141430227 Chr.3: 113273273-113273273 on Build GRCh38
rs148670622 Chr.3: 113273204-113273204 on Build GRCh38
rs376112481 Chr.3: 113273232-113273232 on Build GRCh38
rs1002362887 Chr.3: 113273607-113273607 on Build GRCh38
rs754897087 Chr.3: 113273263-113273263 on Build GRCh38
rs370137874 Chr.3: 113273326-113273326 on Build GRCh38
rs1017659989 Chr.3: 113273546-113273546 on Build GRCh38
rs751240657 Chr.3: 113273172-113273172 on Build GRCh38
rs757611037 Chr.3: 113273277-113273277 on Build GRCh38
rs758161859 Chr.3: 113273304-113273304 on Build GRCh38
rs780414960 Chr.3: 113273287-113273287 on Build GRCh38
rs755540363 Chr.3: 113273363-113273363 on Build GRCh38
rs900605959 Chr.3: 113273548-113273548 on Build GRCh38
rs765361722 Chr.3: 113273223-113273223 on Build GRCh38
rs199618661 Chr.3: 113273355-113273355 on Build GRCh38
rs568473890 Chr.3: 113273443-113273443 on Build GRCh38
rs751737249 Chr.3: 113273336-113273336 on Build GRCh38
rs1040129108 Chr.3: 113273187-113273187 on Build GRCh38
rs779844168 Chr.3: 113273207-113273207 on Build GRCh38
rs541680773 Chr.3: 113273225-113273225 on Build GRCh38
rs560351015 Chr.3: 113273239-113273239 on Build GRCh38
rs556070204 Chr.3: 113273159-113273159 on Build GRCh38
rs369070396 Chr.3: 113273257-113273257 on Build GRCh38
rs78110980 Chr.3: 113273530-113273530 on Build GRCh38
rs761462481 Chr.3: 113273238-113273238 on Build GRCh38
rs373269156 Chr.3: 113273217-113273217 on Build GRCh38
rs144269826 Chr.3: 113273166-113273166 on Build GRCh38
rs763842993 Chr.3: 113273170-113273170 on Build GRCh38
rs1045880141 Chr.3: 113273392-113273392 on Build GRCh38
rs566283417 Chr.3: 113273327-113273327 on Build GRCh38
rs142142926 Chr.3: 113273209-113273209 on Build GRCh38
rs1053820345 Chr.3: 113273489-113273489 on Build GRCh38
rs566322186 Chr.3: 113273616-113273616 on Build GRCh38
rs1052664755 Chr.3: 113273478-113273478 on Build GRCh38
rs771602382 Chr.3: 113273390-113273390 on Build GRCh38
rs142197082 Chr.3: 113273262-113273262 on Build GRCh38
rs1011659174 Chr.3: 113273244-113273244 on Build GRCh38
rs371052682 Chr.3: 113273251-113273251 on Build GRCh38
rs775826071 Chr.3: 113273231-113273231 on Build GRCh38
rs761303691 Chr.3: 113273305-113273305 on Build GRCh38
rs781542207 Chr.3: 113273281-113273281 on Build GRCh38
rs147853055 Chr.3: 113273269-113273269 on Build GRCh38
rs757377586 Chr.3: 113273347-113273347 on Build GRCh38
rs905323751 Chr.3: 113273575-113273575 on Build GRCh38
rs372345745 Chr.3: 113273237-113273237 on Build GRCh38

View Full Product Details