Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 8413006-8413597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35B3
Entrez Gene ID
51000
Gene Name
solute carrier family 35 member B3
Gene Aliases
C6orf196, CGI-19, PAPST2
UniGene
Hs.285847
Gene Chromosome Location
Chr.6: 8413066-8435606 on Build GRCh38
Transcripts
NM_001142541.2 NM_015948.4 NR_109913.1 NR_109914.1 XM_005249156.3 XM_005249157.2 XM_005249158.1 XM_006715102.3 XM_006715103.3 XM_011514648.1 XM_017010910.1 XM_017010911.1 XM_017010912.1 XM_017010913.1 XM_017010914.1 XM_017010915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4856383 c.1187C>T p.T396M Chr.6: 8413568-8413568 on Build GRCh38
4799834 c.1194A>G p.A398A Chr.6: 8413561-8413561 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045722324 Chr.6: 8413386-8413386 on Build GRCh38
rs997225744 Chr.6: 8413541-8413541 on Build GRCh38
rs192836441 Chr.6: 8413408-8413408 on Build GRCh38
rs892950077 Chr.6: 8413482-8413482 on Build GRCh38
rs559359155 Chr.6: 8413443-8413443 on Build GRCh38
rs555066455 Chr.6: 8413214-8413214 on Build GRCh38
rs371917944 Chr.6: 8413446-8413446 on Build GRCh38
rs3799250 Chr.6: 8413150-8413150 on Build GRCh38
rs893169052 Chr.6: 8413108-8413108 on Build GRCh38
rs548491919 Chr.6: 8413145-8413149 on Build GRCh38
rs1007969541 Chr.6: 8413129-8413129 on Build GRCh38
rs367808428 Chr.6: 8413508-8413508 on Build GRCh38
rs41302899 Chr.6: 8413407-8413407 on Build GRCh38
rs138022408 Chr.6: 8413151-8413151 on Build GRCh38
rs558975652 Chr.6: 8413402-8413402 on Build GRCh38
rs909617402 Chr.6: 8413317-8413317 on Build GRCh38
rs915548359 Chr.6: 8413352-8413352 on Build GRCh38
rs957466066 Chr.6: 8413115-8413115 on Build GRCh38
rs749707666 Chr.6: 8413345-8413345 on Build GRCh38
rs1024330387 Chr.6: 8413040-8413040 on Build GRCh38
rs781412205 Chr.6: 8413565-8413565 on Build GRCh38
rs144682817 Chr.6: 8413567-8413567 on Build GRCh38
rs754670471 Chr.6: 8413212-8413212 on Build GRCh38
rs991547308 Chr.6: 8413346-8413346 on Build GRCh38
rs537683810 Chr.6: 8413400-8413400 on Build GRCh38
rs950423574 Chr.6: 8413562-8413562 on Build GRCh38
rs976638323 Chr.6: 8413209-8413209 on Build GRCh38
rs146315109 Chr.6: 8413101-8413102 on Build GRCh38
rs758789823 Chr.6: 8413526-8413526 on Build GRCh38
rs746990857 Chr.6: 8413441-8413441 on Build GRCh38
rs534204460 Chr.6: 8413333-8413333 on Build GRCh38
rs536311991 Chr.6: 8413210-8413210 on Build GRCh38
rs201176928 Chr.6: 8413550-8413550 on Build GRCh38
rs907084768 Chr.6: 8413044-8413044 on Build GRCh38
rs576537280 Chr.6: 8413331-8413331 on Build GRCh38
rs373312181 Chr.6: 8413530-8413530 on Build GRCh38
rs951155869 Chr.6: 8413215-8413215 on Build GRCh38
rs142499546 Chr.6: 8413546-8413546 on Build GRCh38
rs755583811 Chr.6: 8413505-8413505 on Build GRCh38
rs772035132 Chr.6: 8413362-8413362 on Build GRCh38
rs769872030 Chr.6: 8413568-8413568 on Build GRCh38
rs1011310848 Chr.6: 8413139-8413139 on Build GRCh38
rs747954614 Chr.6: 8413361-8413361 on Build GRCh38
rs15300 Chr.6: 8413179-8413179 on Build GRCh38
rs563433331 Chr.6: 8413519-8413519 on Build GRCh38
rs982539176 Chr.6: 8413232-8413232 on Build GRCh38

View Full Product Details