Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27432129-27432696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTK2B
Entrez Gene ID
2185
Gene Name
protein tyrosine kinase 2 beta
Gene Aliases
CADTK, CAKB, FADK2, FAK2, PKB, PTK, PYK2, RAFTK
UniGene
Hs.491322
Gene Chromosome Location
Chr.8: 27311482-27459391 on Build GRCh38
Transcripts
NM_004103.4 NM_173174.2 NM_173175.2 NM_173176.2 XM_005273447.4 XM_005273448.4 XM_011544441.2 XM_011544442.2 XM_017013214.1 XM_017013215.1 XM_017013216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3925088 c.888C>T p.P296P Chr.8: 27432262-27432262 on Build GRCh38
1728161 c.165G>T p.R55S Chr.8: 27432301-27432301 on Build GRCh38
5984399 c.199C>T p.Q67* Chr.8: 27432335-27432335 on Build GRCh38
5835536 c.979_980insC p.Q329fs*14 Chr.8: 27432353-27432354 on Build GRCh38
5988792 c.124-1G>T p.? Chr.8: 27432259-27432259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949588235 Chr.8: 27432575-27432575 on Build GRCh38
rs545561945 Chr.8: 27432160-27432160 on Build GRCh38
rs200116993 Chr.8: 27432274-27432274 on Build GRCh38
rs913621435 Chr.8: 27432434-27432434 on Build GRCh38
rs142980389 Chr.8: 27432360-27432360 on Build GRCh38
rs1055328717 Chr.8: 27432528-27432528 on Build GRCh38
rs41316468 Chr.8: 27432501-27432501 on Build GRCh38
rs748144988 Chr.8: 27432399-27432399 on Build GRCh38
rs769379468 Chr.8: 27432401-27432401 on Build GRCh38
rs752764255 Chr.8: 27432267-27432267 on Build GRCh38
rs201945609 Chr.8: 27432385-27432385 on Build GRCh38
rs1037168578 Chr.8: 27432576-27432576 on Build GRCh38
rs1032243554 Chr.8: 27432557-27432557 on Build GRCh38
rs552990505 Chr.8: 27432208-27432208 on Build GRCh38
rs759756524 Chr.8: 27432329-27432329 on Build GRCh38
rs367987503 Chr.8: 27432281-27432281 on Build GRCh38
rs965961492 Chr.8: 27432662-27432662 on Build GRCh38
rs770605978 Chr.8: 27432412-27432412 on Build GRCh38
rs944409112 Chr.8: 27432195-27432195 on Build GRCh38
rs1000734407 Chr.8: 27432552-27432552 on Build GRCh38
rs1007455901 Chr.8: 27432648-27432648 on Build GRCh38
rs910403311 Chr.8: 27432189-27432189 on Build GRCh38
rs761128691 Chr.8: 27432381-27432381 on Build GRCh38
rs780003250 Chr.8: 27432606-27432606 on Build GRCh38
rs557356317 Chr.8: 27432173-27432173 on Build GRCh38
rs902336990 Chr.8: 27432545-27432545 on Build GRCh38
rs751972320 Chr.8: 27432244-27432244 on Build GRCh38
rs187929803 Chr.8: 27432256-27432256 on Build GRCh38
rs774793270 Chr.8: 27432339-27432339 on Build GRCh38
rs559397091 Chr.8: 27432456-27432456 on Build GRCh38
rs756168261 Chr.8: 27432352-27432352 on Build GRCh38
rs979098010 Chr.8: 27432483-27432483 on Build GRCh38
rs577746675 Chr.8: 27432323-27432323 on Build GRCh38
rs755516438 Chr.8: 27432250-27432250 on Build GRCh38
rs139089526 Chr.8: 27432351-27432351 on Build GRCh38
rs370642375 Chr.8: 27432383-27432383 on Build GRCh38
rs779654031 Chr.8: 27432379-27432379 on Build GRCh38
rs766585609 Chr.8: 27432337-27432337 on Build GRCh38
rs889811237 Chr.8: 27432626-27432626 on Build GRCh38
rs142835408 Chr.8: 27432204-27432204 on Build GRCh38
rs961382278 Chr.8: 27432172-27432172 on Build GRCh38
rs763194097 Chr.8: 27432227-27432227 on Build GRCh38
rs200025953 Chr.8: 27432275-27432275 on Build GRCh38
rs756257912 Chr.8: 27432271-27432271 on Build GRCh38
rs763183706 Chr.8: 27432427-27432427 on Build GRCh38
rs996012790 Chr.8: 27432587-27432587 on Build GRCh38
rs777532166 Chr.8: 27432180-27432180 on Build GRCh38
rs1008352097 Chr.8: 27432488-27432488 on Build GRCh38
rs543628348 Chr.8: 27432346-27432346 on Build GRCh38
rs2303882 Chr.8: 27432446-27432446 on Build GRCh38
rs766814814 Chr.8: 27432233-27432233 on Build GRCh38
rs759952753 Chr.8: 27432342-27432342 on Build GRCh38
rs1017568814 Chr.8: 27432661-27432661 on Build GRCh38
rs752060023 Chr.8: 27432490-27432490 on Build GRCh38
rs746852655 Chr.8: 27432309-27432309 on Build GRCh38
rs975876285 Chr.8: 27432663-27432663 on Build GRCh38
rs368879406 Chr.8: 27432209-27432209 on Build GRCh38
rs942254707 Chr.8: 27432377-27432377 on Build GRCh38
rs772784294 Chr.8: 27432407-27432407 on Build GRCh38
rs776766099 Chr.8: 27432315-27432315 on Build GRCh38
rs564271202 Chr.8: 27432321-27432321 on Build GRCh38
rs749929617 Chr.8: 27432232-27432233 on Build GRCh38
rs959253996 Chr.8: 27432565-27432565 on Build GRCh38
rs143285330 Chr.8: 27432310-27432310 on Build GRCh38
rs764184338 Chr.8: 27432355-27432355 on Build GRCh38
rs551204665 Chr.8: 27432581-27432581 on Build GRCh38
rs945059735 Chr.8: 27432220-27432220 on Build GRCh38
rs754886376 Chr.8: 27432394-27432394 on Build GRCh38
rs776687198 Chr.8: 27432286-27432286 on Build GRCh38
rs561872084 Chr.8: 27432428-27432428 on Build GRCh38
rs985941178 Chr.8: 27432186-27432186 on Build GRCh38
rs2303881 Chr.8: 27432418-27432418 on Build GRCh38
rs111383360 Chr.8: 27432572-27432572 on Build GRCh38
rs1024947480 Chr.8: 27432621-27432621 on Build GRCh38
rs764988584 Chr.8: 27432368-27432368 on Build GRCh38
rs779925728 Chr.8: 27432308-27432308 on Build GRCh38
rs890803761 Chr.8: 27432466-27432466 on Build GRCh38
rs781242371 Chr.8: 27432396-27432396 on Build GRCh38
rs34612102 Chr.8: 27432182-27432183 on Build GRCh38
rs748835098 Chr.8: 27432408-27432408 on Build GRCh38
rs182314206 Chr.8: 27432181-27432181 on Build GRCh38
rs965889617 Chr.8: 27432420-27432420 on Build GRCh38
rs897153010 Chr.8: 27432314-27432314 on Build GRCh38
rs966570637 Chr.8: 27432249-27432249 on Build GRCh38

View Full Product Details