Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12897162-12897741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC3
Entrez Gene ID
83643
Gene Name
coiled-coil domain containing 3
Gene Aliases
-
UniGene
Hs.498720
Gene Chromosome Location
Chr.10: 12896625-13099773 on Build GRCh38
Transcripts
NM_001282658.1 NM_031455.3

SNP Detail

SNP ID Chromosome Location
rs1034944705 Chr.10: 12897199-12897199 on Build GRCh38
rs372851665 Chr.10: 12897529-12897531 on Build GRCh38
rs767101107 Chr.10: 12897390-12897390 on Build GRCh38
rs550581572 Chr.10: 12897423-12897423 on Build GRCh38
rs1004188466 Chr.10: 12897670-12897670 on Build GRCh38
rs183939790 Chr.10: 12897267-12897267 on Build GRCh38
rs900220293 Chr.10: 12897270-12897270 on Build GRCh38
rs1051492090 Chr.10: 12897493-12897493 on Build GRCh38
rs894677391 Chr.10: 12897605-12897605 on Build GRCh38
rs896445265 Chr.10: 12897282-12897282 on Build GRCh38
rs778871963 Chr.10: 12897195-12897196 on Build GRCh38
rs540282731 Chr.10: 12897295-12897295 on Build GRCh38
rs1019145551 Chr.10: 12897548-12897548 on Build GRCh38
rs551692564 Chr.10: 12897613-12897614 on Build GRCh38
rs543626598 Chr.10: 12897456-12897456 on Build GRCh38
rs531488033 Chr.10: 12897417-12897417 on Build GRCh38
rs890156837 Chr.10: 12897341-12897341 on Build GRCh38
rs188713706 Chr.10: 12897468-12897468 on Build GRCh38
rs1056365264 Chr.10: 12897604-12897604 on Build GRCh38
rs1020522245 Chr.10: 12897349-12897349 on Build GRCh38
rs907771766 Chr.10: 12897427-12897427 on Build GRCh38
rs751290040 Chr.10: 12897201-12897201 on Build GRCh38
rs945739660 Chr.10: 12897529-12897529 on Build GRCh38
rs756706946 Chr.10: 12897383-12897383 on Build GRCh38
rs1034532829 Chr.10: 12897372-12897372 on Build GRCh38
rs1024304171 Chr.10: 12897401-12897401 on Build GRCh38
rs111965452 Chr.10: 12897694-12897694 on Build GRCh38
rs41291305 Chr.10: 12897681-12897681 on Build GRCh38
rs565495509 Chr.10: 12897658-12897658 on Build GRCh38
rs989424358 Chr.10: 12897361-12897361 on Build GRCh38
rs941882658 Chr.10: 12897187-12897187 on Build GRCh38
rs886230665 Chr.10: 12897420-12897420 on Build GRCh38
rs748335331 Chr.10: 12897450-12897452 on Build GRCh38
rs1024421898 Chr.10: 12897652-12897652 on Build GRCh38
rs1010435559 Chr.10: 12897348-12897348 on Build GRCh38
rs886285277 Chr.10: 12897182-12897182 on Build GRCh38
rs138575629 Chr.10: 12897710-12897711 on Build GRCh38
rs952081069 Chr.10: 12897575-12897575 on Build GRCh38
rs148278349 Chr.10: 12897553-12897553 on Build GRCh38
rs1014337529 Chr.10: 12897644-12897644 on Build GRCh38
rs76880294 Chr.10: 12897363-12897363 on Build GRCh38
rs963341073 Chr.10: 12897432-12897432 on Build GRCh38
rs560098247 Chr.10: 12897300-12897300 on Build GRCh38
rs931718569 Chr.10: 12897485-12897485 on Build GRCh38
rs921894625 Chr.10: 12897566-12897566 on Build GRCh38
rs570389750 Chr.10: 12897509-12897509 on Build GRCh38
rs531862831 Chr.10: 12897412-12897412 on Build GRCh38
rs992966216 Chr.10: 12897400-12897400 on Build GRCh38
rs533257070 Chr.10: 12897481-12897481 on Build GRCh38
rs529127197 Chr.10: 12897333-12897333 on Build GRCh38
rs921945524 Chr.10: 12897398-12897398 on Build GRCh38
rs1044965785 Chr.10: 12897707-12897707 on Build GRCh38
rs768646739 Chr.10: 12897617-12897618 on Build GRCh38
rs1000746719 Chr.10: 12897364-12897364 on Build GRCh38
rs536072500 Chr.10: 12897554-12897554 on Build GRCh38
rs1000457242 Chr.10: 12897565-12897565 on Build GRCh38
rs1051329830 Chr.10: 12897332-12897332 on Build GRCh38
rs963585153 Chr.10: 12897692-12897692 on Build GRCh38
rs562830178 Chr.10: 12897413-12897413 on Build GRCh38
rs369006506 Chr.10: 12897574-12897574 on Build GRCh38
rs530780938 Chr.10: 12897285-12897285 on Build GRCh38
rs569585457 Chr.10: 12897676-12897676 on Build GRCh38
rs550579224 Chr.10: 12897459-12897459 on Build GRCh38
rs940531417 Chr.10: 12897426-12897426 on Build GRCh38
rs371649149 Chr.10: 12897510-12897510 on Build GRCh38

View Full Product Details