Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111720314-111720899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R1B
Entrez Gene ID
5519
Gene Name
protein phosphatase 2 scaffold subunit Abeta
Gene Aliases
PP2A-Abeta, PR65B
UniGene
Hs.269128
Gene Chromosome Location
Chr.11: 111694569-111766445 on Build GRCh38
Transcripts
XM_017017958.1 XM_017017959.1 NM_015191.2 XM_017017417.1 XM_017017418.1

Gene Symbol
SIK2
Entrez Gene ID
23235
Gene Name
salt inducible kinase 2
Gene Aliases
LOH11CR1I, QIK, SIK-2, SNF1LK2
UniGene
Hs.269128
Gene Chromosome Location
Chr.11: 111602391-111726917 on Build GRCh38
Transcripts
XM_017017958.1 XM_017017959.1 NM_015191.2 XM_017017417.1 XM_017017418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5747326 c.1780+2T>C p.? Chr.11: 111720764-111720764 on Build GRCh38
1225733 c.1528C>A p.L510I Chr.11: 111720510-111720510 on Build GRCh38
4726078 c.1648T>G p.S550A Chr.11: 111720630-111720630 on Build GRCh38
5771167 c.1518C>T p.D506D Chr.11: 111720500-111720500 on Build GRCh38
4421965 c.1502T>C p.I501T Chr.11: 111720484-111720484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992170779 Chr.11: 111720419-111720419 on Build GRCh38
rs369707716 Chr.11: 111720779-111720779 on Build GRCh38
rs763496004 Chr.11: 111720642-111720642 on Build GRCh38
rs763750371 Chr.11: 111720454-111720454 on Build GRCh38
rs375806207 Chr.11: 111720800-111720800 on Build GRCh38
rs774872028 Chr.11: 111720796-111720796 on Build GRCh38
rs1042370650 Chr.11: 111720449-111720449 on Build GRCh38
rs766787024 Chr.11: 111720438-111720438 on Build GRCh38
rs765056611 Chr.11: 111720469-111720469 on Build GRCh38
rs761208814 Chr.11: 111720643-111720643 on Build GRCh38
rs765388415 Chr.11: 111720869-111720869 on Build GRCh38
rs372375521 Chr.11: 111720621-111720621 on Build GRCh38
rs764788389 Chr.11: 111720810-111720810 on Build GRCh38
rs1004315116 Chr.11: 111720668-111720669 on Build GRCh38
rs763697358 Chr.11: 111720626-111720626 on Build GRCh38
rs372524597 Chr.11: 111720799-111720799 on Build GRCh38
rs113067223 Chr.11: 111720558-111720558 on Build GRCh38
rs974249006 Chr.11: 111720381-111720381 on Build GRCh38
rs749858828 Chr.11: 111720580-111720580 on Build GRCh38
rs1029396126 Chr.11: 111720639-111720639 on Build GRCh38
rs574153579 Chr.11: 111720444-111720444 on Build GRCh38
rs760254866 Chr.11: 111720446-111720446 on Build GRCh38
rs750426639 Chr.11: 111720482-111720482 on Build GRCh38
rs971645225 Chr.11: 111720771-111720771 on Build GRCh38
rs79329900 Chr.11: 111720866-111720866 on Build GRCh38
rs769964011 Chr.11: 111720536-111720536 on Build GRCh38
rs753261608 Chr.11: 111720581-111720581 on Build GRCh38
rs761459308 Chr.11: 111720804-111720804 on Build GRCh38
rs748458348 Chr.11: 111720752-111720752 on Build GRCh38
rs777841376 Chr.11: 111720774-111720774 on Build GRCh38
rs756143004 Chr.11: 111720761-111720761 on Build GRCh38
rs901130677 Chr.11: 111720491-111720491 on Build GRCh38
rs751913897 Chr.11: 111720795-111720795 on Build GRCh38
rs765910823 Chr.11: 111720506-111720506 on Build GRCh38
rs763203934 Chr.11: 111720437-111720437 on Build GRCh38
rs181921625 Chr.11: 111720349-111720349 on Build GRCh38
rs764862542 Chr.11: 111720651-111720651 on Build GRCh38
rs780065873 Chr.11: 111720489-111720489 on Build GRCh38
rs773578194 Chr.11: 111720566-111720566 on Build GRCh38
rs761867247 Chr.11: 111720868-111720869 on Build GRCh38
rs781246925 Chr.11: 111720734-111720734 on Build GRCh38
rs375896845 Chr.11: 111720713-111720713 on Build GRCh38
rs771432649 Chr.11: 111720586-111720586 on Build GRCh38
rs369609954 Chr.11: 111720691-111720691 on Build GRCh38
rs558980001 Chr.11: 111720797-111720797 on Build GRCh38
rs1005278667 Chr.11: 111720567-111720567 on Build GRCh38
rs748142533 Chr.11: 111720527-111720527 on Build GRCh38
rs758349007 Chr.11: 111720712-111720712 on Build GRCh38
rs990651226 Chr.11: 111720400-111720400 on Build GRCh38
rs759419734 Chr.11: 111720873-111720873 on Build GRCh38
rs368912230 Chr.11: 111720853-111720853 on Build GRCh38
rs750326950 Chr.11: 111720656-111720656 on Build GRCh38
rs201616979 Chr.11: 111720492-111720492 on Build GRCh38
rs943365259 Chr.11: 111720440-111720440 on Build GRCh38
rs890862471 Chr.11: 111720650-111720650 on Build GRCh38
rs781504260 Chr.11: 111720514-111720514 on Build GRCh38
rs145846639 Chr.11: 111720411-111720411 on Build GRCh38
rs910527348 Chr.11: 111720417-111720417 on Build GRCh38
rs759072318 Chr.11: 111720423-111720423 on Build GRCh38
rs1017099854 Chr.11: 111720617-111720617 on Build GRCh38
rs770585395 Chr.11: 111720352-111720352 on Build GRCh38
rs146609137 Chr.11: 111720874-111720874 on Build GRCh38
rs199644978 Chr.11: 111720622-111720622 on Build GRCh38
rs555719228 Chr.11: 111720427-111720427 on Build GRCh38
rs1054329136 Chr.11: 111720542-111720542 on Build GRCh38
rs772910998 Chr.11: 111720851-111720851 on Build GRCh38
rs776051348 Chr.11: 111720363-111720363 on Build GRCh38
rs369706750 Chr.11: 111720590-111720590 on Build GRCh38
rs1046963022 Chr.11: 111720538-111720538 on Build GRCh38
rs754857866 Chr.11: 111720728-111720728 on Build GRCh38
rs753239574 Chr.11: 111720455-111720455 on Build GRCh38
rs185077107 Chr.11: 111720457-111720457 on Build GRCh38

View Full Product Details