Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41048506-41049094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCMH1
Entrez Gene ID
22955
Gene Name
sex comb on midleg homolog 1 (Drosophila)
Gene Aliases
Scml3
UniGene
Hs.571874
Gene Chromosome Location
Chr.1: 41027199-41242155 on Build GRCh38
Transcripts
NM_001031694.2 NM_001172218.1 NM_001172219.1 NM_001172220.1 NM_001172221.1 NM_001172222.2 NM_012236.3 NR_135092.1 XM_006710462.2 XM_006710464.1 XM_011541032.2 XM_011541033.2 XM_011541034.1 XM_011541035.2 XM_011541036.2 XM_011541039.1 XM_011541040.2 XM_011541043.2 XM_011541044.1 XM_011541045.2 XM_017000698.1 XM_017000699.1 XM_017000700.1 XM_017000701.1 XM_017000702.1 XM_017000703.1 XM_017000704.1 XM_017000705.1 XM_017000706.1 XM_017000707.1 XM_017000708.1 XM_017000709.1 XM_017000710.1 XM_017000711.1 XM_017000712.1 XM_017000713.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4723960 c.1105A>G p.K369E Chr.1: 41048720-41048720 on Build GRCh38
4411205 c.1229C>T p.A410V Chr.1: 41048767-41048767 on Build GRCh38
7134035 c.1031C>G p.S344C Chr.1: 41048794-41048794 on Build GRCh38
2169505 c.1199C>T p.A400V Chr.1: 41048797-41048797 on Build GRCh38
909545 c.1085A>G p.Y362C Chr.1: 41048881-41048881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993579667 Chr.1: 41048958-41048958 on Build GRCh38
rs890931899 Chr.1: 41049034-41049034 on Build GRCh38
rs908228594 Chr.1: 41048600-41048600 on Build GRCh38
rs982908179 Chr.1: 41048594-41048594 on Build GRCh38
rs766342413 Chr.1: 41048976-41048976 on Build GRCh38
rs184938736 Chr.1: 41048546-41048546 on Build GRCh38
rs758754119 Chr.1: 41048901-41048901 on Build GRCh38
rs866575987 Chr.1: 41048939-41048939 on Build GRCh38
rs1010800361 Chr.1: 41048533-41048533 on Build GRCh38
rs766171836 Chr.1: 41048733-41048733 on Build GRCh38
rs142187533 Chr.1: 41048783-41048783 on Build GRCh38
rs759445717 Chr.1: 41048881-41048881 on Build GRCh38
rs1029509333 Chr.1: 41049021-41049021 on Build GRCh38
rs910752223 Chr.1: 41048630-41048630 on Build GRCh38
rs772643140 Chr.1: 41048776-41048776 on Build GRCh38
rs773073621 Chr.1: 41048856-41048856 on Build GRCh38
rs1015342370 Chr.1: 41049070-41049070 on Build GRCh38
rs546900944 Chr.1: 41048877-41048877 on Build GRCh38
rs981636710 Chr.1: 41048781-41048781 on Build GRCh38
rs371495684 Chr.1: 41048668-41048668 on Build GRCh38
rs760818371 Chr.1: 41048986-41048986 on Build GRCh38
rs755732244 Chr.1: 41048751-41048751 on Build GRCh38
rs996686959 Chr.1: 41048570-41048570 on Build GRCh38
rs773459329 Chr.1: 41048572-41048572 on Build GRCh38
rs776823395 Chr.1: 41048835-41048835 on Build GRCh38
rs145683152 Chr.1: 41048896-41048896 on Build GRCh38
rs112636959 Chr.1: 41048786-41048786 on Build GRCh38
rs759167321 Chr.1: 41048611-41048611 on Build GRCh38
rs997131468 Chr.1: 41048990-41048990 on Build GRCh38
rs986404500 Chr.1: 41049000-41049000 on Build GRCh38
rs572879089 Chr.1: 41048542-41048542 on Build GRCh38
rs151194966 Chr.1: 41048760-41048760 on Build GRCh38
rs754513115 Chr.1: 41048736-41048736 on Build GRCh38
rs750802911 Chr.1: 41048900-41048900 on Build GRCh38
rs376970948 Chr.1: 41048800-41048800 on Build GRCh38
rs1030372001 Chr.1: 41048968-41048968 on Build GRCh38
rs893167613 Chr.1: 41048853-41048853 on Build GRCh38
rs537974913 Chr.1: 41049043-41049043 on Build GRCh38
rs186734971 Chr.1: 41048894-41048894 on Build GRCh38
rs148739697 Chr.1: 41048735-41048735 on Build GRCh38
rs561862007 Chr.1: 41048603-41048603 on Build GRCh38
rs141985362 Chr.1: 41048844-41048844 on Build GRCh38
rs962406202 Chr.1: 41049074-41049074 on Build GRCh38
rs945738777 Chr.1: 41048701-41048701 on Build GRCh38
rs368195731 Chr.1: 41048748-41048748 on Build GRCh38
rs1003990279 Chr.1: 41049044-41049044 on Build GRCh38
rs955157388 Chr.1: 41048580-41048580 on Build GRCh38
rs745878448 Chr.1: 41048677-41048677 on Build GRCh38
rs752161618 Chr.1: 41048888-41048888 on Build GRCh38
rs936815138 Chr.1: 41048632-41048632 on Build GRCh38
rs767052739 Chr.1: 41048885-41048885 on Build GRCh38
rs934776711 Chr.1: 41048836-41048836 on Build GRCh38
rs762588000 Chr.1: 41048862-41048862 on Build GRCh38
rs1054229714 Chr.1: 41048658-41048658 on Build GRCh38
rs191547953 Chr.1: 41049028-41049028 on Build GRCh38
rs1022154163 Chr.1: 41048543-41048543 on Build GRCh38
rs762161709 Chr.1: 41048843-41048843 on Build GRCh38
rs138961580 Chr.1: 41048747-41048747 on Build GRCh38
rs138234894 Chr.1: 41048954-41048954 on Build GRCh38
rs962354112 Chr.1: 41048656-41048656 on Build GRCh38
rs779280545 Chr.1: 41048759-41048759 on Build GRCh38
rs1053112950 Chr.1: 41048837-41048837 on Build GRCh38
rs766193595 Chr.1: 41048863-41048863 on Build GRCh38
rs768909374 Chr.1: 41048832-41048832 on Build GRCh38

View Full Product Details