Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68125183-68125778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG12-AS1
Entrez Gene ID
100289178
Gene Name
GNG12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.677603
Gene Chromosome Location
Chr.1: 67832288-68202987 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 NM_001193334.1 NM_024911.6 XM_011542191.2 XM_011542192.2 XM_017002390.1

Gene Symbol
WLS
Entrez Gene ID
79971
Gene Name
wntless Wnt ligand secretion mediator
Gene Aliases
C1orf139, EVI, GPR177, MRP, mig-14
UniGene
Hs.647659
Gene Chromosome Location
Chr.1: 68098459-68233000 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 NM_001193334.1 NM_024911.6 XM_011542191.2 XM_011542192.2 XM_017002390.1

SNP Detail

SNP ID Chromosome Location
rs41287844 Chr.1: 68125512-68125512 on Build GRCh38
rs999030700 Chr.1: 68125709-68125709 on Build GRCh38
rs75116509 Chr.1: 68125378-68125378 on Build GRCh38
rs944029960 Chr.1: 68125368-68125368 on Build GRCh38
rs924389160 Chr.1: 68125474-68125474 on Build GRCh38
rs544835151 Chr.1: 68125745-68125746 on Build GRCh38
rs894505384 Chr.1: 68125637-68125637 on Build GRCh38
rs939893032 Chr.1: 68125551-68125551 on Build GRCh38
rs971546733 Chr.1: 68125697-68125697 on Build GRCh38
rs975789751 Chr.1: 68125574-68125575 on Build GRCh38
rs115461003 Chr.1: 68125475-68125475 on Build GRCh38
rs1046687072 Chr.1: 68125730-68125730 on Build GRCh38
rs899752786 Chr.1: 68125204-68125204 on Build GRCh38
rs932460965 Chr.1: 68125237-68125237 on Build GRCh38
rs1055229202 Chr.1: 68125747-68125747 on Build GRCh38
rs534492526 Chr.1: 68125384-68125384 on Build GRCh38
rs994774615 Chr.1: 68125617-68125617 on Build GRCh38
rs756695541 Chr.1: 68125702-68125706 on Build GRCh38
rs1046792 Chr.1: 68125636-68125636 on Build GRCh38
rs34670698 Chr.1: 68125545-68125546 on Build GRCh38
rs779542466 Chr.1: 68125606-68125606 on Build GRCh38
rs994533816 Chr.1: 68125477-68125477 on Build GRCh38
rs568949897 Chr.1: 68125302-68125302 on Build GRCh38
rs577402732 Chr.1: 68125398-68125403 on Build GRCh38
rs74081379 Chr.1: 68125276-68125276 on Build GRCh38
rs973189612 Chr.1: 68125288-68125288 on Build GRCh38
rs890992484 Chr.1: 68125280-68125280 on Build GRCh38
rs191974585 Chr.1: 68125351-68125351 on Build GRCh38
rs1013566608 Chr.1: 68125651-68125651 on Build GRCh38
rs945647089 Chr.1: 68125415-68125415 on Build GRCh38
rs540418050 Chr.1: 68125699-68125699 on Build GRCh38
rs567534054 Chr.1: 68125410-68125410 on Build GRCh38
rs7062 Chr.1: 68125570-68125570 on Build GRCh38
rs374476437 Chr.1: 68125444-68125444 on Build GRCh38
rs1046835 Chr.1: 68125490-68125490 on Build GRCh38
rs269350 Chr.1: 68125722-68125722 on Build GRCh38
rs751917264 Chr.1: 68125559-68125559 on Build GRCh38
rs528849751 Chr.1: 68125581-68125581 on Build GRCh38
rs1046852 Chr.1: 68125421-68125421 on Build GRCh38
rs754254759 Chr.1: 68125532-68125532 on Build GRCh38
rs542611320 Chr.1: 68125647-68125647 on Build GRCh38
rs531423305 Chr.1: 68125724-68125724 on Build GRCh38
rs906209303 Chr.1: 68125430-68125430 on Build GRCh38
rs577463582 Chr.1: 68125501-68125501 on Build GRCh38
rs556356550 Chr.1: 68125441-68125441 on Build GRCh38
rs767591435 Chr.1: 68125429-68125429 on Build GRCh38
rs1024597548 Chr.1: 68125662-68125662 on Build GRCh38
rs892618153 Chr.1: 68125411-68125411 on Build GRCh38
rs1015002816 Chr.1: 68125283-68125283 on Build GRCh38
rs556902885 Chr.1: 68125281-68125281 on Build GRCh38
rs6695028 Chr.1: 68125357-68125357 on Build GRCh38
rs568370289 Chr.1: 68125350-68125350 on Build GRCh38
rs1026204208 Chr.1: 68125297-68125297 on Build GRCh38
rs750487877 Chr.1: 68125462-68125462 on Build GRCh38
rs906756637 Chr.1: 68125736-68125736 on Build GRCh38
rs1051046736 Chr.1: 68125521-68125521 on Build GRCh38
rs1049542734 Chr.1: 68125633-68125633 on Build GRCh38
rs543964530 Chr.1: 68125489-68125489 on Build GRCh38
rs1002240248 Chr.1: 68125434-68125434 on Build GRCh38
rs554532397 Chr.1: 68125403-68125403 on Build GRCh38
rs959232273 Chr.1: 68125395-68125395 on Build GRCh38
rs989560446 Chr.1: 68125700-68125702 on Build GRCh38
rs921261456 Chr.1: 68125580-68125580 on Build GRCh38
rs906185962 Chr.1: 68125238-68125239 on Build GRCh38
rs35053415 Chr.1: 68125251-68125252 on Build GRCh38

View Full Product Details