Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9877990-9878587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIDEC
Entrez Gene ID
63924
Gene Name
cell death inducing DFFA like effector c
Gene Aliases
CIDE-3, CIDE3, FPLD5, FSP27
UniGene
Hs.567562
Gene Chromosome Location
Chr.3: 9866710-9880254 on Build GRCh38
Transcripts
NM_001199551.1 NM_001199552.1 NM_001199623.1 NM_001321142.1 NM_001321143.1 NM_001321144.1 NM_022094.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4749798 c.10G>A p.A4T Chr.3: 9878477-9878477 on Build GRCh38
2950746 c.20C>T p.S7F Chr.3: 9878467-9878467 on Build GRCh38
4158700 c.46C>T p.L16F Chr.3: 9878441-9878441 on Build GRCh38
4749797 c.10G>A p.A4T Chr.3: 9878477-9878477 on Build GRCh38
4158699 c.46C>T p.L16F Chr.3: 9878441-9878441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78314436 Chr.3: 9878444-9878444 on Build GRCh38
rs768704792 Chr.3: 9878556-9878556 on Build GRCh38
rs1019429226 Chr.3: 9878170-9878170 on Build GRCh38
rs910493100 Chr.3: 9878505-9878505 on Build GRCh38
rs112252671 Chr.3: 9878430-9878430 on Build GRCh38
rs957117903 Chr.3: 9878526-9878526 on Build GRCh38
rs758898250 Chr.3: 9878388-9878388 on Build GRCh38
rs1037012144 Chr.3: 9878377-9878377 on Build GRCh38
rs1029924160 Chr.3: 9878470-9878470 on Build GRCh38
rs777278439 Chr.3: 9878521-9878521 on Build GRCh38
rs745695964 Chr.3: 9878540-9878540 on Build GRCh38
rs868411171 Chr.3: 9878420-9878420 on Build GRCh38
rs1045629935 Chr.3: 9878250-9878250 on Build GRCh38
rs770512970 Chr.3: 9878448-9878448 on Build GRCh38
rs927682880 Chr.3: 9878372-9878372 on Build GRCh38
rs7631805 Chr.3: 9878143-9878143 on Build GRCh38
rs756925680 Chr.3: 9878532-9878532 on Build GRCh38
rs375015077 Chr.3: 9878118-9878118 on Build GRCh38
rs901966007 Chr.3: 9878177-9878177 on Build GRCh38
rs1048983036 Chr.3: 9878133-9878133 on Build GRCh38
rs372098782 Chr.3: 9878034-9878034 on Build GRCh38
rs746959494 Chr.3: 9878547-9878547 on Build GRCh38
rs775474059 Chr.3: 9878467-9878467 on Build GRCh38
rs912905985 Chr.3: 9878242-9878242 on Build GRCh38
rs546019069 Chr.3: 9878017-9878017 on Build GRCh38
rs943353452 Chr.3: 9878529-9878529 on Build GRCh38
rs772659984 Chr.3: 9878406-9878406 on Build GRCh38
rs774070829 Chr.3: 9878451-9878451 on Build GRCh38
rs368499736 Chr.3: 9878506-9878506 on Build GRCh38
rs1016039750 Chr.3: 9878024-9878024 on Build GRCh38
rs780580716 Chr.3: 9878389-9878389 on Build GRCh38
rs771920426 Chr.3: 9878542-9878542 on Build GRCh38
rs764297186 Chr.3: 9878477-9878477 on Build GRCh38
rs925724848 Chr.3: 9878169-9878169 on Build GRCh38
rs550778646 Chr.3: 9878050-9878050 on Build GRCh38
rs1029808533 Chr.3: 9878121-9878121 on Build GRCh38
rs34192167 Chr.3: 9878108-9878108 on Build GRCh38
rs930510077 Chr.3: 9878130-9878130 on Build GRCh38
rs971811671 Chr.3: 9878446-9878446 on Build GRCh38
rs776798648 Chr.3: 9878478-9878478 on Build GRCh38
rs754440049 Chr.3: 9878292-9878292 on Build GRCh38
rs755409925 Chr.3: 9878518-9878518 on Build GRCh38
rs557130147 Chr.3: 9878428-9878428 on Build GRCh38
rs775104397 Chr.3: 9878544-9878544 on Build GRCh38
rs758769971 Chr.3: 9878504-9878504 on Build GRCh38
rs753445910 Chr.3: 9878244-9878244 on Build GRCh38
rs553018745 Chr.3: 9878322-9878322 on Build GRCh38
rs1005543973 Chr.3: 9878016-9878016 on Build GRCh38
rs998969650 Chr.3: 9878187-9878187 on Build GRCh38
rs546870625 Chr.3: 9878562-9878563 on Build GRCh38
rs985171256 Chr.3: 9878166-9878166 on Build GRCh38
rs779667177 Chr.3: 9878215-9878215 on Build GRCh38
rs765494548 Chr.3: 9878485-9878485 on Build GRCh38
rs555769737 Chr.3: 9878146-9878146 on Build GRCh38
rs17851444 Chr.3: 9878454-9878454 on Build GRCh38
rs56111543 Chr.3: 9878357-9878357 on Build GRCh38
rs567505799 Chr.3: 9878207-9878207 on Build GRCh38
rs988484288 Chr.3: 9878226-9878226 on Build GRCh38
rs374888110 Chr.3: 9878558-9878558 on Build GRCh38
rs1006072971 Chr.3: 9878364-9878364 on Build GRCh38
rs748783569 Chr.3: 9878528-9878528 on Build GRCh38
rs371554594 Chr.3: 9878536-9878536 on Build GRCh38
rs777225567 Chr.3: 9878429-9878429 on Build GRCh38
rs62247501 Chr.3: 9878563-9878563 on Build GRCh38
rs1031838724 Chr.3: 9878200-9878200 on Build GRCh38
rs577702648 Chr.3: 9878325-9878325 on Build GRCh38
rs771940288 Chr.3: 9878460-9878460 on Build GRCh38
rs762188260 Chr.3: 9878484-9878484 on Build GRCh38
rs980898662 Chr.3: 9878020-9878020 on Build GRCh38
rs116397880 Chr.3: 9878284-9878284 on Build GRCh38
rs145181634 Chr.3: 9878495-9878495 on Build GRCh38

View Full Product Details