Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61188665-61189205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
UniGene
Hs.655602
Gene Chromosome Location
Chr.2: 61177418-61191210 on Build GRCh38
Transcripts
NM_001321305.1 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1 NM_014709.3

Gene Symbol
USP34
Entrez Gene ID
9736
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
-
UniGene
Hs.644708
Gene Chromosome Location
Chr.2: 61187455-61470714 on Build GRCh38
Transcripts
NM_001321305.1 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1 NM_014709.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
106756 c.10022G>A p.R3341K Chr.2: 61188921-61188921 on Build GRCh38
1022059 c.10067G>A p.R3356H Chr.2: 61188676-61188676 on Build GRCh38
4624838 c.10047A>G p.A3349A Chr.2: 61188696-61188696 on Build GRCh38
5862675 c.10061G>A p.R3354K Chr.2: 61188682-61188682 on Build GRCh38
1306913 c.10005G>A p.E3335E Chr.2: 61188938-61188938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944132741 Chr.2: 61188817-61188817 on Build GRCh38
rs139589738 Chr.2: 61189167-61189167 on Build GRCh38
rs765196343 Chr.2: 61189093-61189093 on Build GRCh38
rs191286986 Chr.2: 61189124-61189124 on Build GRCh38
rs747798160 Chr.2: 61188977-61188977 on Build GRCh38
rs752350929 Chr.2: 61188718-61188719 on Build GRCh38
rs752201352 Chr.2: 61188915-61188915 on Build GRCh38
rs772611714 Chr.2: 61188992-61188992 on Build GRCh38
rs946240783 Chr.2: 61188792-61188792 on Build GRCh38
rs1057249444 Chr.2: 61189171-61189171 on Build GRCh38
rs751610064 Chr.2: 61189034-61189034 on Build GRCh38
rs150846404 Chr.2: 61189157-61189157 on Build GRCh38
rs1002964594 Chr.2: 61188767-61188767 on Build GRCh38
rs1039720183 Chr.2: 61188703-61188705 on Build GRCh38
rs757317357 Chr.2: 61189040-61189040 on Build GRCh38
rs931507702 Chr.2: 61188758-61188758 on Build GRCh38
rs1057431377 Chr.2: 61188768-61188768 on Build GRCh38
rs541184731 Chr.2: 61188899-61188899 on Build GRCh38
rs990448898 Chr.2: 61188772-61188772 on Build GRCh38
rs375328150 Chr.2: 61189000-61189000 on Build GRCh38
rs912706198 Chr.2: 61188804-61188804 on Build GRCh38
rs554348858 Chr.2: 61188805-61188805 on Build GRCh38
rs753599649 Chr.2: 61188913-61188913 on Build GRCh38
rs531770634 Chr.2: 61189061-61189061 on Build GRCh38
rs575670399 Chr.2: 61188771-61188771 on Build GRCh38
rs760534552 Chr.2: 61188898-61188898 on Build GRCh38
rs559287513 Chr.2: 61188955-61188955 on Build GRCh38
rs1021580181 Chr.2: 61188842-61188842 on Build GRCh38
rs777707567 Chr.2: 61188949-61188949 on Build GRCh38
rs202199016 Chr.2: 61188748-61188748 on Build GRCh38
rs751787145 Chr.2: 61189118-61189118 on Build GRCh38
rs749597720 Chr.2: 61189072-61189072 on Build GRCh38
rs564470263 Chr.2: 61189055-61189055 on Build GRCh38
rs771390663 Chr.2: 61188761-61188761 on Build GRCh38
rs764315934 Chr.2: 61189028-61189028 on Build GRCh38
rs746273735 Chr.2: 61188960-61188960 on Build GRCh38
rs755265940 Chr.2: 61188723-61188723 on Build GRCh38
rs1044562928 Chr.2: 61188762-61188762 on Build GRCh38
rs747457588 Chr.2: 61188752-61188752 on Build GRCh38
rs769740124 Chr.2: 61188821-61188822 on Build GRCh38
rs771351711 Chr.2: 61188956-61188956 on Build GRCh38
rs759834130 Chr.2: 61188875-61188875 on Build GRCh38
rs772245742 Chr.2: 61188742-61188742 on Build GRCh38
rs920156806 Chr.2: 61189063-61189063 on Build GRCh38
rs550350694 Chr.2: 61189156-61189156 on Build GRCh38
rs764251323 Chr.2: 61189116-61189116 on Build GRCh38
rs905925762 Chr.2: 61188766-61188766 on Build GRCh38
rs774051253 Chr.2: 61188975-61188975 on Build GRCh38
rs781601287 Chr.2: 61189045-61189045 on Build GRCh38
rs752944776 Chr.2: 61188930-61188930 on Build GRCh38
rs770124669 Chr.2: 61188962-61188962 on Build GRCh38
rs372146795 Chr.2: 61188895-61188895 on Build GRCh38
rs746539744 Chr.2: 61189085-61189085 on Build GRCh38
rs565509446 Chr.2: 61188765-61188765 on Build GRCh38
rs776676252 Chr.2: 61189014-61189014 on Build GRCh38
rs781687292 Chr.2: 61188959-61188959 on Build GRCh38
rs113660467 Chr.2: 61189119-61189119 on Build GRCh38
rs758608115 Chr.2: 61188944-61188944 on Build GRCh38
rs139164389 Chr.2: 61188848-61188848 on Build GRCh38
rs762757085 Chr.2: 61189102-61189102 on Build GRCh38
rs143137074 Chr.2: 61189023-61189023 on Build GRCh38
rs756474245 Chr.2: 61188881-61188882 on Build GRCh38
rs753602835 Chr.2: 61188703-61188703 on Build GRCh38
rs777309577 Chr.2: 61189153-61189153 on Build GRCh38
rs771689109 Chr.2: 61188989-61188989 on Build GRCh38
rs779711208 Chr.2: 61188694-61188694 on Build GRCh38
rs776860151 Chr.2: 61188866-61188866 on Build GRCh38
rs775436188 Chr.2: 61189095-61189095 on Build GRCh38
rs755606236 Chr.2: 61189087-61189087 on Build GRCh38
rs1034505218 Chr.2: 61188981-61188981 on Build GRCh38
rs914832563 Chr.2: 61188775-61188775 on Build GRCh38
rs1008292226 Chr.2: 61188702-61188702 on Build GRCh38
rs759664079 Chr.2: 61189081-61189082 on Build GRCh38
rs200471425 Chr.2: 61189015-61189015 on Build GRCh38
rs938760648 Chr.2: 61189158-61189158 on Build GRCh38
rs146305168 Chr.2: 61188841-61188841 on Build GRCh38
rs766260000 Chr.2: 61189013-61189013 on Build GRCh38
rs534624755 Chr.2: 61188765-61188766 on Build GRCh38
rs891217279 Chr.2: 61188692-61188692 on Build GRCh38
rs1041925599 Chr.2: 61188798-61188798 on Build GRCh38
rs772882532 Chr.2: 61189082-61189082 on Build GRCh38
rs748267814 Chr.2: 61188732-61188732 on Build GRCh38
rs577828703 Chr.2: 61189016-61189016 on Build GRCh38
rs367931795 Chr.2: 61188883-61188883 on Build GRCh38
rs980202965 Chr.2: 61189125-61189125 on Build GRCh38
rs779316354 Chr.2: 61188925-61188925 on Build GRCh38
rs754452993 Chr.2: 61189107-61189108 on Build GRCh38
rs780401510 Chr.2: 61189067-61189067 on Build GRCh38
rs768331160 Chr.2: 61188876-61188876 on Build GRCh38
rs1002663257 Chr.2: 61188966-61188966 on Build GRCh38
rs371427641 Chr.2: 61189024-61189024 on Build GRCh38
rs761365222 Chr.2: 61188882-61188882 on Build GRCh38
rs771646059 Chr.2: 61189073-61189073 on Build GRCh38
rs776766244 Chr.2: 61189086-61189086 on Build GRCh38
rs145063616 Chr.2: 61188921-61188921 on Build GRCh38

View Full Product Details