Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69421673-69422221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF23
Entrez Gene ID
9493
Gene Name
kinesin family member 23
Gene Aliases
CHO1, KNSL5, MKLP-1, MKLP1
UniGene
Hs.270845
Gene Chromosome Location
Chr.15: 69414246-69448427 on Build GRCh38
Transcripts
NM_001281301.1 NM_004856.6 NM_138555.3 XM_005254796.2 XM_005254797.2 XM_005254798.2 XM_005254799.2 XM_011522238.1 XM_011522239.2 XM_017022733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1255825 c.305A>C p.H102P Chr.15: 69421741-69421741 on Build GRCh38
4779271 c.431G>C p.G144A Chr.15: 69422106-69422106 on Build GRCh38
4696052 c.341C>T p.T114M Chr.15: 69422016-69422016 on Build GRCh38
4484692 c.283C>T p.P95S Chr.15: 69421719-69421719 on Build GRCh38
4950774 c.316+2_316+3delTA p.? Chr.15: 69421754-69421755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775864608 Chr.15: 69421722-69421722 on Build GRCh38
rs774076969 Chr.15: 69422051-69422051 on Build GRCh38
rs751196574 Chr.15: 69421693-69421693 on Build GRCh38
rs780887490 Chr.15: 69421791-69421791 on Build GRCh38
rs772638653 Chr.15: 69421958-69421958 on Build GRCh38
rs766423729 Chr.15: 69422005-69422005 on Build GRCh38
rs766340385 Chr.15: 69422049-69422049 on Build GRCh38
rs771624403 Chr.15: 69421707-69421707 on Build GRCh38
rs573840379 Chr.15: 69422179-69422179 on Build GRCh38
rs747385783 Chr.15: 69421971-69421971 on Build GRCh38
rs549286242 Chr.15: 69421732-69421732 on Build GRCh38
rs760809801 Chr.15: 69421724-69421724 on Build GRCh38
rs199854965 Chr.15: 69421988-69421988 on Build GRCh38
rs183300516 Chr.15: 69421887-69421887 on Build GRCh38
rs773221237 Chr.15: 69422036-69422036 on Build GRCh38
rs914396912 Chr.15: 69421723-69421723 on Build GRCh38
rs61751120 Chr.15: 69421729-69421729 on Build GRCh38
rs878889642 Chr.15: 69421827-69421827 on Build GRCh38
rs896751320 Chr.15: 69421924-69421924 on Build GRCh38
rs761980281 Chr.15: 69422007-69422007 on Build GRCh38
rs753393722 Chr.15: 69422100-69422100 on Build GRCh38
rs758885320 Chr.15: 69422106-69422106 on Build GRCh38
rs780304497 Chr.15: 69421962-69421963 on Build GRCh38
rs761855392 Chr.15: 69421734-69421734 on Build GRCh38
rs34969957 Chr.15: 69422008-69422008 on Build GRCh38
rs1056759155 Chr.15: 69421814-69421814 on Build GRCh38
rs754657140 Chr.15: 69421778-69421778 on Build GRCh38
rs370109100 Chr.15: 69421733-69421733 on Build GRCh38
rs948904102 Chr.15: 69422163-69422163 on Build GRCh38
rs755780903 Chr.15: 69422090-69422090 on Build GRCh38
rs61752336 Chr.15: 69421749-69421749 on Build GRCh38
rs372849416 Chr.15: 69422076-69422076 on Build GRCh38
rs768824023 Chr.15: 69422082-69422082 on Build GRCh38
rs374913132 Chr.15: 69422183-69422183 on Build GRCh38
rs747369638 Chr.15: 69422146-69422146 on Build GRCh38
rs997409755 Chr.15: 69421781-69421781 on Build GRCh38
rs775747735 Chr.15: 69421702-69421702 on Build GRCh38
rs759354494 Chr.15: 69422054-69422054 on Build GRCh38
rs755397176 Chr.15: 69422151-69422151 on Build GRCh38
rs888957986 Chr.15: 69421799-69421799 on Build GRCh38
rs1052766722 Chr.15: 69421741-69421741 on Build GRCh38
rs751304465 Chr.15: 69421776-69421776 on Build GRCh38
rs752304859 Chr.15: 69421798-69421798 on Build GRCh38
rs750388701 Chr.15: 69421756-69421756 on Build GRCh38
rs754874229 Chr.15: 69422052-69422052 on Build GRCh38
rs59689652 Chr.15: 69421866-69421866 on Build GRCh38
rs755655896 Chr.15: 69421802-69421802 on Build GRCh38
rs572081732 Chr.15: 69421844-69421844 on Build GRCh38
rs374755675 Chr.15: 69421770-69421770 on Build GRCh38
rs753971521 Chr.15: 69421913-69421913 on Build GRCh38
rs772597901 Chr.15: 69421719-69421719 on Build GRCh38
rs557894020 Chr.15: 69421956-69421956 on Build GRCh38
rs763699866 Chr.15: 69422099-69422099 on Build GRCh38
rs200683749 Chr.15: 69422039-69422039 on Build GRCh38
rs1041485717 Chr.15: 69421736-69421736 on Build GRCh38
rs544467574 Chr.15: 69422017-69422017 on Build GRCh38
rs137922672 Chr.15: 69421710-69421710 on Build GRCh38
rs777380027 Chr.15: 69421944-69421944 on Build GRCh38
rs747693155 Chr.15: 69421705-69421705 on Build GRCh38
rs375858716 Chr.15: 69422155-69422155 on Build GRCh38
rs767117625 Chr.15: 69422060-69422060 on Build GRCh38
rs754603079 Chr.15: 69421697-69421697 on Build GRCh38
rs189246681 Chr.15: 69422140-69422140 on Build GRCh38
rs1019582196 Chr.15: 69422193-69422193 on Build GRCh38
rs758450286 Chr.15: 69421941-69421942 on Build GRCh38
rs780349117 Chr.15: 69421964-69421964 on Build GRCh38
rs762825884 Chr.15: 69421767-69421767 on Build GRCh38

View Full Product Details