Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30092087-30092611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL3
Entrez Gene ID
83719
Gene Name
yippee like 3
Gene Aliases
-
UniGene
Hs.513491
Gene Chromosome Location
Chr.16: 30092314-30096216 on Build GRCh38
Transcripts
NM_001145524.1 NM_031477.4

SNP Detail

SNP ID Chromosome Location
rs139287306 Chr.16: 30092551-30092551 on Build GRCh38
rs967758517 Chr.16: 30092372-30092372 on Build GRCh38
rs1007712573 Chr.16: 30092139-30092139 on Build GRCh38
rs762560791 Chr.16: 30092579-30092579 on Build GRCh38
rs1036170663 Chr.16: 30092312-30092312 on Build GRCh38
rs766215953 Chr.16: 30092581-30092581 on Build GRCh38
rs914960639 Chr.16: 30092111-30092111 on Build GRCh38
rs530807504 Chr.16: 30092321-30092321 on Build GRCh38
rs1044572 Chr.16: 30092440-30092440 on Build GRCh38
rs764752504 Chr.16: 30092139-30092139 on Build GRCh38
rs780617234 Chr.16: 30092163-30092163 on Build GRCh38
rs543476786 Chr.16: 30092122-30092122 on Build GRCh38
rs180950054 Chr.16: 30092225-30092225 on Build GRCh38
rs923599704 Chr.16: 30092451-30092452 on Build GRCh38
rs1044634 Chr.16: 30092353-30092353 on Build GRCh38
rs549009773 Chr.16: 30092338-30092338 on Build GRCh38
rs375624212 Chr.16: 30092223-30092223 on Build GRCh38
rs571831865 Chr.16: 30092546-30092546 on Build GRCh38
rs904243638 Chr.16: 30092489-30092489 on Build GRCh38
rs557817502 Chr.16: 30092560-30092560 on Build GRCh38
rs990252048 Chr.16: 30092362-30092362 on Build GRCh38
rs373954330 Chr.16: 30092523-30092523 on Build GRCh38
rs941848109 Chr.16: 30092310-30092310 on Build GRCh38
rs938992137 Chr.16: 30092451-30092451 on Build GRCh38
rs567570907 Chr.16: 30092339-30092339 on Build GRCh38
rs909323704 Chr.16: 30092290-30092290 on Build GRCh38
rs992372489 Chr.16: 30092246-30092246 on Build GRCh38
rs915015884 Chr.16: 30092368-30092369 on Build GRCh38
rs1012054180 Chr.16: 30092230-30092230 on Build GRCh38
rs956877602 Chr.16: 30092552-30092552 on Build GRCh38
rs1044584 Chr.16: 30092414-30092414 on Build GRCh38
rs916472678 Chr.16: 30092458-30092458 on Build GRCh38
rs370215390 Chr.16: 30092280-30092280 on Build GRCh38
rs1011729028 Chr.16: 30092589-30092589 on Build GRCh38
rs561960539 Chr.16: 30092132-30092132 on Build GRCh38
rs559516347 Chr.16: 30092261-30092261 on Build GRCh38
rs939257005 Chr.16: 30092211-30092211 on Build GRCh38
rs888779294 Chr.16: 30092454-30092454 on Build GRCh38
rs1007249586 Chr.16: 30092456-30092456 on Build GRCh38
rs1004912619 Chr.16: 30092273-30092273 on Build GRCh38
rs1029212045 Chr.16: 30092316-30092316 on Build GRCh38
rs535333355 Chr.16: 30092386-30092386 on Build GRCh38
rs769001937 Chr.16: 30092359-30092359 on Build GRCh38
rs548216647 Chr.16: 30092585-30092585 on Build GRCh38
rs563478907 Chr.16: 30092283-30092283 on Build GRCh38
rs1021411758 Chr.16: 30092145-30092145 on Build GRCh38
rs576245433 Chr.16: 30092567-30092567 on Build GRCh38
rs1051715201 Chr.16: 30092436-30092436 on Build GRCh38
rs1021778873 Chr.16: 30092474-30092474 on Build GRCh38
rs902456282 Chr.16: 30092172-30092172 on Build GRCh38
rs953558429 Chr.16: 30092337-30092337 on Build GRCh38
rs956930017 Chr.16: 30092237-30092237 on Build GRCh38
rs535836986 Chr.16: 30092215-30092215 on Build GRCh38
rs933188559 Chr.16: 30092407-30092407 on Build GRCh38
rs551833779 Chr.16: 30092282-30092282 on Build GRCh38
rs1004965002 Chr.16: 30092562-30092562 on Build GRCh38
rs1029961457 Chr.16: 30092125-30092125 on Build GRCh38
rs533242264 Chr.16: 30092268-30092268 on Build GRCh38
rs887838807 Chr.16: 30092257-30092257 on Build GRCh38

View Full Product Details