Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 129073392-129073962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132D
Entrez Gene ID
121256
Gene Name
transmembrane protein 132D
Gene Aliases
MOLT, PPP1R153
UniGene
Hs.507268
Gene Chromosome Location
Chr.12: 129071726-129903667 on Build GRCh38
Transcripts
NM_133448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3811458 c.1873G>A p.E625K Chr.12: 129073916-129073916 on Build GRCh38
3457992 c.1828G>A p.D610N Chr.12: 129073961-129073961 on Build GRCh38
3457990 c.1849G>A p.D617N Chr.12: 129073940-129073940 on Build GRCh38
6803299 c.3251A>T p.D1084V Chr.12: 129073924-129073924 on Build GRCh38
6310925 c.1858C>A p.P620T Chr.12: 129073931-129073931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773957559 Chr.12: 129073862-129073862 on Build GRCh38
rs759382977 Chr.12: 129073901-129073901 on Build GRCh38
rs768481651 Chr.12: 129073698-129073698 on Build GRCh38
rs544459164 Chr.12: 129073753-129073753 on Build GRCh38
rs756953292 Chr.12: 129073503-129073503 on Build GRCh38
rs376010677 Chr.12: 129073449-129073449 on Build GRCh38
rs530079204 Chr.12: 129073808-129073808 on Build GRCh38
rs150644035 Chr.12: 129073578-129073579 on Build GRCh38
rs1021116259 Chr.12: 129073934-129073934 on Build GRCh38
rs1034253297 Chr.12: 129073429-129073429 on Build GRCh38
rs916090596 Chr.12: 129073699-129073699 on Build GRCh38
rs985783591 Chr.12: 129073729-129073729 on Build GRCh38
rs571589123 Chr.12: 129073550-129073550 on Build GRCh38
rs909765418 Chr.12: 129073732-129073732 on Build GRCh38
rs536064040 Chr.12: 129073799-129073800 on Build GRCh38
rs751593339 Chr.12: 129073921-129073921 on Build GRCh38
rs774206202 Chr.12: 129073799-129073799 on Build GRCh38
rs905525757 Chr.12: 129073801-129073801 on Build GRCh38
rs545290833 Chr.12: 129073722-129073722 on Build GRCh38
rs747461491 Chr.12: 129073852-129073852 on Build GRCh38
rs765164006 Chr.12: 129073906-129073906 on Build GRCh38
rs980984791 Chr.12: 129073428-129073428 on Build GRCh38
rs764238814 Chr.12: 129073920-129073920 on Build GRCh38
rs532610657 Chr.12: 129073929-129073929 on Build GRCh38
rs566948994 Chr.12: 129073460-129073460 on Build GRCh38
rs142279858 Chr.12: 129073449-129073450 on Build GRCh38
rs1034339269 Chr.12: 129073450-129073462 on Build GRCh38
rs913949925 Chr.12: 129073439-129073439 on Build GRCh38
rs373019991 Chr.12: 129073448-129073448 on Build GRCh38
rs1001170804 Chr.12: 129073804-129073804 on Build GRCh38
rs368838475 Chr.12: 129073447-129073447 on Build GRCh38
rs1007331141 Chr.12: 129073784-129073784 on Build GRCh38
rs747860987 Chr.12: 129073871-129073871 on Build GRCh38
rs758753338 Chr.12: 129073917-129073917 on Build GRCh38
rs1011042559 Chr.12: 129073602-129073602 on Build GRCh38
rs552827155 Chr.12: 129073706-129073706 on Build GRCh38
rs918356810 Chr.12: 129073797-129073797 on Build GRCh38
rs142143418 Chr.12: 129073463-129073464 on Build GRCh38
rs551304457 Chr.12: 129073930-129073930 on Build GRCh38
rs1045409333 Chr.12: 129073800-129073800 on Build GRCh38
rs1053237518 Chr.12: 129073559-129073559 on Build GRCh38
rs985970101 Chr.12: 129073442-129073442 on Build GRCh38
rs372535592 Chr.12: 129073601-129073601 on Build GRCh38
rs1049791403 Chr.12: 129073727-129073727 on Build GRCh38
rs146776647 Chr.12: 129073437-129073437 on Build GRCh38
rs888274270 Chr.12: 129073541-129073541 on Build GRCh38
rs777826021 Chr.12: 129073720-129073720 on Build GRCh38
rs1026403237 Chr.12: 129073805-129073805 on Build GRCh38
rs965765978 Chr.12: 129073597-129073597 on Build GRCh38
rs916059345 Chr.12: 129073488-129073488 on Build GRCh38
rs753211291 Chr.12: 129073436-129073436 on Build GRCh38
rs139764768 Chr.12: 129073506-129073506 on Build GRCh38
rs200525995 Chr.12: 129073854-129073854 on Build GRCh38
rs1013891799 Chr.12: 129073572-129073572 on Build GRCh38
rs1025951129 Chr.12: 129073659-129073659 on Build GRCh38
rs770189816 Chr.12: 129073860-129073860 on Build GRCh38
rs376759792 Chr.12: 129073446-129073446 on Build GRCh38
rs752965079 Chr.12: 129073909-129073909 on Build GRCh38
rs374661593 Chr.12: 129073647-129073647 on Build GRCh38
rs906816016 Chr.12: 129073585-129073585 on Build GRCh38
rs186940947 Chr.12: 129073578-129073578 on Build GRCh38
rs749190657 Chr.12: 129073692-129073692 on Build GRCh38
rs533653423 Chr.12: 129073591-129073591 on Build GRCh38
rs550251624 Chr.12: 129073721-129073721 on Build GRCh38
rs761992274 Chr.12: 129073701-129073701 on Build GRCh38
rs770149931 Chr.12: 129073874-129073874 on Build GRCh38
rs199659256 Chr.12: 129073475-129073476 on Build GRCh38
rs758523299 Chr.12: 129073832-129073832 on Build GRCh38
rs570219941 Chr.12: 129073764-129073764 on Build GRCh38
rs894497938 Chr.12: 129073471-129073472 on Build GRCh38
rs777977120 Chr.12: 129073851-129073851 on Build GRCh38
rs781558381 Chr.12: 129073855-129073855 on Build GRCh38
rs544956906 Chr.12: 129073620-129073620 on Build GRCh38
rs893221919 Chr.12: 129073563-129073564 on Build GRCh38
rs932542970 Chr.12: 129073507-129073507 on Build GRCh38
rs766299752 Chr.12: 129073882-129073882 on Build GRCh38
rs558691041 Chr.12: 129073723-129073723 on Build GRCh38
rs575539866 Chr.12: 129073726-129073726 on Build GRCh38
rs760105459 Chr.12: 129073879-129073879 on Build GRCh38
rs984970708 Chr.12: 129073485-129073486 on Build GRCh38
rs145404523 Chr.12: 129073564-129073564 on Build GRCh38
rs776879974 Chr.12: 129073724-129073724 on Build GRCh38
rs776455207 Chr.12: 129073888-129073888 on Build GRCh38
rs896879643 Chr.12: 129073566-129073566 on Build GRCh38
rs779733905 Chr.12: 129073611-129073611 on Build GRCh38
rs746194139 Chr.12: 129073856-129073856 on Build GRCh38
rs561245845 Chr.12: 129073757-129073757 on Build GRCh38
rs759711088 Chr.12: 129073781-129073781 on Build GRCh38
rs1049615690 Chr.12: 129073511-129073511 on Build GRCh38
rs951905938 Chr.12: 129073675-129073675 on Build GRCh38
rs984501696 Chr.12: 129073688-129073688 on Build GRCh38
rs572870029 Chr.12: 129073707-129073707 on Build GRCh38
rs765505864 Chr.12: 129073825-129073825 on Build GRCh38
rs907239759 Chr.12: 129073523-129073523 on Build GRCh38
rs144460624 Chr.12: 129073529-129073529 on Build GRCh38
rs527388696 Chr.12: 129073868-129073868 on Build GRCh38
rs981713324 Chr.12: 129073702-129073702 on Build GRCh38
rs191406150 Chr.12: 129073831-129073831 on Build GRCh38
rs555850748 Chr.12: 129073878-129073878 on Build GRCh38
rs777508987 Chr.12: 129073861-129073862 on Build GRCh38
rs1033893195 Chr.12: 129073592-129073592 on Build GRCh38

View Full Product Details