Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 130354409-130354969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIWIL1
Entrez Gene ID
9271
Gene Name
piwi like RNA-mediated gene silencing 1
Gene Aliases
CT80.1, HIWI, MIWI, PIWI
UniGene
Hs.405659
Gene Chromosome Location
Chr.12: 130337887-130412708 on Build GRCh38
Transcripts
NM_001190971.1 NM_004764.4 XM_011539002.2 XM_011539003.2 XM_011539004.2 XM_011539005.1 XM_011539006.2 XM_017020229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1605778 c.1135C>A p.P379T Chr.12: 130354627-130354627 on Build GRCh38
692291 c.1054C>A p.Q352K Chr.12: 130354546-130354546 on Build GRCh38
6746958 c.1208T>C p.M403T Chr.12: 130354924-130354924 on Build GRCh38
1739901 c.1088T>G p.V363G Chr.12: 130354580-130354580 on Build GRCh38
3792354 c.1139C>G p.A380G Chr.12: 130354631-130354631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747762614 Chr.12: 130354892-130354892 on Build GRCh38
rs1014080344 Chr.12: 130354843-130354843 on Build GRCh38
rs766692063 Chr.12: 130354590-130354590 on Build GRCh38
rs748671026 Chr.12: 130354515-130354515 on Build GRCh38
rs766303134 Chr.12: 130354695-130354696 on Build GRCh38
rs767366713 Chr.12: 130354497-130354497 on Build GRCh38
rs768241314 Chr.12: 130354668-130354668 on Build GRCh38
rs928677048 Chr.12: 130354782-130354782 on Build GRCh38
rs769540084 Chr.12: 130354521-130354521 on Build GRCh38
rs771202187 Chr.12: 130354625-130354625 on Build GRCh38
rs1013549299 Chr.12: 130354523-130354523 on Build GRCh38
rs778554091 Chr.12: 130354873-130354873 on Build GRCh38
rs957182133 Chr.12: 130354679-130354679 on Build GRCh38
rs756400104 Chr.12: 130354501-130354501 on Build GRCh38
rs945703690 Chr.12: 130354721-130354721 on Build GRCh38
rs771488646 Chr.12: 130354905-130354905 on Build GRCh38
rs576896988 Chr.12: 130354519-130354520 on Build GRCh38
rs1049673643 Chr.12: 130354535-130354535 on Build GRCh38
rs778273394 Chr.12: 130354711-130354711 on Build GRCh38
rs544030644 Chr.12: 130354487-130354487 on Build GRCh38
rs905175422 Chr.12: 130354863-130354863 on Build GRCh38
rs201349482 Chr.12: 130354494-130354494 on Build GRCh38
rs768421961 Chr.12: 130354444-130354444 on Build GRCh38
rs10848087 Chr.12: 130354620-130354620 on Build GRCh38
rs372705871 Chr.12: 130354864-130354864 on Build GRCh38
rs4759660 Chr.12: 130354506-130354506 on Build GRCh38
rs755473652 Chr.12: 130354510-130354510 on Build GRCh38
rs776454826 Chr.12: 130354922-130354922 on Build GRCh38
rs774380989 Chr.12: 130354539-130354539 on Build GRCh38
rs368357352 Chr.12: 130354698-130354698 on Build GRCh38
rs374570688 Chr.12: 130354498-130354498 on Build GRCh38
rs781712952 Chr.12: 130354853-130354853 on Build GRCh38
rs35377460 Chr.12: 130354746-130354747 on Build GRCh38
rs761820280 Chr.12: 130354680-130354680 on Build GRCh38
rs760453817 Chr.12: 130354687-130354687 on Build GRCh38
rs146983606 Chr.12: 130354761-130354761 on Build GRCh38
rs757589987 Chr.12: 130354841-130354841 on Build GRCh38
rs952223496 Chr.12: 130354737-130354737 on Build GRCh38
rs914389072 Chr.12: 130354854-130354854 on Build GRCh38
rs551381051 Chr.12: 130354700-130354700 on Build GRCh38
rs762136772 Chr.12: 130354738-130354738 on Build GRCh38
rs754014143 Chr.12: 130354692-130354692 on Build GRCh38
rs772381837 Chr.12: 130354518-130354518 on Build GRCh38
rs752827820 Chr.12: 130354701-130354701 on Build GRCh38
rs956307815 Chr.12: 130354804-130354804 on Build GRCh38
rs778990087 Chr.12: 130354604-130354604 on Build GRCh38
rs748970480 Chr.12: 130354666-130354666 on Build GRCh38
rs371671008 Chr.12: 130354875-130354875 on Build GRCh38
rs369600464 Chr.12: 130354601-130354601 on Build GRCh38
rs909413266 Chr.12: 130354785-130354785 on Build GRCh38
rs375538757 Chr.12: 130354524-130354524 on Build GRCh38
rs375915792 Chr.12: 130354943-130354943 on Build GRCh38
rs762482687 Chr.12: 130354529-130354529 on Build GRCh38
rs758823682 Chr.12: 130354710-130354710 on Build GRCh38
rs200413984 Chr.12: 130354600-130354600 on Build GRCh38
rs777982419 Chr.12: 130354619-130354619 on Build GRCh38
rs1041042351 Chr.12: 130354455-130354455 on Build GRCh38
rs775148722 Chr.12: 130354638-130354638 on Build GRCh38
rs748618197 Chr.12: 130354606-130354606 on Build GRCh38
rs571601131 Chr.12: 130354722-130354722 on Build GRCh38
rs756278220 Chr.12: 130354583-130354583 on Build GRCh38
rs778109943 Chr.12: 130354920-130354920 on Build GRCh38
rs902053597 Chr.12: 130354462-130354462 on Build GRCh38
rs544937707 Chr.12: 130354850-130354850 on Build GRCh38
rs752017518 Chr.12: 130354712-130354712 on Build GRCh38
rs758988547 Chr.12: 130354614-130354614 on Build GRCh38
rs529941222 Chr.12: 130354557-130354557 on Build GRCh38
rs922921274 Chr.12: 130354760-130354760 on Build GRCh38
rs185995248 Chr.12: 130354937-130354937 on Build GRCh38
rs779329497 Chr.12: 130354513-130354513 on Build GRCh38
rs1050618100 Chr.12: 130354762-130354762 on Build GRCh38
rs878933165 Chr.12: 130354819-130354819 on Build GRCh38
rs368086386 Chr.12: 130354919-130354919 on Build GRCh38
rs942179934 Chr.12: 130354805-130354805 on Build GRCh38
rs773146996 Chr.12: 130354908-130354908 on Build GRCh38
rs750281432 Chr.12: 130354573-130354573 on Build GRCh38
rs199577850 Chr.12: 130354681-130354681 on Build GRCh38
rs970085065 Chr.12: 130354468-130354468 on Build GRCh38
rs773702953 Chr.12: 130354474-130354474 on Build GRCh38
rs748917485 Chr.12: 130354861-130354861 on Build GRCh38
rs761873436 Chr.12: 130354556-130354556 on Build GRCh38
rs774045226 Chr.12: 130354673-130354673 on Build GRCh38
rs762798932 Chr.12: 130354545-130354545 on Build GRCh38
rs772879502 Chr.12: 130354686-130354686 on Build GRCh38
rs200918174 Chr.12: 130354703-130354703 on Build GRCh38
rs2173082 Chr.12: 130354442-130354442 on Build GRCh38
rs1036407771 Chr.12: 130354809-130354809 on Build GRCh38
rs550883502 Chr.12: 130354821-130354821 on Build GRCh38
rs932148154 Chr.12: 130354470-130354470 on Build GRCh38
rs764322395 Chr.12: 130354517-130354517 on Build GRCh38
rs763709895 Chr.12: 130354531-130354531 on Build GRCh38
rs371846241 Chr.12: 130354505-130354505 on Build GRCh38
rs752917743 Chr.12: 130354938-130354938 on Build GRCh38

View Full Product Details